Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165311743-165312263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN2A
Entrez Gene ID
6326
Gene Name
sodium voltage-gated channel alpha subunit 2
Gene Aliases
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2
UniGene
Hs.93485
Gene Chromosome Location
Chr.2: 165208056-165392310 on Build GRCh38
Transcripts
NM_001040142.1 NM_001040143.1 NM_021007.2 XM_005246753.3 XM_005246755.4 XM_017004655.1 XM_017004656.1 XM_017004657.1 XM_017004658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173070 c.988G>C p.E330Q Chr.2: 165312042-165312042 on Build GRCh38
4086194 c.1000G>A p.D334N Chr.2: 165312054-165312054 on Build GRCh38
6088081 c.991G>T p.G331W Chr.2: 165312045-165312045 on Build GRCh38
6088080 c.991G>T p.G331W Chr.2: 165312045-165312045 on Build GRCh38
7173071 c.988G>C p.E330Q Chr.2: 165312042-165312042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903420024 Chr.2: 165311973-165311973 on Build GRCh38
rs753672474 Chr.2: 165311985-165311985 on Build GRCh38
rs982574434 Chr.2: 165312228-165312228 on Build GRCh38
rs556487979 Chr.2: 165311818-165311818 on Build GRCh38
rs759187531 Chr.2: 165312115-165312115 on Build GRCh38
rs750968741 Chr.2: 165312014-165312014 on Build GRCh38
rs1020253622 Chr.2: 165311961-165311961 on Build GRCh38
rs771619853 Chr.2: 165312116-165312116 on Build GRCh38
rs1024442274 Chr.2: 165312144-165312144 on Build GRCh38
rs1026718663 Chr.2: 165312209-165312209 on Build GRCh38
rs919729029 Chr.2: 165311835-165311835 on Build GRCh38
rs186838716 Chr.2: 165312150-165312150 on Build GRCh38
rs751688495 Chr.2: 165312006-165312006 on Build GRCh38
rs778228350 Chr.2: 165312086-165312086 on Build GRCh38
rs771610189 Chr.2: 165312102-165312102 on Build GRCh38
rs974198115 Chr.2: 165312179-165312179 on Build GRCh38
rs143785800 Chr.2: 165311859-165311859 on Build GRCh38
rs547425175 Chr.2: 165312176-165312176 on Build GRCh38
rs748413349 Chr.2: 165312047-165312047 on Build GRCh38
rs182616186 Chr.2: 165311970-165311970 on Build GRCh38
rs571977829 Chr.2: 165311889-165311889 on Build GRCh38
rs951239918 Chr.2: 165312212-165312212 on Build GRCh38
rs1018317363 Chr.2: 165312151-165312151 on Build GRCh38
rs939395773 Chr.2: 165311864-165311864 on Build GRCh38
rs773982382 Chr.2: 165312107-165312107 on Build GRCh38
rs888933984 Chr.2: 165311819-165311819 on Build GRCh38
rs1056861694 Chr.2: 165311883-165311883 on Build GRCh38
rs1043918754 Chr.2: 165311965-165311965 on Build GRCh38
rs911007694 Chr.2: 165311769-165311769 on Build GRCh38
rs763965582 Chr.2: 165311983-165311983 on Build GRCh38
rs948098014 Chr.2: 165311932-165311932 on Build GRCh38
rs770003814 Chr.2: 165312050-165312050 on Build GRCh38
rs757218387 Chr.2: 165311986-165311986 on Build GRCh38
rs774950838 Chr.2: 165312134-165312134 on Build GRCh38
rs146355214 Chr.2: 165312158-165312158 on Build GRCh38
rs3214604 Chr.2: 165311782-165311784 on Build GRCh38
rs929905509 Chr.2: 165311837-165311837 on Build GRCh38
rs910507834 Chr.2: 165312238-165312238 on Build GRCh38
rs189591560 Chr.2: 165311890-165311890 on Build GRCh38
rs1033303305 Chr.2: 165312059-165312059 on Build GRCh38
rs557410328 Chr.2: 165311894-165311894 on Build GRCh38
rs971700697 Chr.2: 165312172-165312172 on Build GRCh38
rs577252162 Chr.2: 165311943-165311943 on Build GRCh38
rs1007900162 Chr.2: 165312122-165312122 on Build GRCh38
rs998106381 Chr.2: 165311931-165311931 on Build GRCh38
rs943250239 Chr.2: 165311854-165311854 on Build GRCh38
rs964099506 Chr.2: 165311796-165311796 on Build GRCh38
rs749778303 Chr.2: 165312098-165312098 on Build GRCh38
rs766101104 Chr.2: 165311997-165312000 on Build GRCh38
rs755180885 Chr.2: 165312032-165312032 on Build GRCh38
rs1046994948 Chr.2: 165311839-165311839 on Build GRCh38
rs903994289 Chr.2: 165311969-165311969 on Build GRCh38
rs781204054 Chr.2: 165312036-165312036 on Build GRCh38
rs760400769 Chr.2: 165312137-165312137 on Build GRCh38
rs796053175 Chr.2: 165312081-165312081 on Build GRCh38
rs752555632 Chr.2: 165311952-165311952 on Build GRCh38
rs2304016 Chr.2: 165311993-165311993 on Build GRCh38
rs149611312 Chr.2: 165311781-165311783 on Build GRCh38
rs959126625 Chr.2: 165312126-165312126 on Build GRCh38

View Full Product Details