Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169806860-169807274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSB
Entrez Gene ID
6741
Gene Name
Sjogren syndrome antigen B
Gene Aliases
LARP3, La, La/SSB
UniGene
Hs.632535
Gene Chromosome Location
Chr.2: 169798812-169812064 on Build GRCh38
Transcripts
NM_001294145.1 NM_003142.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009369 c.529G>T p.E177* Chr.2: 169807046-169807046 on Build GRCh38
6436291 c.538C>A p.L180M Chr.2: 169807055-169807055 on Build GRCh38
6291904 c.498A>G p.K166K Chr.2: 169807015-169807015 on Build GRCh38
1009368 c.453+2T>C p.? Chr.2: 169806894-169806894 on Build GRCh38
6790684 c.454-9delT p.? Chr.2: 169806962-169806962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894726884 Chr.2: 169807165-169807165 on Build GRCh38
rs751104851 Chr.2: 169806950-169806950 on Build GRCh38
rs557175075 Chr.2: 169807155-169807155 on Build GRCh38
rs886989126 Chr.2: 169807225-169807225 on Build GRCh38
rs1021757058 Chr.2: 169806962-169806962 on Build GRCh38
rs1032452156 Chr.2: 169806968-169806968 on Build GRCh38
rs750270859 Chr.2: 169806905-169806906 on Build GRCh38
rs372919350 Chr.2: 169807136-169807136 on Build GRCh38
rs968801676 Chr.2: 169807208-169807208 on Build GRCh38
rs1051448 Chr.2: 169806893-169806893 on Build GRCh38
rs762784654 Chr.2: 169806938-169806939 on Build GRCh38
rs991642774 Chr.2: 169806937-169806937 on Build GRCh38
rs987815986 Chr.2: 169807099-169807099 on Build GRCh38
rs552982135 Chr.2: 169807186-169807186 on Build GRCh38
rs373415341 Chr.2: 169806916-169806916 on Build GRCh38
rs186344908 Chr.2: 169807028-169807028 on Build GRCh38
rs138612495 Chr.2: 169807219-169807219 on Build GRCh38
rs910453439 Chr.2: 169807102-169807102 on Build GRCh38
rs1047405070 Chr.2: 169807144-169807144 on Build GRCh38
rs760452732 Chr.2: 169807013-169807013 on Build GRCh38
rs770257590 Chr.2: 169807128-169807128 on Build GRCh38
rs765542277 Chr.2: 169807021-169807021 on Build GRCh38
rs766140838 Chr.2: 169806940-169806941 on Build GRCh38
rs942421060 Chr.2: 169807137-169807137 on Build GRCh38
rs369947209 Chr.2: 169807049-169807049 on Build GRCh38
rs377554846 Chr.2: 169806925-169806925 on Build GRCh38
rs181856742 Chr.2: 169807024-169807024 on Build GRCh38
rs771917892 Chr.2: 169806941-169806941 on Build GRCh38
rs1021766760 Chr.2: 169807206-169807206 on Build GRCh38
rs756488413 Chr.2: 169807069-169807069 on Build GRCh38
rs1031735449 Chr.2: 169807241-169807241 on Build GRCh38
rs144350886 Chr.2: 169807036-169807036 on Build GRCh38
rs12467132 Chr.2: 169807008-169807008 on Build GRCh38
rs373048377 Chr.2: 169807055-169807055 on Build GRCh38
rs954901032 Chr.2: 169807242-169807242 on Build GRCh38
rs574812366 Chr.2: 169807112-169807112 on Build GRCh38
rs753095047 Chr.2: 169807062-169807062 on Build GRCh38
rs772044776 Chr.2: 169807118-169807118 on Build GRCh38
rs914195389 Chr.2: 169807108-169807108 on Build GRCh38
rs781552255 Chr.2: 169807057-169807057 on Build GRCh38
rs759469414 Chr.2: 169806930-169806930 on Build GRCh38
rs977883230 Chr.2: 169807222-169807222 on Build GRCh38
rs770592828 Chr.2: 169807088-169807088 on Build GRCh38
rs537210708 Chr.2: 169807154-169807154 on Build GRCh38
rs369277813 Chr.2: 169806906-169806906 on Build GRCh38
rs777462195 Chr.2: 169806901-169806901 on Build GRCh38
rs906169294 Chr.2: 169807150-169807150 on Build GRCh38
rs775290380 Chr.2: 169807006-169807006 on Build GRCh38
rs750722452 Chr.2: 169807027-169807027 on Build GRCh38
rs769419286 Chr.2: 169806888-169806888 on Build GRCh38
rs576924111 Chr.2: 169807164-169807164 on Build GRCh38
rs777376871 Chr.2: 169807078-169807078 on Build GRCh38
rs754582732 Chr.2: 169807094-169807094 on Build GRCh38
rs748777515 Chr.2: 169806908-169806908 on Build GRCh38
rs748904183 Chr.2: 169807085-169807085 on Build GRCh38
rs778337894 Chr.2: 169807097-169807097 on Build GRCh38
rs751610012 Chr.2: 169807054-169807054 on Build GRCh38

View Full Product Details