Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173906988-173907493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP3
Entrez Gene ID
6670
Gene Name
Sp3 transcription factor
Gene Aliases
SPR2
UniGene
Hs.531587
Gene Chromosome Location
Chr.2: 173906459-173965702 on Build GRCh38
Transcripts
NM_001017371.4 NM_001172712.1 NM_003111.4

SNP Detail

SNP ID Chromosome Location
rs1025188907 Chr.2: 173907225-173907225 on Build GRCh38
rs983899090 Chr.2: 173907118-173907118 on Build GRCh38
rs752341813 Chr.2: 173907012-173907012 on Build GRCh38
rs1048113602 Chr.2: 173907448-173907449 on Build GRCh38
rs999520041 Chr.2: 173907245-173907245 on Build GRCh38
rs936152251 Chr.2: 173907045-173907045 on Build GRCh38
rs958112493 Chr.2: 173907079-173907079 on Build GRCh38
rs923987783 Chr.2: 173907073-173907073 on Build GRCh38
rs958143653 Chr.2: 173907262-173907262 on Build GRCh38
rs1012480416 Chr.2: 173907062-173907062 on Build GRCh38
rs895310103 Chr.2: 173907454-173907454 on Build GRCh38
rs570364697 Chr.2: 173907070-173907070 on Build GRCh38
rs1030952794 Chr.2: 173907248-173907248 on Build GRCh38
rs895290690 Chr.2: 173907204-173907204 on Build GRCh38
rs1054601245 Chr.2: 173907355-173907355 on Build GRCh38
rs7589313 Chr.2: 173907183-173907183 on Build GRCh38
rs115642843 Chr.2: 173907419-173907419 on Build GRCh38
rs912521641 Chr.2: 173907368-173907370 on Build GRCh38
rs778688069 Chr.2: 173907182-173907182 on Build GRCh38
rs893574048 Chr.2: 173907375-173907375 on Build GRCh38
rs151009102 Chr.2: 173907207-173907207 on Build GRCh38
rs748326042 Chr.2: 173907331-173907331 on Build GRCh38
rs755317014 Chr.2: 173907296-173907296 on Build GRCh38
rs534559261 Chr.2: 173907311-173907311 on Build GRCh38
rs868267054 Chr.2: 173907072-173907072 on Build GRCh38
rs750852641 Chr.2: 173907076-173907076 on Build GRCh38
rs943648465 Chr.2: 173907174-173907174 on Build GRCh38
rs112896232 Chr.2: 173907014-173907014 on Build GRCh38
rs749297023 Chr.2: 173907398-173907398 on Build GRCh38
rs575335069 Chr.2: 173907067-173907067 on Build GRCh38
rs190747420 Chr.2: 173907167-173907167 on Build GRCh38
rs758127683 Chr.2: 173907244-173907244 on Build GRCh38
rs1018333450 Chr.2: 173907071-173907071 on Build GRCh38
rs539358979 Chr.2: 173907084-173907084 on Build GRCh38
rs116765826 Chr.2: 173907379-173907379 on Build GRCh38
rs781550734 Chr.2: 173907309-173907309 on Build GRCh38
rs912223387 Chr.2: 173907168-173907168 on Build GRCh38
rs1036729600 Chr.2: 173907401-173907401 on Build GRCh38
rs970722431 Chr.2: 173907069-173907069 on Build GRCh38
rs989975206 Chr.2: 173907082-173907082 on Build GRCh38
rs551595921 Chr.2: 173907212-173907212 on Build GRCh38
rs375218338 Chr.2: 173907361-173907361 on Build GRCh38
rs924841385 Chr.2: 173907332-173907332 on Build GRCh38
rs914393600 Chr.2: 173907103-173907103 on Build GRCh38
rs567604880 Chr.2: 173907447-173907447 on Build GRCh38
rs749143175 Chr.2: 173907342-173907342 on Build GRCh38
rs1025505484 Chr.2: 173907112-173907112 on Build GRCh38
rs751831533 Chr.2: 173907260-173907260 on Build GRCh38
rs903387985 Chr.2: 173907030-173907030 on Build GRCh38
rs757272411 Chr.2: 173907132-173907132 on Build GRCh38
rs142822571 Chr.2: 173907114-173907114 on Build GRCh38
rs991510283 Chr.2: 173907184-173907184 on Build GRCh38
rs1053558261 Chr.2: 173907049-173907049 on Build GRCh38
rs186682808 Chr.2: 173907142-173907142 on Build GRCh38
rs7562969 Chr.2: 173907430-173907430 on Build GRCh38

View Full Product Details