Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174422710-174423222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRN3
Entrez Gene ID
79634
Gene Name
secernin 3
Gene Aliases
SES3
UniGene
Hs.470679
Gene Chromosome Location
Chr.2: 174395729-174430071 on Build GRCh38
Transcripts
NM_001193528.1 NM_024583.4 XM_005246853.2 XM_005246854.4 XM_005246855.2 XM_005246856.2 XM_005246857.2 XM_011511839.2 XM_017004909.1 XM_017004910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6281581 c.771A>G p.E257E Chr.2: 174422901-174422901 on Build GRCh38
1590905 c.769C>T p.R257* Chr.2: 174422920-174422920 on Build GRCh38
6270511 c.907G>T p.D303Y Chr.2: 174423037-174423037 on Build GRCh38
1590906 c.762A>G p.E254E Chr.2: 174422913-174422913 on Build GRCh38
7220744 c.896+3G>A p.? Chr.2: 174423050-174423050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373001341 Chr.2: 174422796-174422796 on Build GRCh38
rs1053825889 Chr.2: 174423152-174423152 on Build GRCh38
rs757603980 Chr.2: 174422869-174422869 on Build GRCh38
rs1038175461 Chr.2: 174422745-174422745 on Build GRCh38
rs137877902 Chr.2: 174422951-174422951 on Build GRCh38
rs187598711 Chr.2: 174423043-174423043 on Build GRCh38
rs919749724 Chr.2: 174423157-174423157 on Build GRCh38
rs754486074 Chr.2: 174423000-174423000 on Build GRCh38
rs999210021 Chr.2: 174423131-174423131 on Build GRCh38
rs983829134 Chr.2: 174423085-174423085 on Build GRCh38
rs777076118 Chr.2: 174422966-174422966 on Build GRCh38
rs371689574 Chr.2: 174422843-174422843 on Build GRCh38
rs757014041 Chr.2: 174422920-174422920 on Build GRCh38
rs569449324 Chr.2: 174423008-174423008 on Build GRCh38
rs1032107408 Chr.2: 174422852-174422852 on Build GRCh38
rs937421425 Chr.2: 174423034-174423034 on Build GRCh38
rs1014274666 Chr.2: 174422886-174422886 on Build GRCh38
rs1046125247 Chr.2: 174422806-174422806 on Build GRCh38
rs771087793 Chr.2: 174422841-174422841 on Build GRCh38
rs1055331006 Chr.2: 174422780-174422780 on Build GRCh38
rs111549211 Chr.2: 174423108-174423108 on Build GRCh38
rs769183816 Chr.2: 174422956-174422956 on Build GRCh38
rs372009590 Chr.2: 174422924-174422924 on Build GRCh38
rs764820727 Chr.2: 174422889-174422889 on Build GRCh38
rs762097918 Chr.2: 174422978-174422978 on Build GRCh38
rs200861361 Chr.2: 174423066-174423066 on Build GRCh38
rs1054828881 Chr.2: 174422845-174422845 on Build GRCh38
rs746098699 Chr.2: 174422938-174422938 on Build GRCh38
rs973940996 Chr.2: 174423142-174423142 on Build GRCh38
rs1032471675 Chr.2: 174423021-174423021 on Build GRCh38
rs375309399 Chr.2: 174422987-174422987 on Build GRCh38
rs918610387 Chr.2: 174422948-174422948 on Build GRCh38
rs779004549 Chr.2: 174422929-174422929 on Build GRCh38
rs34170959 Chr.2: 174422999-174422999 on Build GRCh38
rs536481088 Chr.2: 174423026-174423026 on Build GRCh38
rs147537313 Chr.2: 174422917-174422917 on Build GRCh38
rs777009399 Chr.2: 174423090-174423090 on Build GRCh38
rs776043910 Chr.2: 174422855-174422855 on Build GRCh38
rs779710465 Chr.2: 174422903-174422903 on Build GRCh38
rs570817462 Chr.2: 174423100-174423100 on Build GRCh38
rs566461857 Chr.2: 174422830-174422830 on Build GRCh38
rs2290158 Chr.2: 174423050-174423050 on Build GRCh38
rs746123505 Chr.2: 174423055-174423055 on Build GRCh38
rs764490668 Chr.2: 174422860-174422860 on Build GRCh38
rs761209587 Chr.2: 174422953-174422953 on Build GRCh38
rs888504447 Chr.2: 174422747-174422747 on Build GRCh38
rs937741152 Chr.2: 174422785-174422785 on Build GRCh38
rs772197215 Chr.2: 174422941-174422941 on Build GRCh38
rs1007059988 Chr.2: 174422880-174422880 on Build GRCh38
rs201412954 Chr.2: 174422879-174422879 on Build GRCh38
rs754171822 Chr.2: 174422862-174422862 on Build GRCh38
rs767566854 Chr.2: 174422853-174422853 on Build GRCh38
rs769516492 Chr.2: 174422926-174422926 on Build GRCh38
rs749558523 Chr.2: 174422840-174422840 on Build GRCh38
rs949305465 Chr.2: 174423056-174423056 on Build GRCh38
rs770801211 Chr.2: 174423123-174423123 on Build GRCh38
rs895104614 Chr.2: 174422794-174422794 on Build GRCh38
rs377145764 Chr.2: 174422921-174422921 on Build GRCh38
rs765571983 Chr.2: 174422983-174422983 on Build GRCh38
rs747115497 Chr.2: 174423071-174423071 on Build GRCh38
rs375004342 Chr.2: 174422793-174422793 on Build GRCh38
rs772250241 Chr.2: 174423058-174423058 on Build GRCh38
rs753589368 Chr.2: 174422807-174422809 on Build GRCh38
rs951401922 Chr.2: 174422990-174422990 on Build GRCh38
rs111766947 Chr.2: 174422781-174422781 on Build GRCh38
rs192931480 Chr.2: 174422996-174422996 on Build GRCh38
rs571298327 Chr.2: 174422992-174422992 on Build GRCh38
rs1013501741 Chr.2: 174422799-174422799 on Build GRCh38
rs548479043 Chr.2: 174422767-174422767 on Build GRCh38
rs373970660 Chr.2: 174422849-174422849 on Build GRCh38
rs761154718 Chr.2: 174422856-174422856 on Build GRCh38
rs780864881 Chr.2: 174422748-174422748 on Build GRCh38
rs139815824 Chr.2: 174422891-174422891 on Build GRCh38

View Full Product Details