Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175179972-175180471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5G3
Entrez Gene ID
518
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Gene Aliases
P3
UniGene
Hs.429
Gene Chromosome Location
Chr.2: 175176258-175181762 on Build GRCh38
Transcripts
NM_001002258.4 NM_001190329.1 NM_001689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4920093 c.74T>A p.I25N Chr.2: 175180144-175180144 on Build GRCh38
1641710 c.40-10delT p.? Chr.2: 175180188-175180188 on Build GRCh38
5620337 c.109A>G p.R37G Chr.2: 175180109-175180109 on Build GRCh38
6607957 c.95G>A p.R32Q Chr.2: 175180123-175180123 on Build GRCh38
3771155 c.94C>T p.R32* Chr.2: 175180124-175180124 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371342791 Chr.2: 175180202-175180202 on Build GRCh38
rs34091276 Chr.2: 175180386-175180386 on Build GRCh38
rs914451829 Chr.2: 175180204-175180204 on Build GRCh38
rs751702566 Chr.2: 175180116-175180116 on Build GRCh38
rs912384757 Chr.2: 175180226-175180226 on Build GRCh38
rs983087440 Chr.2: 175180407-175180407 on Build GRCh38
rs550863029 Chr.2: 175180405-175180405 on Build GRCh38
rs953940766 Chr.2: 175180208-175180208 on Build GRCh38
rs902412469 Chr.2: 175180195-175180195 on Build GRCh38
rs955962968 Chr.2: 175180043-175180043 on Build GRCh38
rs76971553 Chr.2: 175180364-175180364 on Build GRCh38
rs768171823 Chr.2: 175180123-175180123 on Build GRCh38
rs115096123 Chr.2: 175180088-175180088 on Build GRCh38
rs768375380 Chr.2: 175180186-175180186 on Build GRCh38
rs779770959 Chr.2: 175180118-175180132 on Build GRCh38
rs879755837 Chr.2: 175180394-175180394 on Build GRCh38
rs759712161 Chr.2: 175180223-175180223 on Build GRCh38
rs576390385 Chr.2: 175180197-175180198 on Build GRCh38
rs919276554 Chr.2: 175180350-175180351 on Build GRCh38
rs76731943 Chr.2: 175180256-175180256 on Build GRCh38
rs923509637 Chr.2: 175180030-175180030 on Build GRCh38
rs562851671 Chr.2: 175180147-175180147 on Build GRCh38
rs888071529 Chr.2: 175180372-175180372 on Build GRCh38
rs144490355 Chr.2: 175180135-175180135 on Build GRCh38
rs112555046 Chr.2: 175180180-175180180 on Build GRCh38
rs908980392 Chr.2: 175180084-175180084 on Build GRCh38
rs182114394 Chr.2: 175180078-175180078 on Build GRCh38
rs934699973 Chr.2: 175180216-175180216 on Build GRCh38
rs76567292 Chr.2: 175180161-175180161 on Build GRCh38
rs746796011 Chr.2: 175180188-175180188 on Build GRCh38
rs766633186 Chr.2: 175180111-175180111 on Build GRCh38
rs887956941 Chr.2: 175180156-175180156 on Build GRCh38
rs776481245 Chr.2: 175180194-175180194 on Build GRCh38
rs151324447 Chr.2: 175180061-175180061 on Build GRCh38
rs759623853 Chr.2: 175180120-175180120 on Build GRCh38
rs761642633 Chr.2: 175180196-175180196 on Build GRCh38
rs976400700 Chr.2: 175180009-175180009 on Build GRCh38
rs543134495 Chr.2: 175180152-175180152 on Build GRCh38
rs990042446 Chr.2: 175180047-175180047 on Build GRCh38
rs532866764 Chr.2: 175180403-175180403 on Build GRCh38
rs1049306395 Chr.2: 175180365-175180365 on Build GRCh38
rs540926879 Chr.2: 175180385-175180385 on Build GRCh38
rs369817332 Chr.2: 175180071-175180071 on Build GRCh38
rs566491563 Chr.2: 175180007-175180007 on Build GRCh38
rs944802570 Chr.2: 175180127-175180127 on Build GRCh38
rs201787994 Chr.2: 175180035-175180035 on Build GRCh38
rs372765094 Chr.2: 175180149-175180149 on Build GRCh38
rs533821173 Chr.2: 175180016-175180016 on Build GRCh38
rs774675726 Chr.2: 175180203-175180203 on Build GRCh38
rs773353629 Chr.2: 175180080-175180080 on Build GRCh38
rs1030605271 Chr.2: 175180400-175180400 on Build GRCh38
rs778171387 Chr.2: 175180128-175180128 on Build GRCh38
rs768466127 Chr.2: 175180187-175180188 on Build GRCh38
rs753201238 Chr.2: 175180124-175180124 on Build GRCh38
rs752545228 Chr.2: 175180160-175180160 on Build GRCh38
rs929267392 Chr.2: 175180350-175180350 on Build GRCh38
rs893230811 Chr.2: 175180249-175180249 on Build GRCh38
rs1053077964 Chr.2: 175180238-175180238 on Build GRCh38
rs998080561 Chr.2: 175180399-175180399 on Build GRCh38
rs746941539 Chr.2: 175180162-175180162 on Build GRCh38
rs1040437936 Chr.2: 175180317-175180317 on Build GRCh38
rs755968861 Chr.2: 175180308-175180308 on Build GRCh38
rs1015843757 Chr.2: 175180414-175180414 on Build GRCh38

View Full Product Details