Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176129512-176130035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD8
Entrez Gene ID
3234
Gene Name
homeobox D8
Gene Aliases
HOX4, HOX4E, HOX5.4
UniGene
Hs.301963
Gene Chromosome Location
Chr.2: 176129694-176132695 on Build GRCh38
Transcripts
NM_001199746.1 NM_001199747.1 NM_019558.3

Gene Symbol
LOC100129455
Entrez Gene ID
100129455
Gene Name
uncharacterized LOC100129455
Gene Aliases
-
UniGene
Hs.636572
Gene Chromosome Location
Chr.2: 176127085-176129631 on Build GRCh38
Transcripts
NM_001199746.1 NM_001199747.1 NM_019558.3

SNP Detail

SNP ID Chromosome Location
rs141068157 Chr.2: 176130000-176130000 on Build GRCh38
rs1010570819 Chr.2: 176129676-176129676 on Build GRCh38
rs980847350 Chr.2: 176129702-176129702 on Build GRCh38
rs550045449 Chr.2: 176129720-176129720 on Build GRCh38
rs768187327 Chr.2: 176129679-176129679 on Build GRCh38
rs939259449 Chr.2: 176129667-176129667 on Build GRCh38
rs954865428 Chr.2: 176129681-176129681 on Build GRCh38
rs892104069 Chr.2: 176129546-176129546 on Build GRCh38
rs1022053744 Chr.2: 176129556-176129556 on Build GRCh38
rs997253273 Chr.2: 176129844-176129844 on Build GRCh38
rs1043674374 Chr.2: 176129756-176129756 on Build GRCh38
rs772599850 Chr.2: 176129610-176129610 on Build GRCh38
rs1009106062 Chr.2: 176129683-176129683 on Build GRCh38
rs973498406 Chr.2: 176129617-176129617 on Build GRCh38
rs886129495 Chr.2: 176129871-176129874 on Build GRCh38
rs973358055 Chr.2: 176129696-176129696 on Build GRCh38
rs529259495 Chr.2: 176129770-176129770 on Build GRCh38
rs879720943 Chr.2: 176129583-176129583 on Build GRCh38
rs892177517 Chr.2: 176129675-176129675 on Build GRCh38
rs981360496 Chr.2: 176129705-176129705 on Build GRCh38
rs913939857 Chr.2: 176129733-176129733 on Build GRCh38
rs550351167 Chr.2: 176129698-176129698 on Build GRCh38
rs910083678 Chr.2: 176129609-176129609 on Build GRCh38
rs945285569 Chr.2: 176129649-176129649 on Build GRCh38
rs943296331 Chr.2: 176129979-176129979 on Build GRCh38
rs568390529 Chr.2: 176129729-176129729 on Build GRCh38
rs968945931 Chr.2: 176129559-176129559 on Build GRCh38
rs1045320507 Chr.2: 176129660-176129660 on Build GRCh38
rs966731869 Chr.2: 176129956-176129956 on Build GRCh38
rs1029269224 Chr.2: 176129575-176129575 on Build GRCh38
rs960859466 Chr.2: 176129934-176129934 on Build GRCh38
rs975731304 Chr.2: 176129588-176129588 on Build GRCh38
rs847163 Chr.2: 176129895-176129895 on Build GRCh38
rs547591313 Chr.2: 176129789-176129789 on Build GRCh38
rs920662996 Chr.2: 176129631-176129631 on Build GRCh38
rs1052388467 Chr.2: 176129734-176129734 on Build GRCh38
rs899559738 Chr.2: 176129678-176129678 on Build GRCh38
rs892434187 Chr.2: 176129633-176129633 on Build GRCh38
rs901648687 Chr.2: 176129832-176129832 on Build GRCh38
rs1052312957 Chr.2: 176129672-176129672 on Build GRCh38
rs947937538 Chr.2: 176129658-176129658 on Build GRCh38
rs749325797 Chr.2: 176129534-176129534 on Build GRCh38
rs1025911040 Chr.2: 176129591-176129591 on Build GRCh38
rs746710263 Chr.2: 176129677-176129677 on Build GRCh38
rs992284842 Chr.2: 176129953-176129953 on Build GRCh38
rs979847091 Chr.2: 176129969-176129969 on Build GRCh38
rs1002420999 Chr.2: 176129922-176129922 on Build GRCh38
rs913983995 Chr.2: 176129955-176129955 on Build GRCh38
rs775969710 Chr.2: 176129644-176129644 on Build GRCh38
rs926687642 Chr.2: 176129616-176129616 on Build GRCh38
rs1015127065 Chr.2: 176129691-176129691 on Build GRCh38
rs975301050 Chr.2: 176129565-176129565 on Build GRCh38
rs1033856674 Chr.2: 176129925-176129925 on Build GRCh38
rs566125600 Chr.2: 176129814-176129814 on Build GRCh38
rs962048854 Chr.2: 176129694-176129694 on Build GRCh38
rs748290187 Chr.2: 176129732-176129749 on Build GRCh38
rs974676158 Chr.2: 176129991-176129991 on Build GRCh38
rs545944847 Chr.2: 176129606-176129606 on Build GRCh38
rs1025942300 Chr.2: 176129684-176129684 on Build GRCh38
rs960784821 Chr.2: 176129724-176129724 on Build GRCh38
rs1006287258 Chr.2: 176129887-176129887 on Build GRCh38
rs779301705 Chr.2: 176129566-176129566 on Build GRCh38
rs903726444 Chr.2: 176129668-176129668 on Build GRCh38
rs925181449 Chr.2: 176129775-176129775 on Build GRCh38
rs980902991 Chr.2: 176129612-176129612 on Build GRCh38
rs990117472 Chr.2: 176129829-176129829 on Build GRCh38
rs527887130 Chr.2: 176129603-176129603 on Build GRCh38
rs963308377 Chr.2: 176129906-176129906 on Build GRCh38
rs927791303 Chr.2: 176129664-176129664 on Build GRCh38
rs1022882937 Chr.2: 176129869-176129869 on Build GRCh38
rs951632264 Chr.2: 176129593-176129593 on Build GRCh38
rs1056498183 Chr.2: 176129620-176129621 on Build GRCh38
rs757296842 Chr.2: 176129543-176129543 on Build GRCh38
rs957449262 Chr.2: 176129822-176129822 on Build GRCh38
rs903651743 Chr.2: 176129563-176129563 on Build GRCh38
rs1044029661 Chr.2: 176129651-176129651 on Build GRCh38
rs1029586858 Chr.2: 176129680-176129680 on Build GRCh38
rs1010665576 Chr.2: 176129554-176129554 on Build GRCh38
rs920584135 Chr.2: 176129697-176129697 on Build GRCh38
rs746332012 Chr.2: 176129600-176129600 on Build GRCh38
rs955147064 Chr.2: 176129581-176129581 on Build GRCh38
rs913804766 Chr.2: 176129755-176129755 on Build GRCh38
rs847162 Chr.2: 176130011-176130011 on Build GRCh38
rs531922295 Chr.2: 176129711-176129711 on Build GRCh38

View Full Product Details