Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178832041-178832543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC141
Entrez Gene ID
285025
Gene Name
coiled-coil domain containing 141
Gene Aliases
CAMDI
UniGene
Hs.324341
Gene Chromosome Location
Chr.2: 178825308-179050134 on Build GRCh38
Transcripts
NM_173648.3 XM_011510992.2 XM_011510993.2 XM_017003867.1 XM_017003868.1 XM_017003869.1 XM_017003870.1

SNP Detail

SNP ID Chromosome Location
rs12693169 Chr.2: 178832399-178832399 on Build GRCh38
rs141512722 Chr.2: 178832400-178832400 on Build GRCh38
rs540080617 Chr.2: 178832165-178832165 on Build GRCh38
rs907139188 Chr.2: 178832213-178832213 on Build GRCh38
rs957219061 Chr.2: 178832115-178832115 on Build GRCh38
rs762991315 Chr.2: 178832254-178832254 on Build GRCh38
rs1020978365 Chr.2: 178832118-178832118 on Build GRCh38
rs918337763 Chr.2: 178832366-178832366 on Build GRCh38
rs959613442 Chr.2: 178832205-178832205 on Build GRCh38
rs55935293 Chr.2: 178832483-178832484 on Build GRCh38
rs953647733 Chr.2: 178832387-178832387 on Build GRCh38
rs1045587974 Chr.2: 178832212-178832212 on Build GRCh38
rs571068791 Chr.2: 178832142-178832142 on Build GRCh38
rs998481913 Chr.2: 178832068-178832068 on Build GRCh38
rs74954944 Chr.2: 178832100-178832100 on Build GRCh38
rs889700443 Chr.2: 178832085-178832085 on Build GRCh38
rs986300465 Chr.2: 178832396-178832396 on Build GRCh38
rs540268653 Chr.2: 178832415-178832415 on Build GRCh38
rs751846198 Chr.2: 178832411-178832411 on Build GRCh38
rs531436238 Chr.2: 178832476-178832476 on Build GRCh38
rs982127603 Chr.2: 178832172-178832172 on Build GRCh38
rs942597319 Chr.2: 178832414-178832414 on Build GRCh38
rs1020571939 Chr.2: 178832478-178832478 on Build GRCh38
rs541623033 Chr.2: 178832479-178832479 on Build GRCh38
rs2046776 Chr.2: 178832463-178832463 on Build GRCh38
rs1004042171 Chr.2: 178832482-178832482 on Build GRCh38
rs933716264 Chr.2: 178832507-178832507 on Build GRCh38
rs866629298 Chr.2: 178832511-178832511 on Build GRCh38
rs1010916149 Chr.2: 178832116-178832116 on Build GRCh38
rs567184078 Chr.2: 178832236-178832236 on Build GRCh38
rs915417638 Chr.2: 178832111-178832111 on Build GRCh38
rs1036397652 Chr.2: 178832141-178832141 on Build GRCh38
rs1048875443 Chr.2: 178832185-178832185 on Build GRCh38
rs74642120 Chr.2: 178832237-178832237 on Build GRCh38
rs893079515 Chr.2: 178832422-178832422 on Build GRCh38
rs910023562 Chr.2: 178832247-178832247 on Build GRCh38
rs998467158 Chr.2: 178832228-178832228 on Build GRCh38
rs892763340 Chr.2: 178832514-178832514 on Build GRCh38
rs143379106 Chr.2: 178832201-178832201 on Build GRCh38
rs79196305 Chr.2: 178832491-178832491 on Build GRCh38
rs139983321 Chr.2: 178832449-178832449 on Build GRCh38
rs207462681 Chr.2: 178832257-178832257 on Build GRCh38
rs957124234 Chr.2: 178832315-178832315 on Build GRCh38
rs149644403 Chr.2: 178832077-178832077 on Build GRCh38
rs1007227127 Chr.2: 178832095-178832095 on Build GRCh38
rs918756680 Chr.2: 178832504-178832504 on Build GRCh38
rs527739022 Chr.2: 178832488-178832488 on Build GRCh38
rs866375409 Chr.2: 178832493-178832493 on Build GRCh38
rs772181466 Chr.2: 178832484-178832484 on Build GRCh38
rs1049590898 Chr.2: 178832442-178832442 on Build GRCh38
rs941495819 Chr.2: 178832309-178832309 on Build GRCh38
rs777939064 Chr.2: 178832465-178832466 on Build GRCh38
rs2046775 Chr.2: 178832484-178832484 on Build GRCh38
rs562555442 Chr.2: 178832475-178832475 on Build GRCh38
rs962993019 Chr.2: 178832487-178832487 on Build GRCh38
rs1012773057 Chr.2: 178832209-178832209 on Build GRCh38
rs1045576796 Chr.2: 178832481-178832481 on Build GRCh38
rs536043789 Chr.2: 178832330-178832330 on Build GRCh38
rs147495917 Chr.2: 178832419-178832419 on Build GRCh38
rs894272141 Chr.2: 178832189-178832189 on Build GRCh38
rs944649937 Chr.2: 178832122-178832122 on Build GRCh38
rs866446734 Chr.2: 178832455-178832455 on Build GRCh38
rs59389117 Chr.2: 178832483-178832483 on Build GRCh38
rs868718648 Chr.2: 178832485-178832485 on Build GRCh38
rs1017177862 Chr.2: 178832486-178832486 on Build GRCh38
rs2839769 Chr.2: 178832317-178832317 on Build GRCh38
rs546050846 Chr.2: 178832473-178832473 on Build GRCh38
rs561889263 Chr.2: 178832483-178832483 on Build GRCh38
rs1035439858 Chr.2: 178832178-178832178 on Build GRCh38
rs934600563 Chr.2: 178832502-178832502 on Build GRCh38
rs902563557 Chr.2: 178832480-178832480 on Build GRCh38
rs1010022867 Chr.2: 178832466-178832466 on Build GRCh38

View Full Product Details