Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 179444737-179445258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF385B
Entrez Gene ID
151126
Gene Name
zinc finger protein 385B
Gene Aliases
ZNF533
UniGene
Hs.655005
Gene Chromosome Location
Chr.2: 179441982-179864550 on Build GRCh38
Transcripts
NM_001113397.1 NM_001113398.1 NM_001282725.1 NM_152520.4 NR_104234.1 XM_011510713.2 XM_011510714.2 XM_011510715.2 XM_011510716.2 XM_011510717.2 XM_011510719.2 XM_011510720.2 XM_011510721.2 XM_017003435.1 XM_017003436.1 XM_017003437.1 XM_017003438.1 XM_017003439.1 XM_017003440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4089375 c.949C>T p.R317W Chr.2: 179444896-179444896 on Build GRCh38
4778377 c.1197+4A>C p.? Chr.2: 179444872-179444872 on Build GRCh38
6846059 c.946C>T p.Q316* Chr.2: 179444899-179444899 on Build GRCh38
3990890 c.969A>T p.A323A Chr.2: 179444876-179444876 on Build GRCh38
6456006 c.1108C>T p.R370* Chr.2: 179444965-179444965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779245294 Chr.2: 179444906-179444906 on Build GRCh38
rs1001963440 Chr.2: 179445112-179445112 on Build GRCh38
rs760630158 Chr.2: 179444941-179444941 on Build GRCh38
rs568969068 Chr.2: 179445109-179445109 on Build GRCh38
rs759319215 Chr.2: 179445035-179445036 on Build GRCh38
rs761355854 Chr.2: 179444861-179444861 on Build GRCh38
rs997298277 Chr.2: 179444938-179444938 on Build GRCh38
rs779574586 Chr.2: 179444889-179444889 on Build GRCh38
rs183122712 Chr.2: 179444981-179444981 on Build GRCh38
rs777904953 Chr.2: 179444968-179444968 on Build GRCh38
rs749729993 Chr.2: 179444898-179444898 on Build GRCh38
rs768693941 Chr.2: 179444942-179444942 on Build GRCh38
rs887661170 Chr.2: 179444815-179444815 on Build GRCh38
rs1027607851 Chr.2: 179444810-179444810 on Build GRCh38
rs942814480 Chr.2: 179444930-179444930 on Build GRCh38
rs1034749320 Chr.2: 179445134-179445134 on Build GRCh38
rs965687670 Chr.2: 179444915-179444915 on Build GRCh38
rs574453009 Chr.2: 179445176-179445176 on Build GRCh38
rs1014568409 Chr.2: 179444890-179444890 on Build GRCh38
rs538099800 Chr.2: 179445145-179445145 on Build GRCh38
rs1021030120 Chr.2: 179445151-179445151 on Build GRCh38
rs950038792 Chr.2: 179445142-179445142 on Build GRCh38
rs765003835 Chr.2: 179444862-179444862 on Build GRCh38
rs772237623 Chr.2: 179444916-179444916 on Build GRCh38
rs776282714 Chr.2: 179444949-179444949 on Build GRCh38
rs778385833 Chr.2: 179444988-179444988 on Build GRCh38
rs750744643 Chr.2: 179444826-179444826 on Build GRCh38
rs538441153 Chr.2: 179445031-179445031 on Build GRCh38
rs905722574 Chr.2: 179444776-179444776 on Build GRCh38
rs763216067 Chr.2: 179444836-179444836 on Build GRCh38
rs760712382 Chr.2: 179444829-179444829 on Build GRCh38
rs751033754 Chr.2: 179445090-179445090 on Build GRCh38
rs967693566 Chr.2: 179445163-179445163 on Build GRCh38
rs746750739 Chr.2: 179445002-179445002 on Build GRCh38
rs750056621 Chr.2: 179444964-179444964 on Build GRCh38
rs571494357 Chr.2: 179444854-179444854 on Build GRCh38
rs905032152 Chr.2: 179445093-179445093 on Build GRCh38
rs761704803 Chr.2: 179444950-179444950 on Build GRCh38
rs1045856702 Chr.2: 179445173-179445173 on Build GRCh38
rs3828332 Chr.2: 179445035-179445035 on Build GRCh38
rs568414639 Chr.2: 179444868-179444868 on Build GRCh38
rs753677755 Chr.2: 179444858-179444858 on Build GRCh38
rs988786358 Chr.2: 179445034-179445034 on Build GRCh38
rs1047494335 Chr.2: 179445006-179445006 on Build GRCh38
rs754131573 Chr.2: 179445005-179445005 on Build GRCh38
rs367571052 Chr.2: 179444986-179444986 on Build GRCh38
rs1038250826 Chr.2: 179445056-179445056 on Build GRCh38
rs926199782 Chr.2: 179445189-179445189 on Build GRCh38
rs926109423 Chr.2: 179444921-179444921 on Build GRCh38
rs762602913 Chr.2: 179444965-179444965 on Build GRCh38
rs746114859 Chr.2: 179444909-179444909 on Build GRCh38
rs138370654 Chr.2: 179444852-179444852 on Build GRCh38
rs202072834 Chr.2: 179444900-179444900 on Build GRCh38
rs775714592 Chr.2: 179444931-179444931 on Build GRCh38
rs1001772364 Chr.2: 179444781-179444781 on Build GRCh38
rs200150920 Chr.2: 179444896-179444896 on Build GRCh38
rs145623392 Chr.2: 179444888-179444888 on Build GRCh38
rs927362236 Chr.2: 179445224-179445224 on Build GRCh38
rs764770018 Chr.2: 179444954-179444954 on Build GRCh38
rs756630059 Chr.2: 179444895-179444895 on Build GRCh38
rs1029156951 Chr.2: 179444959-179444959 on Build GRCh38
rs554592162 Chr.2: 179445167-179445167 on Build GRCh38

View Full Product Details