Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189560856-189561366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC40A1
Entrez Gene ID
30061
Gene Name
solute carrier family 40 member 1
Gene Aliases
FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3
UniGene
Hs.643005
Gene Chromosome Location
Chr.2: 189560590-189580811 on Build GRCh38
Transcripts
NM_014585.5 XM_005246505.1 XM_017003938.1

SNP Detail

SNP ID Chromosome Location
rs935138958 Chr.2: 189561023-189561023 on Build GRCh38
rs930400498 Chr.2: 189561232-189561232 on Build GRCh38
rs985575990 Chr.2: 189561313-189561313 on Build GRCh38
rs1015120544 Chr.2: 189561105-189561105 on Build GRCh38
rs994996376 Chr.2: 189561174-189561174 on Build GRCh38
rs958663617 Chr.2: 189561021-189561021 on Build GRCh38
rs1020433271 Chr.2: 189560994-189560994 on Build GRCh38
rs931035208 Chr.2: 189560952-189560952 on Build GRCh38
rs985243278 Chr.2: 189560953-189560956 on Build GRCh38
rs773624861 Chr.2: 189561108-189561108 on Build GRCh38
rs565741596 Chr.2: 189561142-189561143 on Build GRCh38
rs886055359 Chr.2: 189561232-189561232 on Build GRCh38
rs567324403 Chr.2: 189561186-189561186 on Build GRCh38
rs562536353 Chr.2: 189560888-189560888 on Build GRCh38
rs1045161932 Chr.2: 189561069-189561069 on Build GRCh38
rs147807428 Chr.2: 189560999-189560999 on Build GRCh38
rs917191527 Chr.2: 189561045-189561045 on Build GRCh38
rs893657861 Chr.2: 189561038-189561038 on Build GRCh38
rs919066031 Chr.2: 189561222-189561222 on Build GRCh38
rs953582371 Chr.2: 189561296-189561296 on Build GRCh38
rs991497059 Chr.2: 189561032-189561032 on Build GRCh38
rs371205681 Chr.2: 189561156-189561156 on Build GRCh38
rs906503918 Chr.2: 189561090-189561090 on Build GRCh38
rs972333472 Chr.2: 189561122-189561122 on Build GRCh38
rs910846589 Chr.2: 189560957-189560957 on Build GRCh38
rs569348005 Chr.2: 189561295-189561295 on Build GRCh38
rs1000214695 Chr.2: 189561000-189561000 on Build GRCh38
rs911015902 Chr.2: 189561324-189561324 on Build GRCh38
rs563974199 Chr.2: 189560953-189560953 on Build GRCh38
rs549021644 Chr.2: 189561051-189561051 on Build GRCh38
rs537552855 Chr.2: 189561225-189561225 on Build GRCh38
rs774721281 Chr.2: 189561007-189561007 on Build GRCh38
rs766516631 Chr.2: 189561170-189561170 on Build GRCh38
rs530886053 Chr.2: 189561081-189561081 on Build GRCh38
rs566807636 Chr.2: 189560975-189560975 on Build GRCh38
rs367652885 Chr.2: 189561125-189561125 on Build GRCh38
rs772418551 Chr.2: 189561046-189561046 on Build GRCh38
rs549887682 Chr.2: 189561256-189561256 on Build GRCh38
rs533203775 Chr.2: 189560896-189560896 on Build GRCh38
rs1040980769 Chr.2: 189560976-189560976 on Build GRCh38
rs760814102 Chr.2: 189561147-189561147 on Build GRCh38
rs748527134 Chr.2: 189561016-189561016 on Build GRCh38
rs189968883 Chr.2: 189560965-189560965 on Build GRCh38
rs1003537811 Chr.2: 189561098-189561098 on Build GRCh38

View Full Product Details