Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199911165-199911663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf69
Entrez Gene ID
205327
Gene Name
chromosome 2 open reading frame 69
Gene Aliases
-
UniGene
Hs.154494
Gene Chromosome Location
Chr.2: 199911256-199928273 on Build GRCh38
Transcripts
NM_153689.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6497785 c.3G>C p.M1I Chr.2: 199911441-199911441 on Build GRCh38
6204402 c.70G>T p.A24S Chr.2: 199911508-199911508 on Build GRCh38
399089 c.177C>A p.T59T Chr.2: 199911615-199911615 on Build GRCh38
4620784 c.4T>C p.W2R Chr.2: 199911442-199911442 on Build GRCh38
3749627 c.192T>G p.D64E Chr.2: 199911630-199911630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530626964 Chr.2: 199911298-199911298 on Build GRCh38
rs146692252 Chr.2: 199911290-199911290 on Build GRCh38
rs994774174 Chr.2: 199911254-199911254 on Build GRCh38
rs959031553 Chr.2: 199911191-199911191 on Build GRCh38
rs747333931 Chr.2: 199911471-199911471 on Build GRCh38
rs532672769 Chr.2: 199911268-199911268 on Build GRCh38
rs886452471 Chr.2: 199911371-199911371 on Build GRCh38
rs762288716 Chr.2: 199911593-199911593 on Build GRCh38
rs1052226714 Chr.2: 199911364-199911364 on Build GRCh38
rs945212998 Chr.2: 199911225-199911225 on Build GRCh38
rs996936635 Chr.2: 199911607-199911607 on Build GRCh38
rs866052049 Chr.2: 199911626-199911626 on Build GRCh38
rs777065957 Chr.2: 199911509-199911509 on Build GRCh38
rs772200305 Chr.2: 199911316-199911316 on Build GRCh38
rs921258487 Chr.2: 199911328-199911329 on Build GRCh38
rs540650009 Chr.2: 199911545-199911545 on Build GRCh38
rs937498852 Chr.2: 199911615-199911615 on Build GRCh38
rs776086262 Chr.2: 199911395-199911395 on Build GRCh38
rs767030169 Chr.2: 199911527-199911527 on Build GRCh38
rs921478144 Chr.2: 199911468-199911468 on Build GRCh38
rs987178109 Chr.2: 199911486-199911486 on Build GRCh38
rs538524137 Chr.2: 199911378-199911378 on Build GRCh38
rs936498628 Chr.2: 199911332-199911332 on Build GRCh38
rs746356835 Chr.2: 199911628-199911628 on Build GRCh38
rs753818494 Chr.2: 199911569-199911569 on Build GRCh38
rs1038961463 Chr.2: 199911334-199911334 on Build GRCh38
rs753547843 Chr.2: 199911394-199911394 on Build GRCh38
rs540730599 Chr.2: 199911288-199911288 on Build GRCh38
rs540256555 Chr.2: 199911214-199911214 on Build GRCh38
rs997551553 Chr.2: 199911279-199911279 on Build GRCh38
rs1037791612 Chr.2: 199911384-199911384 on Build GRCh38
rs1020542975 Chr.2: 199911632-199911632 on Build GRCh38
rs925068545 Chr.2: 199911328-199911328 on Build GRCh38
rs991812241 Chr.2: 199911315-199911315 on Build GRCh38
rs1034698725 Chr.2: 199911410-199911410 on Build GRCh38
rs917700068 Chr.2: 199911220-199911220 on Build GRCh38
rs748368142 Chr.2: 199911426-199911426 on Build GRCh38
rs565435469 Chr.2: 199911552-199911552 on Build GRCh38
rs974597838 Chr.2: 199911543-199911543 on Build GRCh38
rs955658665 Chr.2: 199911484-199911484 on Build GRCh38
rs772982593 Chr.2: 199911637-199911637 on Build GRCh38
rs1022042628 Chr.2: 199911577-199911577 on Build GRCh38
rs779068208 Chr.2: 199911402-199911402 on Build GRCh38
rs748855175 Chr.2: 199911515-199911515 on Build GRCh38
rs1032126923 Chr.2: 199911521-199911521 on Build GRCh38
rs577488855 Chr.2: 199911588-199911588 on Build GRCh38
rs777904112 Chr.2: 199911591-199911591 on Build GRCh38
rs1014473935 Chr.2: 199911502-199911502 on Build GRCh38
rs959031351 Chr.2: 199911310-199911310 on Build GRCh38
rs34642678 Chr.2: 199911242-199911243 on Build GRCh38
rs565618527 Chr.2: 199911237-199911237 on Build GRCh38
rs963595477 Chr.2: 199911304-199911304 on Build GRCh38
rs1022407764 Chr.2: 199911443-199911443 on Build GRCh38
rs563083229 Chr.2: 199911633-199911633 on Build GRCh38
rs567361029 Chr.2: 199911317-199911317 on Build GRCh38
rs552674272 Chr.2: 199911353-199911353 on Build GRCh38
rs1016815567 Chr.2: 199911303-199911303 on Build GRCh38
rs769103816 Chr.2: 199911472-199911472 on Build GRCh38
rs1034571173 Chr.2: 199911309-199911309 on Build GRCh38
rs534654554 Chr.2: 199911320-199911320 on Build GRCh38
rs1051105559 Chr.2: 199911345-199911345 on Build GRCh38
rs755050592 Chr.2: 199911570-199911570 on Build GRCh38
rs757604720 Chr.2: 199911616-199911616 on Build GRCh38
rs551188628 Chr.2: 199911281-199911281 on Build GRCh38
rs1025039325 Chr.2: 199911512-199911512 on Build GRCh38
rs760609777 Chr.2: 199911639-199911639 on Build GRCh38
rs994743929 Chr.2: 199911381-199911381 on Build GRCh38
rs934065986 Chr.2: 199911188-199911188 on Build GRCh38
rs1036581005 Chr.2: 199911357-199911357 on Build GRCh38
rs781715546 Chr.2: 199911617-199911617 on Build GRCh38
rs571553456 Chr.2: 199911356-199911356 on Build GRCh38
rs773176679 Chr.2: 199911541-199911541 on Build GRCh38
rs755068015 Chr.2: 199911397-199911397 on Build GRCh38
rs536354517 Chr.2: 199911516-199911516 on Build GRCh38
rs572997218 Chr.2: 199911526-199911526 on Build GRCh38
rs896102238 Chr.2: 199911464-199911464 on Build GRCh38
rs1038972866 Chr.2: 199911246-199911246 on Build GRCh38
rs747576112 Chr.2: 199911631-199911631 on Build GRCh38
rs936549634 Chr.2: 199911243-199911243 on Build GRCh38
rs940706762 Chr.2: 199911595-199911595 on Build GRCh38
rs575046114 Chr.2: 199911469-199911469 on Build GRCh38
rs771430930 Chr.2: 199911474-199911474 on Build GRCh38
rs1009726408 Chr.2: 199911566-199911566 on Build GRCh38
rs554682984 Chr.2: 199911519-199911519 on Build GRCh38
rs975636316 Chr.2: 199911455-199911455 on Build GRCh38
rs892160938 Chr.2: 199911558-199911558 on Build GRCh38
rs904854916 Chr.2: 199911391-199911391 on Build GRCh38
rs62178345 Chr.2: 199911630-199911630 on Build GRCh38
rs899281796 Chr.2: 199911613-199911613 on Build GRCh38
rs758737747 Chr.2: 199911445-199911445 on Build GRCh38
rs778131370 Chr.2: 199911470-199911470 on Build GRCh38
rs747388975 Chr.2: 199911611-199911611 on Build GRCh38
rs991864737 Chr.2: 199911206-199911206 on Build GRCh38
rs549058438 Chr.2: 199911308-199911308 on Build GRCh38
rs956629186 Chr.2: 199911423-199911423 on Build GRCh38
rs977887267 Chr.2: 199911322-199911322 on Build GRCh38
rs766478215 Chr.2: 199911556-199911556 on Build GRCh38
rs557139782 Chr.2: 199911396-199911396 on Build GRCh38
rs1034549409 Chr.2: 199911398-199911398 on Build GRCh38
rs1051652995 Chr.2: 199911189-199911189 on Build GRCh38
rs758678127 Chr.2: 199911589-199911589 on Build GRCh38
rs747542092 Chr.2: 199911438-199911438 on Build GRCh38
rs868667563 Chr.2: 199911586-199911586 on Build GRCh38
rs1035220275 Chr.2: 199911301-199911301 on Build GRCh38
rs1042645171 Chr.2: 199911229-199911229 on Build GRCh38

View Full Product Details