Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201185665-201186179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
UniGene
Hs.5353
Gene Chromosome Location
Chr.2: 201182881-201229406 on Build GRCh38
Transcripts
NM_001206524.1 NM_001206542.1 NM_001230.4 NM_001306083.1 NM_032974.4 NM_032976.3 NM_032977.3 XM_005246907.2 XM_006712796.3 XM_011511990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2906944 c.42A>C p.K14N Chr.2: 201185819-201185819 on Build GRCh38
2906949 c.106G>A p.V36M Chr.2: 201185883-201185883 on Build GRCh38
719806 c.202G>A p.E68K Chr.2: 201185979-201185979 on Build GRCh38
4090162 c.289T>C p.C97R Chr.2: 201186066-201186066 on Build GRCh38
239158 c.346A>T p.R116* Chr.2: 201186123-201186123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773417904 Chr.2: 201185940-201185940 on Build GRCh38
rs185792819 Chr.2: 201186032-201186032 on Build GRCh38
rs925956116 Chr.2: 201186073-201186073 on Build GRCh38
rs766784091 Chr.2: 201185907-201185907 on Build GRCh38
rs200935960 Chr.2: 201185797-201185797 on Build GRCh38
rs140246829 Chr.2: 201186132-201186132 on Build GRCh38
rs773803576 Chr.2: 201185891-201185891 on Build GRCh38
rs1047225163 Chr.2: 201185913-201185913 on Build GRCh38
rs114625983 Chr.2: 201185951-201185951 on Build GRCh38
rs751728796 Chr.2: 201185858-201185858 on Build GRCh38
rs762557222 Chr.2: 201185915-201185915 on Build GRCh38
rs774630500 Chr.2: 201185745-201185745 on Build GRCh38
rs761521100 Chr.2: 201186122-201186122 on Build GRCh38
rs559658034 Chr.2: 201185923-201185923 on Build GRCh38
rs200012872 Chr.2: 201185919-201185919 on Build GRCh38
rs769066879 Chr.2: 201185847-201185847 on Build GRCh38
rs769634137 Chr.2: 201185844-201185844 on Build GRCh38
rs1804650 Chr.2: 201185934-201185934 on Build GRCh38
rs140813639 Chr.2: 201185839-201185839 on Build GRCh38
rs750385913 Chr.2: 201185930-201185930 on Build GRCh38
rs375838979 Chr.2: 201185871-201185871 on Build GRCh38
rs761126176 Chr.2: 201185763-201185763 on Build GRCh38
rs146696981 Chr.2: 201186003-201186003 on Build GRCh38
rs199511092 Chr.2: 201185729-201185729 on Build GRCh38
rs201601111 Chr.2: 201186102-201186102 on Build GRCh38
rs764280980 Chr.2: 201185744-201185744 on Build GRCh38
rs942382015 Chr.2: 201185825-201185825 on Build GRCh38
rs763058483 Chr.2: 201185742-201185742 on Build GRCh38
rs767039731 Chr.2: 201186050-201186050 on Build GRCh38
rs3900115 Chr.2: 201185954-201185954 on Build GRCh38
rs764759465 Chr.2: 201185777-201185777 on Build GRCh38
rs773537509 Chr.2: 201185976-201185976 on Build GRCh38
rs761234746 Chr.2: 201185988-201185988 on Build GRCh38
rs1002649181 Chr.2: 201185963-201185963 on Build GRCh38
rs146684884 Chr.2: 201185903-201185903 on Build GRCh38
rs748398291 Chr.2: 201186088-201186088 on Build GRCh38
rs139417609 Chr.2: 201185953-201185953 on Build GRCh38
rs771377939 Chr.2: 201186002-201186002 on Build GRCh38
rs1000439250 Chr.2: 201185722-201185722 on Build GRCh38
rs552561721 Chr.2: 201185826-201185826 on Build GRCh38
rs768180020 Chr.2: 201185760-201185760 on Build GRCh38
rs757161113 Chr.2: 201185829-201185829 on Build GRCh38
rs752402003 Chr.2: 201185796-201185796 on Build GRCh38
rs774542679 Chr.2: 201185859-201185859 on Build GRCh38
rs779212325 Chr.2: 201186087-201186087 on Build GRCh38
rs189450587 Chr.2: 201186148-201186148 on Build GRCh38
rs777256257 Chr.2: 201185798-201185798 on Build GRCh38
rs991951998 Chr.2: 201186016-201186016 on Build GRCh38
rs192255542 Chr.2: 201185754-201185754 on Build GRCh38
rs531649047 Chr.2: 201186072-201186072 on Build GRCh38
rs745721366 Chr.2: 201185727-201185727 on Build GRCh38
rs750732272 Chr.2: 201185761-201185761 on Build GRCh38
rs148026756 Chr.2: 201186048-201186048 on Build GRCh38
rs775888850 Chr.2: 201186119-201186119 on Build GRCh38
rs975524888 Chr.2: 201186121-201186121 on Build GRCh38
rs768033545 Chr.2: 201186083-201186083 on Build GRCh38
rs377184631 Chr.2: 201186134-201186134 on Build GRCh38
rs770473281 Chr.2: 201186074-201186075 on Build GRCh38
rs370492434 Chr.2: 201186059-201186059 on Build GRCh38
rs770155279 Chr.2: 201186112-201186112 on Build GRCh38
rs759913674 Chr.2: 201186024-201186024 on Build GRCh38
rs749507462 Chr.2: 201185846-201185846 on Build GRCh38
rs199600895 Chr.2: 201186109-201186109 on Build GRCh38
rs551818122 Chr.2: 201186036-201186036 on Build GRCh38
rs780080444 Chr.2: 201185845-201185845 on Build GRCh38
rs751018783 Chr.2: 201185921-201185921 on Build GRCh38
rs536573363 Chr.2: 201186126-201186126 on Build GRCh38
rs775811329 Chr.2: 201185734-201185734 on Build GRCh38
rs939291713 Chr.2: 201186014-201186014 on Build GRCh38
rs754607963 Chr.2: 201186130-201186130 on Build GRCh38
rs746161446 Chr.2: 201186110-201186110 on Build GRCh38
rs912946102 Chr.2: 201185714-201185714 on Build GRCh38
rs756047946 Chr.2: 201186054-201186054 on Build GRCh38
rs777907841 Chr.2: 201186095-201186095 on Build GRCh38
rs7608787 Chr.2: 201186133-201186133 on Build GRCh38
rs753639297 Chr.2: 201186060-201186060 on Build GRCh38
rs200530667 Chr.2: 201186091-201186091 on Build GRCh38
rs753330049 Chr.2: 201186103-201186103 on Build GRCh38
rs772200260 Chr.2: 201185962-201185962 on Build GRCh38
rs559979934 Chr.2: 201185838-201185838 on Build GRCh38
rs778361451 Chr.2: 201185755-201185755 on Build GRCh38
rs752505651 Chr.2: 201186145-201186145 on Build GRCh38
rs749820662 Chr.2: 201186021-201186023 on Build GRCh38
rs749967724 Chr.2: 201186053-201186053 on Build GRCh38
rs908733923 Chr.2: 201186137-201186137 on Build GRCh38
rs772411929 Chr.2: 201185874-201185874 on Build GRCh38
rs780923455 Chr.2: 201185924-201185924 on Build GRCh38
rs201782416 Chr.2: 201185866-201185866 on Build GRCh38
rs942374240 Chr.2: 201186104-201186104 on Build GRCh38

View Full Product Details