Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203433720-203434266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAPH1
Entrez Gene ID
65059
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Gene Aliases
ALS2CR18, ALS2CR9, LPD, PREL-2, PREL2, RMO1, RalGDS/AF-6
UniGene
Hs.471162
Gene Chromosome Location
Chr.2: 203433682-203535335 on Build GRCh38
Transcripts
NM_213589.2 XM_005246777.3 XM_005246779.4 XM_006712695.3 XM_011511646.2 XM_011511647.1 XM_011511648.1

SNP Detail

SNP ID Chromosome Location
rs934306326 Chr.2: 203433786-203433786 on Build GRCh38
rs990361725 Chr.2: 203434055-203434055 on Build GRCh38
rs372249473 Chr.2: 203434057-203434058 on Build GRCh38
rs76899339 Chr.2: 203434031-203434031 on Build GRCh38
rs199519643 Chr.2: 203434055-203434056 on Build GRCh38
rs900703963 Chr.2: 203433969-203433973 on Build GRCh38
rs567216499 Chr.2: 203434054-203434055 on Build GRCh38
rs534403657 Chr.2: 203433897-203433897 on Build GRCh38
rs573120043 Chr.2: 203434105-203434105 on Build GRCh38
rs938739843 Chr.2: 203433849-203433849 on Build GRCh38
rs372040819 Chr.2: 203434075-203434080 on Build GRCh38
rs531130435 Chr.2: 203433829-203433829 on Build GRCh38
rs933519868 Chr.2: 203434209-203434210 on Build GRCh38
rs963346435 Chr.2: 203434043-203434043 on Build GRCh38
rs1052896606 Chr.2: 203433787-203433787 on Build GRCh38
rs775849397 Chr.2: 203433875-203433876 on Build GRCh38
rs974516020 Chr.2: 203434175-203434180 on Build GRCh38
rs1052332474 Chr.2: 203434213-203434213 on Build GRCh38
rs549119800 Chr.2: 203433869-203433869 on Build GRCh38
rs1016318707 Chr.2: 203433841-203433841 on Build GRCh38
rs10650425 Chr.2: 203434080-203434081 on Build GRCh38
rs200542761 Chr.2: 203434040-203434040 on Build GRCh38
rs546704483 Chr.2: 203434013-203434013 on Build GRCh38
rs67160931 Chr.2: 203434059-203434060 on Build GRCh38
rs71408937 Chr.2: 203434040-203434041 on Build GRCh38
rs879131031 Chr.2: 203434015-203434015 on Build GRCh38
rs950772999 Chr.2: 203433833-203433834 on Build GRCh38
rs538594533 Chr.2: 203434029-203434029 on Build GRCh38
rs969939147 Chr.2: 203434053-203434060 on Build GRCh38
rs551768978 Chr.2: 203433808-203433808 on Build GRCh38
rs10650424 Chr.2: 203434058-203434059 on Build GRCh38
rs201489616 Chr.2: 203434037-203434037 on Build GRCh38
rs981658482 Chr.2: 203434070-203434070 on Build GRCh38
rs563971085 Chr.2: 203433822-203433822 on Build GRCh38
rs940986076 Chr.2: 203434223-203434223 on Build GRCh38
rs938684978 Chr.2: 203434120-203434120 on Build GRCh38
rs563344471 Chr.2: 203434232-203434232 on Build GRCh38
rs1047510 Chr.2: 203434064-203434064 on Build GRCh38
rs145648108 Chr.2: 203434160-203434160 on Build GRCh38
rs887335510 Chr.2: 203433957-203433957 on Build GRCh38
rs953253332 Chr.2: 203433747-203433747 on Build GRCh38
rs1038798263 Chr.2: 203433968-203433968 on Build GRCh38
rs2264901 Chr.2: 203434079-203434079 on Build GRCh38
rs921654152 Chr.2: 203434203-203434204 on Build GRCh38
rs988475766 Chr.2: 203434131-203434131 on Build GRCh38
rs947026347 Chr.2: 203434171-203434171 on Build GRCh38
rs71757799 Chr.2: 203433834-203433834 on Build GRCh38
rs928522963 Chr.2: 203433907-203433907 on Build GRCh38
rs1048567303 Chr.2: 203433906-203433906 on Build GRCh38
rs990007897 Chr.2: 203433788-203433788 on Build GRCh38
rs915785096 Chr.2: 203433875-203433875 on Build GRCh38
rs1012321149 Chr.2: 203434047-203434047 on Build GRCh38
rs906901101 Chr.2: 203433757-203433757 on Build GRCh38
rs1004473580 Chr.2: 203433975-203433975 on Build GRCh38
rs928582159 Chr.2: 203434074-203434074 on Build GRCh38
rs745911946 Chr.2: 203433902-203433902 on Build GRCh38
rs963396423 Chr.2: 203433859-203433860 on Build GRCh38
rs1049002845 Chr.2: 203434218-203434218 on Build GRCh38
rs977785014 Chr.2: 203433827-203433827 on Build GRCh38
rs934362044 Chr.2: 203433930-203433931 on Build GRCh38
rs916052468 Chr.2: 203433988-203434001 on Build GRCh38
rs73990636 Chr.2: 203434044-203434044 on Build GRCh38
rs1023661878 Chr.2: 203434049-203434049 on Build GRCh38
rs201945070 Chr.2: 203434062-203434062 on Build GRCh38
rs578043874 Chr.2: 203434202-203434202 on Build GRCh38
rs1039184722 Chr.2: 203433807-203433807 on Build GRCh38
rs759538507 Chr.2: 203434201-203434201 on Build GRCh38
rs998077335 Chr.2: 203433981-203433981 on Build GRCh38
rs181502320 Chr.2: 203434107-203434107 on Build GRCh38
rs1052742027 Chr.2: 203433931-203433934 on Build GRCh38
rs1028724084 Chr.2: 203434050-203434050 on Build GRCh38
rs914330455 Chr.2: 203433943-203433944 on Build GRCh38
rs940260917 Chr.2: 203434234-203434234 on Build GRCh38
rs376564062 Chr.2: 203434214-203434214 on Build GRCh38
rs567526997 Chr.2: 203433896-203433896 on Build GRCh38
rs571142435 Chr.2: 203434027-203434027 on Build GRCh38
rs2957328 Chr.2: 203434056-203434056 on Build GRCh38
rs1046059495 Chr.2: 203433744-203433744 on Build GRCh38
rs970288455 Chr.2: 203433903-203433903 on Build GRCh38
rs1129558 Chr.2: 203434060-203434060 on Build GRCh38
rs1037910767 Chr.2: 203434229-203434230 on Build GRCh38
rs953202400 Chr.2: 203434052-203434052 on Build GRCh38
rs915967690 Chr.2: 203434161-203434161 on Build GRCh38

View Full Product Details