Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 205716115-205716617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRP2
Entrez Gene ID
8828
Gene Name
neuropilin 2
Gene Aliases
NP2, NPN2, PRO2714, VEGF165R2
UniGene
Hs.471200
Gene Chromosome Location
Chr.2: 205682450-205798133 on Build GRCh38
Transcripts
NM_003872.2 NM_018534.3 NM_201264.1 NM_201266.1 NM_201267.1 NM_201279.1 XM_005246933.3 XM_005246934.3 XM_017005185.1 XM_017005186.1 XM_017005187.1 XM_017005188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6188920 c.314G>A p.C105Y Chr.2: 205716255-205716255 on Build GRCh38
3576600 c.327C>T p.A109A Chr.2: 205716268-205716268 on Build GRCh38
3909371 c.271C>T p.R91W Chr.2: 205716212-205716212 on Build GRCh38
6729831 c.373A>G p.T125A Chr.2: 205716314-205716314 on Build GRCh38
6188919 c.314G>A p.C105Y Chr.2: 205716255-205716255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775295749 Chr.2: 205716408-205716408 on Build GRCh38
rs758549230 Chr.2: 205716238-205716238 on Build GRCh38
rs746365156 Chr.2: 205716294-205716294 on Build GRCh38
rs137895071 Chr.2: 205716261-205716261 on Build GRCh38
rs532731194 Chr.2: 205716331-205716331 on Build GRCh38
rs937135069 Chr.2: 205716512-205716512 on Build GRCh38
rs908897178 Chr.2: 205716189-205716189 on Build GRCh38
rs969270872 Chr.2: 205716542-205716542 on Build GRCh38
rs781637032 Chr.2: 205716149-205716149 on Build GRCh38
rs767624307 Chr.2: 205716382-205716382 on Build GRCh38
rs747518465 Chr.2: 205716147-205716147 on Build GRCh38
rs1023294841 Chr.2: 205716577-205716577 on Build GRCh38
rs1032541503 Chr.2: 205716347-205716347 on Build GRCh38
rs772785643 Chr.2: 205716188-205716188 on Build GRCh38
rs747811301 Chr.2: 205716314-205716314 on Build GRCh38
rs1000584356 Chr.2: 205716591-205716591 on Build GRCh38
rs201159993 Chr.2: 205716165-205716165 on Build GRCh38
rs756581851 Chr.2: 205716388-205716388 on Build GRCh38
rs781592704 Chr.2: 205716289-205716289 on Build GRCh38
rs768445628 Chr.2: 205716418-205716420 on Build GRCh38
rs764261615 Chr.2: 205716361-205716361 on Build GRCh38
rs969492038 Chr.2: 205716585-205716585 on Build GRCh38
rs139797563 Chr.2: 205716355-205716355 on Build GRCh38
rs150835990 Chr.2: 205716359-205716359 on Build GRCh38
rs940381865 Chr.2: 205716192-205716192 on Build GRCh38
rs745414434 Chr.2: 205716254-205716255 on Build GRCh38
rs777614760 Chr.2: 205716394-205716394 on Build GRCh38
rs367901430 Chr.2: 205716199-205716199 on Build GRCh38
rs775207599 Chr.2: 205716358-205716358 on Build GRCh38
rs373526002 Chr.2: 205716399-205716399 on Build GRCh38
rs766182324 Chr.2: 205716226-205716226 on Build GRCh38
rs1013407324 Chr.2: 205716486-205716486 on Build GRCh38
rs759512138 Chr.2: 205716353-205716353 on Build GRCh38
rs745806444 Chr.2: 205716400-205716400 on Build GRCh38
rs370588471 Chr.2: 205716148-205716148 on Build GRCh38
rs756496733 Chr.2: 205716295-205716295 on Build GRCh38
rs750463349 Chr.2: 205716422-205716422 on Build GRCh38
rs746749837 Chr.2: 205716374-205716382 on Build GRCh38
rs141877635 Chr.2: 205716156-205716156 on Build GRCh38
rs772877594 Chr.2: 205716328-205716328 on Build GRCh38
rs762777063 Chr.2: 205716409-205716409 on Build GRCh38
rs1023903034 Chr.2: 205716534-205716534 on Build GRCh38
rs773927775 Chr.2: 205716173-205716173 on Build GRCh38
rs371182108 Chr.2: 205716496-205716496 on Build GRCh38
rs372737222 Chr.2: 205716406-205716406 on Build GRCh38
rs746644568 Chr.2: 205716397-205716397 on Build GRCh38
rs376188302 Chr.2: 205716411-205716411 on Build GRCh38
rs971062951 Chr.2: 205716396-205716396 on Build GRCh38
rs776487113 Chr.2: 205716350-205716350 on Build GRCh38
rs751590695 Chr.2: 205716271-205716271 on Build GRCh38
rs771779558 Chr.2: 205716186-205716186 on Build GRCh38
rs779740453 Chr.2: 205716368-205716369 on Build GRCh38
rs530087986 Chr.2: 205716232-205716232 on Build GRCh38
rs751751923 Chr.2: 205716375-205716375 on Build GRCh38
rs755359753 Chr.2: 205716391-205716391 on Build GRCh38
rs747791031 Chr.2: 205716174-205716174 on Build GRCh38
rs368761335 Chr.2: 205716390-205716390 on Build GRCh38
rs1049360552 Chr.2: 205716523-205716523 on Build GRCh38
rs532033435 Chr.2: 205716594-205716594 on Build GRCh38
rs849541 Chr.2: 205716309-205716309 on Build GRCh38
rs771712741 Chr.2: 205716361-205716361 on Build GRCh38
rs201391637 Chr.2: 205716383-205716383 on Build GRCh38
rs940563565 Chr.2: 205716578-205716578 on Build GRCh38
rs765066964 Chr.2: 205716233-205716233 on Build GRCh38
rs757792703 Chr.2: 205716278-205716278 on Build GRCh38
rs569530464 Chr.2: 205716434-205716434 on Build GRCh38
rs770450540 Chr.2: 205716334-205716334 on Build GRCh38
rs768418889 Chr.2: 205716410-205716410 on Build GRCh38
rs757331205 Chr.2: 205716380-205716380 on Build GRCh38
rs35678298 Chr.2: 205716325-205716325 on Build GRCh38
rs909084928 Chr.2: 205716544-205716545 on Build GRCh38
rs764171906 Chr.2: 205716265-205716265 on Build GRCh38
rs770164649 Chr.2: 205716164-205716164 on Build GRCh38

View Full Product Details