Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207077152-207077660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF7
Entrez Gene ID
8609
Gene Name
Kruppel like factor 7
Gene Aliases
UKLF
UniGene
Hs.471221
Gene Chromosome Location
Chr.2: 207074137-207173851 on Build GRCh38
Transcripts
NM_001270942.1 NM_001270943.1 NM_001270944.1 NM_003709.3 NR_073108.1 XM_005246926.3 XM_006712815.3 XM_011512072.2 XM_011512073.2 XM_011512074.2 XM_017005161.1 XM_017005162.1 XM_017005164.1

SNP Detail

SNP ID Chromosome Location
rs961736342 Chr.2: 207077272-207077272 on Build GRCh38
rs953231898 Chr.2: 207077324-207077324 on Build GRCh38
rs1047539778 Chr.2: 207077200-207077200 on Build GRCh38
rs907659940 Chr.2: 207077490-207077490 on Build GRCh38
rs776809889 Chr.2: 207077526-207077526 on Build GRCh38
rs759811338 Chr.2: 207077593-207077593 on Build GRCh38
rs931671248 Chr.2: 207077518-207077518 on Build GRCh38
rs974067209 Chr.2: 207077435-207077437 on Build GRCh38
rs887489564 Chr.2: 207077224-207077224 on Build GRCh38
rs1021462210 Chr.2: 207077350-207077350 on Build GRCh38
rs900143829 Chr.2: 207077245-207077245 on Build GRCh38
rs746738145 Chr.2: 207077366-207077366 on Build GRCh38
rs1022539848 Chr.2: 207077274-207077274 on Build GRCh38
rs896621119 Chr.2: 207077606-207077606 on Build GRCh38
rs1040132952 Chr.2: 207077498-207077498 on Build GRCh38
rs1049018744 Chr.2: 207077494-207077494 on Build GRCh38
rs1043358603 Chr.2: 207077525-207077525 on Build GRCh38
rs997627412 Chr.2: 207077545-207077545 on Build GRCh38
rs544140140 Chr.2: 207077359-207077359 on Build GRCh38
rs1023651738 Chr.2: 207077409-207077409 on Build GRCh38
rs920534892 Chr.2: 207077441-207077441 on Build GRCh38
rs938705929 Chr.2: 207077435-207077435 on Build GRCh38
rs59137430 Chr.2: 207077540-207077540 on Build GRCh38
rs924601001 Chr.2: 207077370-207077370 on Build GRCh38
rs888993002 Chr.2: 207077574-207077574 on Build GRCh38
rs190421525 Chr.2: 207077241-207077241 on Build GRCh38
rs930565336 Chr.2: 207077444-207077444 on Build GRCh38
rs564701723 Chr.2: 207077353-207077353 on Build GRCh38
rs529865099 Chr.2: 207077489-207077489 on Build GRCh38
rs949003699 Chr.2: 207077486-207077486 on Build GRCh38
rs995787934 Chr.2: 207077254-207077254 on Build GRCh38
rs543764589 Chr.2: 207077190-207077190 on Build GRCh38
rs1014571775 Chr.2: 207077244-207077244 on Build GRCh38
rs773220547 Chr.2: 207077193-207077193 on Build GRCh38
rs770750128 Chr.2: 207077466-207077466 on Build GRCh38
rs986334731 Chr.2: 207077325-207077325 on Build GRCh38
rs181242243 Chr.2: 207077356-207077356 on Build GRCh38
rs560843633 Chr.2: 207077446-207077446 on Build GRCh38
rs139972370 Chr.2: 207077234-207077234 on Build GRCh38
rs968750392 Chr.2: 207077355-207077355 on Build GRCh38
rs961095271 Chr.2: 207077381-207077381 on Build GRCh38
rs186689773 Chr.2: 207077520-207077520 on Build GRCh38
rs904085094 Chr.2: 207077276-207077276 on Build GRCh38
rs1030704429 Chr.2: 207077304-207077304 on Build GRCh38
rs916180287 Chr.2: 207077458-207077458 on Build GRCh38
rs997417376 Chr.2: 207077280-207077280 on Build GRCh38
rs1030006806 Chr.2: 207077567-207077567 on Build GRCh38
rs900255456 Chr.2: 207077514-207077514 on Build GRCh38
rs888633323 Chr.2: 207077495-207077495 on Build GRCh38
rs17215792 Chr.2: 207077185-207077185 on Build GRCh38
rs992219580 Chr.2: 207077406-207077406 on Build GRCh38
rs969411534 Chr.2: 207077413-207077413 on Build GRCh38
rs546371793 Chr.2: 207077515-207077515 on Build GRCh38
rs747701565 Chr.2: 207077181-207077181 on Build GRCh38

View Full Product Details