Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 213003074-213003588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
UniGene
Hs.604950
Gene Chromosome Location
Chr.2: 212999684-213152753 on Build GRCh38
Transcripts
NM_001079526.1 NM_016260.2 XM_005246384.4 XM_005246385.3 XM_005246386.4 XM_011510802.2 XM_011510803.2 XM_011510804.2 XM_011510805.2 XM_011510807.2 XM_011510808.2 XM_011510809.2 XM_011510810.2 XM_011510811.2 XM_011510812.2 XM_011510814.2 XM_011510815.2 XM_011510816.2 XM_011510817.2 XM_011510818.2 XM_011510819.2 XM_017003589.1 XM_017003590.1 XM_017003591.1 XM_017003592.1

SNP Detail

SNP ID Chromosome Location
rs765594677 Chr.2: 213003147-213003147 on Build GRCh38
rs1017385348 Chr.2: 213003388-213003388 on Build GRCh38
rs535637321 Chr.2: 213003450-213003450 on Build GRCh38
rs568207317 Chr.2: 213003292-213003292 on Build GRCh38
rs556982446 Chr.2: 213003454-213003454 on Build GRCh38
rs976318856 Chr.2: 213003412-213003412 on Build GRCh38
rs914015446 Chr.2: 213003416-213003416 on Build GRCh38
rs899884282 Chr.2: 213003455-213003455 on Build GRCh38
rs575573498 Chr.2: 213003480-213003480 on Build GRCh38
rs966180826 Chr.2: 213003403-213003403 on Build GRCh38
rs540846419 Chr.2: 213003232-213003232 on Build GRCh38
rs891300664 Chr.2: 213003497-213003497 on Build GRCh38
rs866150211 Chr.2: 213003316-213003316 on Build GRCh38
rs777274827 Chr.2: 213003481-213003481 on Build GRCh38
rs1055035775 Chr.2: 213003532-213003532 on Build GRCh38
rs1007216135 Chr.2: 213003345-213003345 on Build GRCh38
rs925303880 Chr.2: 213003486-213003486 on Build GRCh38
rs1038411689 Chr.2: 213003362-213003362 on Build GRCh38
rs539860878 Chr.2: 213003542-213003542 on Build GRCh38
rs1047407403 Chr.2: 213003314-213003314 on Build GRCh38
rs951670046 Chr.2: 213003318-213003318 on Build GRCh38
rs896543768 Chr.2: 213003159-213003159 on Build GRCh38
rs1009762859 Chr.2: 213003521-213003521 on Build GRCh38
rs904148458 Chr.2: 213003562-213003562 on Build GRCh38
rs1045966432 Chr.2: 213003217-213003217 on Build GRCh38
rs758843642 Chr.2: 213003281-213003281 on Build GRCh38
rs978437942 Chr.2: 213003172-213003172 on Build GRCh38
rs866708822 Chr.2: 213003540-213003540 on Build GRCh38
rs1010250966 Chr.2: 213003103-213003103 on Build GRCh38
rs1021236763 Chr.2: 213003140-213003140 on Build GRCh38
rs1014116088 Chr.2: 213003203-213003203 on Build GRCh38
rs925696808 Chr.2: 213003181-213003181 on Build GRCh38
rs941337615 Chr.2: 213003358-213003358 on Build GRCh38
rs140202223 Chr.2: 213003262-213003262 on Build GRCh38
rs866587939 Chr.2: 213003202-213003202 on Build GRCh38
rs958458942 Chr.2: 213003191-213003191 on Build GRCh38

View Full Product Details