Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218638215-218638714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF142
Entrez Gene ID
7701
Gene Name
zinc finger protein 142
Gene Aliases
pHZ-49
UniGene
Hs.657969
Gene Chromosome Location
Chr.2: 218637592-218659632 on Build GRCh38
Transcripts
NM_001105537.2 NR_073599.1 NR_073600.1 XM_006712730.3 XM_011511784.2 XM_011511785.2 XM_011511786.2 XM_011511787.2 XM_011511788.2 XM_011511789.2 XM_017004871.1 XM_017004872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016412 c.4799G>A p.R1600H Chr.2: 218638604-218638604 on Build GRCh38
6416898 c.4310G>T p.R1437L Chr.2: 218638604-218638604 on Build GRCh38
4133455 c.4459C>A p.P1487T Chr.2: 218638455-218638455 on Build GRCh38
3043664 c.4689G>T p.R1563R Chr.2: 218638714-218638714 on Build GRCh38
1271029 c.4796A>G p.H1599R Chr.2: 218638607-218638607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774418402 Chr.2: 218638380-218638380 on Build GRCh38
rs780208929 Chr.2: 218638566-218638566 on Build GRCh38
rs368249612 Chr.2: 218638478-218638478 on Build GRCh38
rs748842115 Chr.2: 218638527-218638527 on Build GRCh38
rs200270476 Chr.2: 218638429-218638429 on Build GRCh38
rs760564661 Chr.2: 218638654-218638654 on Build GRCh38
rs754873030 Chr.2: 218638309-218638309 on Build GRCh38
rs774221961 Chr.2: 218638578-218638578 on Build GRCh38
rs749859946 Chr.2: 218638463-218638463 on Build GRCh38
rs997089262 Chr.2: 218638601-218638601 on Build GRCh38
rs371270102 Chr.2: 218638613-218638613 on Build GRCh38
rs767470620 Chr.2: 218638585-218638585 on Build GRCh38
rs200894546 Chr.2: 218638604-218638604 on Build GRCh38
rs917147736 Chr.2: 218638447-218638447 on Build GRCh38
rs551096799 Chr.2: 218638399-218638399 on Build GRCh38
rs372192856 Chr.2: 218638518-218638518 on Build GRCh38
rs952819157 Chr.2: 218638357-218638357 on Build GRCh38
rs766136700 Chr.2: 218638596-218638596 on Build GRCh38
rs773078488 Chr.2: 218638651-218638651 on Build GRCh38
rs753180361 Chr.2: 218638607-218638607 on Build GRCh38
rs769805203 Chr.2: 218638636-218638636 on Build GRCh38
rs555746746 Chr.2: 218638410-218638410 on Build GRCh38
rs758738288 Chr.2: 218638612-218638612 on Build GRCh38
rs779663339 Chr.2: 218638523-218638523 on Build GRCh38
rs766395618 Chr.2: 218638308-218638308 on Build GRCh38
rs754322118 Chr.2: 218638465-218638465 on Build GRCh38
rs952934120 Chr.2: 218638377-218638377 on Build GRCh38
rs753335955 Chr.2: 218638603-218638603 on Build GRCh38
rs767687799 Chr.2: 218638301-218638301 on Build GRCh38
rs367737510 Chr.2: 218638554-218638554 on Build GRCh38
rs776057640 Chr.2: 218638280-218638280 on Build GRCh38
rs750264792 Chr.2: 218638454-218638454 on Build GRCh38
rs778828682 Chr.2: 218638314-218638314 on Build GRCh38
rs556339609 Chr.2: 218638555-218638555 on Build GRCh38
rs376838139 Chr.2: 218638551-218638551 on Build GRCh38
rs774741989 Chr.2: 218638473-218638473 on Build GRCh38
rs377006217 Chr.2: 218638676-218638676 on Build GRCh38
rs767847255 Chr.2: 218638257-218638257 on Build GRCh38
rs763998037 Chr.2: 218638428-218638428 on Build GRCh38
rs373369883 Chr.2: 218638605-218638605 on Build GRCh38
rs745358698 Chr.2: 218638460-218638460 on Build GRCh38
rs776199147 Chr.2: 218638359-218638359 on Build GRCh38
rs767276190 Chr.2: 218638510-218638510 on Build GRCh38
rs771163019 Chr.2: 218638564-218638564 on Build GRCh38
rs757676073 Chr.2: 218638678-218638678 on Build GRCh38
rs780703499 Chr.2: 218638338-218638338 on Build GRCh38
rs766120334 Chr.2: 218638657-218638657 on Build GRCh38
rs780711019 Chr.2: 218638459-218638459 on Build GRCh38
rs771072813 Chr.2: 218638617-218638617 on Build GRCh38
rs750266764 Chr.2: 218638587-218638587 on Build GRCh38
rs770325210 Chr.2: 218638337-218638337 on Build GRCh38
rs758344926 Chr.2: 218638536-218638536 on Build GRCh38
rs780675796 Chr.2: 218638462-218638462 on Build GRCh38
rs759539156 Chr.2: 218638568-218638568 on Build GRCh38
rs755448262 Chr.2: 218638533-218638533 on Build GRCh38
rs201658171 Chr.2: 218638497-218638497 on Build GRCh38
rs752812115 Chr.2: 218638618-218638619 on Build GRCh38
rs901335362 Chr.2: 218638337-218638338 on Build GRCh38
rs199685898 Chr.2: 218638366-218638366 on Build GRCh38
rs755960222 Chr.2: 218638321-218638321 on Build GRCh38
rs375239015 Chr.2: 218638615-218638615 on Build GRCh38
rs761137416 Chr.2: 218638340-218638340 on Build GRCh38
rs755664207 Chr.2: 218638517-218638517 on Build GRCh38
rs370013089 Chr.2: 218638595-218638595 on Build GRCh38
rs971765263 Chr.2: 218638450-218638450 on Build GRCh38
rs750142144 Chr.2: 218638512-218638512 on Build GRCh38
rs746125635 Chr.2: 218638685-218638685 on Build GRCh38
rs375904295 Chr.2: 218638582-218638582 on Build GRCh38
rs1028289196 Chr.2: 218638311-218638311 on Build GRCh38
rs193157475 Chr.2: 218638365-218638365 on Build GRCh38
rs757015243 Chr.2: 218638502-218638502 on Build GRCh38
rs778255912 Chr.2: 218638675-218638675 on Build GRCh38
rs760754006 Chr.2: 218638393-218638393 on Build GRCh38
rs758861181 Chr.2: 218638666-218638666 on Build GRCh38
rs1041201492 Chr.2: 218638574-218638574 on Build GRCh38
rs752271205 Chr.2: 218638664-218638664 on Build GRCh38
rs772282172 Chr.2: 218638481-218638481 on Build GRCh38
rs1803383 Chr.2: 218638390-218638390 on Build GRCh38
rs3915806 Chr.2: 218638419-218638419 on Build GRCh38
rs752428952 Chr.2: 218638318-218638318 on Build GRCh38
rs997549214 Chr.2: 218638362-218638362 on Build GRCh38
rs761768678 Chr.2: 218638288-218638288 on Build GRCh38
rs758244416 Chr.2: 218638319-218638319 on Build GRCh38
rs751300245 Chr.2: 218638500-218638500 on Build GRCh38
rs557844517 Chr.2: 218638242-218638242 on Build GRCh38
rs903285561 Chr.2: 218638310-218638310 on Build GRCh38
rs762050305 Chr.2: 218638381-218638381 on Build GRCh38
rs201648820 Chr.2: 218638458-218638458 on Build GRCh38
rs746710790 Chr.2: 218638322-218638322 on Build GRCh38
rs556643601 Chr.2: 218638482-218638482 on Build GRCh38
rs748529642 Chr.2: 218638552-218638552 on Build GRCh38
rs775379914 Chr.2: 218638486-218638486 on Build GRCh38
rs928504409 Chr.2: 218638470-218638470 on Build GRCh38
rs866605255 Chr.2: 218638608-218638608 on Build GRCh38
rs761639163 Chr.2: 218638583-218638583 on Build GRCh38
rs374211078 Chr.2: 218638646-218638646 on Build GRCh38
rs979727406 Chr.2: 218638479-218638479 on Build GRCh38
rs370225990 Chr.2: 218638662-218638662 on Build GRCh38
rs747278562 Chr.2: 218638559-218638559 on Build GRCh38
rs769070195 Chr.2: 218638471-218638471 on Build GRCh38
rs188734714 Chr.2: 218638649-218638649 on Build GRCh38
rs762898553 Chr.2: 218638487-218638487 on Build GRCh38
rs183954984 Chr.2: 218638635-218638635 on Build GRCh38
rs368702779 Chr.2: 218638305-218638305 on Build GRCh38
rs372437071 Chr.2: 218638363-218638363 on Build GRCh38
rs372969531 Chr.2: 218638673-218638673 on Build GRCh38
rs770357064 Chr.2: 218638345-218638345 on Build GRCh38
rs371679801 Chr.2: 218638577-218638577 on Build GRCh38
rs776650467 Chr.2: 218638566-218638566 on Build GRCh38
rs559784734 Chr.2: 218638661-218638661 on Build GRCh38
rs758093986 Chr.2: 218638421-218638421 on Build GRCh38
rs781446620 Chr.2: 218638684-218638684 on Build GRCh38
rs200133906 Chr.2: 218638625-218638625 on Build GRCh38
rs947377737 Chr.2: 218638240-218638240 on Build GRCh38

View Full Product Details