Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219234053-219234565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKZF1
Entrez Gene ID
55139
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
ZNF744
UniGene
Hs.437647
Gene Chromosome Location
Chr.2: 219229757-219236669 on Build GRCh38
Transcripts
NM_001042410.1 NM_001282792.1 NM_018089.2 XM_005246663.2 XM_011511392.2 XM_017004420.1 XM_017004421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5990818 c.1099A>G p.K367E Chr.2: 219234183-219234183 on Build GRCh38
5990819 c.1144A>G p.R382G Chr.2: 219234228-219234228 on Build GRCh38
4461131 c.1193C>T p.S398F Chr.2: 219234277-219234277 on Build GRCh38
3838650 c.1049-1G>A p.? Chr.2: 219234132-219234132 on Build GRCh38
6999409 c.1204+5G>C p.? Chr.2: 219234293-219234293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780469403 Chr.2: 219234245-219234245 on Build GRCh38
rs761220756 Chr.2: 219234345-219234345 on Build GRCh38
rs769601581 Chr.2: 219234293-219234293 on Build GRCh38
rs928268061 Chr.2: 219234400-219234400 on Build GRCh38
rs756514072 Chr.2: 219234240-219234240 on Build GRCh38
rs139977011 Chr.2: 219234082-219234082 on Build GRCh38
rs373225556 Chr.2: 219234282-219234282 on Build GRCh38
rs536678002 Chr.2: 219234414-219234414 on Build GRCh38
rs537274222 Chr.2: 219234466-219234466 on Build GRCh38
rs768176164 Chr.2: 219234355-219234355 on Build GRCh38
rs775103160 Chr.2: 219234196-219234196 on Build GRCh38
rs753935310 Chr.2: 219234091-219234093 on Build GRCh38
rs749417107 Chr.2: 219234179-219234179 on Build GRCh38
rs1010963956 Chr.2: 219234189-219234189 on Build GRCh38
rs780416274 Chr.2: 219234176-219234176 on Build GRCh38
rs377499778 Chr.2: 219234256-219234256 on Build GRCh38
rs750881588 Chr.2: 219234239-219234239 on Build GRCh38
rs550921748 Chr.2: 219234390-219234390 on Build GRCh38
rs1024021158 Chr.2: 219234462-219234462 on Build GRCh38
rs750553376 Chr.2: 219234191-219234192 on Build GRCh38
rs1014013527 Chr.2: 219234419-219234419 on Build GRCh38
rs747362901 Chr.2: 219234158-219234158 on Build GRCh38
rs758651743 Chr.2: 219234227-219234230 on Build GRCh38
rs375422658 Chr.2: 219234135-219234135 on Build GRCh38
rs139055832 Chr.2: 219234406-219234406 on Build GRCh38
rs554415816 Chr.2: 219234495-219234495 on Build GRCh38
rs1007360744 Chr.2: 219234314-219234314 on Build GRCh38
rs1051161722 Chr.2: 219234261-219234261 on Build GRCh38
rs757669034 Chr.2: 219234161-219234161 on Build GRCh38
rs532543952 Chr.2: 219234142-219234142 on Build GRCh38
rs780253015 Chr.2: 219234147-219234147 on Build GRCh38
rs548458187 Chr.2: 219234366-219234366 on Build GRCh38
rs775228175 Chr.2: 219234297-219234297 on Build GRCh38
rs202032166 Chr.2: 219234164-219234164 on Build GRCh38
rs368810674 Chr.2: 219234273-219234273 on Build GRCh38
rs1053019834 Chr.2: 219234225-219234225 on Build GRCh38
rs764559758 Chr.2: 219234144-219234144 on Build GRCh38
rs963633507 Chr.2: 219234094-219234094 on Build GRCh38
rs755239892 Chr.2: 219234387-219234387 on Build GRCh38
rs150725143 Chr.2: 219234137-219234137 on Build GRCh38
rs761213285 Chr.2: 219234358-219234358 on Build GRCh38
rs768121043 Chr.2: 219234238-219234238 on Build GRCh38
rs762467569 Chr.2: 219234198-219234198 on Build GRCh38
rs752021745 Chr.2: 219234387-219234388 on Build GRCh38
rs899059792 Chr.2: 219234394-219234394 on Build GRCh38
rs752862067 Chr.2: 219234402-219234402 on Build GRCh38
rs746173907 Chr.2: 219234167-219234167 on Build GRCh38
rs773971309 Chr.2: 219234453-219234453 on Build GRCh38
rs372924586 Chr.2: 219234243-219234243 on Build GRCh38
rs938245477 Chr.2: 219234403-219234403 on Build GRCh38
rs773734066 Chr.2: 219234230-219234230 on Build GRCh38
rs760434575 Chr.2: 219234123-219234123 on Build GRCh38
rs780279587 Chr.2: 219234227-219234229 on Build GRCh38
rs1042453575 Chr.2: 219234511-219234511 on Build GRCh38
rs201470378 Chr.2: 219234187-219234187 on Build GRCh38
rs754134208 Chr.2: 219234252-219234252 on Build GRCh38
rs115949101 Chr.2: 219234191-219234191 on Build GRCh38
rs113233400 Chr.2: 219234193-219234193 on Build GRCh38
rs764856870 Chr.2: 219234138-219234138 on Build GRCh38
rs773167441 Chr.2: 219234110-219234110 on Build GRCh38
rs752132261 Chr.2: 219234145-219234145 on Build GRCh38
rs748954117 Chr.2: 219234269-219234269 on Build GRCh38
rs567529723 Chr.2: 219234398-219234398 on Build GRCh38
rs754084597 Chr.2: 219234378-219234378 on Build GRCh38
rs774244687 Chr.2: 219234184-219234184 on Build GRCh38
rs199537659 Chr.2: 219234185-219234185 on Build GRCh38
rs752271664 Chr.2: 219234139-219234139 on Build GRCh38
rs980269243 Chr.2: 219234486-219234486 on Build GRCh38
rs972487320 Chr.2: 219234318-219234318 on Build GRCh38
rs779054428 Chr.2: 219234257-219234257 on Build GRCh38
rs765379929 Chr.2: 219234122-219234124 on Build GRCh38
rs201952175 Chr.2: 219234181-219234181 on Build GRCh38
rs367760517 Chr.2: 219234098-219234098 on Build GRCh38
rs745714073 Chr.2: 219234289-219234289 on Build GRCh38
rs749361982 Chr.2: 219234083-219234083 on Build GRCh38
rs902603566 Chr.2: 219234512-219234512 on Build GRCh38
rs934059340 Chr.2: 219234258-219234258 on Build GRCh38
rs879235737 Chr.2: 219234272-219234272 on Build GRCh38
rs998230443 Chr.2: 219234515-219234515 on Build GRCh38
rs189474878 Chr.2: 219234540-219234540 on Build GRCh38
rs970212875 Chr.2: 219234484-219234484 on Build GRCh38
rs761944759 Chr.2: 219234115-219234119 on Build GRCh38
rs776469050 Chr.2: 219234188-219234188 on Build GRCh38
rs926125813 Chr.2: 219234492-219234492 on Build GRCh38
rs531124156 Chr.2: 219234221-219234221 on Build GRCh38
rs377760043 Chr.2: 219234300-219234300 on Build GRCh38
rs918303593 Chr.2: 219234340-219234340 on Build GRCh38
rs763692863 Chr.2: 219234204-219234204 on Build GRCh38
rs966689873 Chr.2: 219234202-219234202 on Build GRCh38
rs756458786 Chr.2: 219234169-219234169 on Build GRCh38
rs773965398 Chr.2: 219234311-219234311 on Build GRCh38
rs777782911 Chr.2: 219234410-219234410 on Build GRCh38
rs750940639 Chr.2: 219234357-219234357 on Build GRCh38
rs141815102 Chr.2: 219234127-219234127 on Build GRCh38
rs528605919 Chr.2: 219234207-219234207 on Build GRCh38

View Full Product Details