Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219249126-219249637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK16
Entrez Gene ID
8576
Gene Name
serine/threonine kinase 16
Gene Aliases
KRCT, MPSK, PKL12, TSF1
UniGene
Hs.153003
Gene Chromosome Location
Chr.2: 219245469-219250337 on Build GRCh38
Transcripts
NM_001008910.2 NR_026909.1 XM_011512067.2

SNP Detail

SNP ID Chromosome Location
rs975572736 Chr.2: 219249606-219249606 on Build GRCh38
rs997068764 Chr.2: 219249371-219249371 on Build GRCh38
rs928532521 Chr.2: 219249151-219249151 on Build GRCh38
rs915843639 Chr.2: 219249557-219249557 on Build GRCh38
rs1029000191 Chr.2: 219249385-219249386 on Build GRCh38
rs931375299 Chr.2: 219249350-219249350 on Build GRCh38
rs1011256158 Chr.2: 219249279-219249279 on Build GRCh38
rs369861070 Chr.2: 219249293-219249293 on Build GRCh38
rs779604905 Chr.2: 219249336-219249336 on Build GRCh38
rs887075432 Chr.2: 219249405-219249405 on Build GRCh38
rs961471692 Chr.2: 219249454-219249454 on Build GRCh38
rs769646195 Chr.2: 219249520-219249520 on Build GRCh38
rs1031724564 Chr.2: 219249263-219249263 on Build GRCh38
rs1020271166 Chr.2: 219249587-219249587 on Build GRCh38
rs563920060 Chr.2: 219249596-219249596 on Build GRCh38
rs944194868 Chr.2: 219249331-219249331 on Build GRCh38
rs781222978 Chr.2: 219249418-219249418 on Build GRCh38
rs757837337 Chr.2: 219249318-219249318 on Build GRCh38
rs75923512 Chr.2: 219249347-219249347 on Build GRCh38
rs750088356 Chr.2: 219249296-219249296 on Build GRCh38
rs894108802 Chr.2: 219249264-219249264 on Build GRCh38
rs988758797 Chr.2: 219249314-219249314 on Build GRCh38
rs1057420000 Chr.2: 219249417-219249417 on Build GRCh38
rs904285178 Chr.2: 219249475-219249475 on Build GRCh38
rs540924426 Chr.2: 219249297-219249297 on Build GRCh38
rs558833993 Chr.2: 219249349-219249349 on Build GRCh38
rs114934283 Chr.2: 219249348-219249348 on Build GRCh38
rs75665317 Chr.2: 219249150-219249150 on Build GRCh38
rs191609682 Chr.2: 219249443-219249443 on Build GRCh38
rs991388262 Chr.2: 219249530-219249530 on Build GRCh38
rs558314884 Chr.2: 219249162-219249162 on Build GRCh38
rs968402227 Chr.2: 219249179-219249179 on Build GRCh38
rs1031337696 Chr.2: 219249527-219249527 on Build GRCh38
rs370333843 Chr.2: 219249208-219249208 on Build GRCh38
rs1048397529 Chr.2: 219249386-219249386 on Build GRCh38
rs547193420 Chr.2: 219249472-219249472 on Build GRCh38
rs972302522 Chr.2: 219249480-219249480 on Build GRCh38
rs928527484 Chr.2: 219249496-219249496 on Build GRCh38
rs533007300 Chr.2: 219249601-219249601 on Build GRCh38
rs571010105 Chr.2: 219249579-219249579 on Build GRCh38
rs1012742925 Chr.2: 219249423-219249423 on Build GRCh38
rs372925835 Chr.2: 219249584-219249584 on Build GRCh38
rs999798823 Chr.2: 219249582-219249582 on Build GRCh38
rs1042771824 Chr.2: 219249573-219249573 on Build GRCh38
rs140893994 Chr.2: 219249567-219249567 on Build GRCh38
rs1012711665 Chr.2: 219249158-219249158 on Build GRCh38
rs934465666 Chr.2: 219249586-219249586 on Build GRCh38
rs1055867083 Chr.2: 219249232-219249232 on Build GRCh38
rs578231671 Chr.2: 219249207-219249207 on Build GRCh38
rs148024515 Chr.2: 219249410-219249410 on Build GRCh38
rs965539627 Chr.2: 219249598-219249598 on Build GRCh38

View Full Product Details