Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219553255-219553780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
UniGene
Hs.526594
Gene Chromosome Location
Chr.2: 219549408-219571573 on Build GRCh38
Transcripts
NM_015311.2 XM_005246424.4 XM_011510857.2 XM_011510863.2 XM_017003696.1 XM_017003697.1 XM_017003698.1 XM_017003699.1 XM_017003700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6732736 c.4968G>A p.L1656L Chr.2: 219553595-219553595 on Build GRCh38
6732737 c.4692G>A p.L1564L Chr.2: 219553595-219553595 on Build GRCh38
6217466 c.4621C>T p.R1541W Chr.2: 219553666-219553666 on Build GRCh38
4781269 c.4675G>A p.E1559K Chr.2: 219553612-219553612 on Build GRCh38
4091753 c.4894G>A p.V1632M Chr.2: 219553669-219553669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771958357 Chr.2: 219553713-219553713 on Build GRCh38
rs962417010 Chr.2: 219553513-219553513 on Build GRCh38
rs201511204 Chr.2: 219553520-219553520 on Build GRCh38
rs777508328 Chr.2: 219553438-219553438 on Build GRCh38
rs769071777 Chr.2: 219553427-219553427 on Build GRCh38
rs770524599 Chr.2: 219553624-219553624 on Build GRCh38
rs752000673 Chr.2: 219553598-219553598 on Build GRCh38
rs776475820 Chr.2: 219553672-219553672 on Build GRCh38
rs754358473 Chr.2: 219553715-219553715 on Build GRCh38
rs969569691 Chr.2: 219553728-219553728 on Build GRCh38
rs2555928 Chr.2: 219553389-219553389 on Build GRCh38
rs576820480 Chr.2: 219553619-219553619 on Build GRCh38
rs766884484 Chr.2: 219553706-219553706 on Build GRCh38
rs551362911 Chr.2: 219553324-219553324 on Build GRCh38
rs760670486 Chr.2: 219553626-219553626 on Build GRCh38
rs935710047 Chr.2: 219553616-219553616 on Build GRCh38
rs765428655 Chr.2: 219553573-219553573 on Build GRCh38
rs35252057 Chr.2: 219553688-219553689 on Build GRCh38
rs866308700 Chr.2: 219553569-219553569 on Build GRCh38
rs1006242538 Chr.2: 219553673-219553673 on Build GRCh38
rs775664438 Chr.2: 219553629-219553629 on Build GRCh38
rs540208891 Chr.2: 219553669-219553669 on Build GRCh38
rs997366169 Chr.2: 219553514-219553514 on Build GRCh38
rs754529663 Chr.2: 219553655-219553655 on Build GRCh38
rs866863146 Chr.2: 219553642-219553642 on Build GRCh38
rs764047202 Chr.2: 219553709-219553710 on Build GRCh38
rs756666231 Chr.2: 219553648-219553648 on Build GRCh38
rs765176936 Chr.2: 219553627-219553627 on Build GRCh38
rs903317350 Chr.2: 219553427-219553432 on Build GRCh38
rs897920873 Chr.2: 219553698-219553698 on Build GRCh38
rs533921662 Chr.2: 219553463-219553463 on Build GRCh38
rs1015035912 Chr.2: 219553477-219553477 on Build GRCh38
rs1055447155 Chr.2: 219553329-219553329 on Build GRCh38
rs370697325 Chr.2: 219553551-219553551 on Build GRCh38
rs759524921 Chr.2: 219553568-219553568 on Build GRCh38
rs571353513 Chr.2: 219553462-219553462 on Build GRCh38
rs763048668 Chr.2: 219553631-219553631 on Build GRCh38
rs200575344 Chr.2: 219553597-219553597 on Build GRCh38
rs1029315678 Chr.2: 219553339-219553339 on Build GRCh38
rs779389667 Chr.2: 219553652-219553652 on Build GRCh38
rs747400429 Chr.2: 219553380-219553380 on Build GRCh38
rs1041218740 Chr.2: 219553725-219553725 on Build GRCh38
rs774155373 Chr.2: 219553547-219553547 on Build GRCh38
rs774043639 Chr.2: 219553636-219553636 on Build GRCh38
rs894162480 Chr.2: 219553391-219553391 on Build GRCh38
rs760983839 Chr.2: 219553705-219553705 on Build GRCh38
rs199707413 Chr.2: 219553620-219553620 on Build GRCh38
rs992332228 Chr.2: 219553541-219553541 on Build GRCh38
rs140825693 Chr.2: 219553612-219553612 on Build GRCh38
rs773186372 Chr.2: 219553562-219553562 on Build GRCh38
rs747860748 Chr.2: 219553665-219553665 on Build GRCh38
rs556686718 Chr.2: 219553613-219553613 on Build GRCh38
rs901237039 Chr.2: 219553480-219553480 on Build GRCh38
rs188544 Chr.2: 219553651-219553651 on Build GRCh38
rs779628741 Chr.2: 219553618-219553618 on Build GRCh38
rs377217282 Chr.2: 219553693-219553693 on Build GRCh38
rs947657768 Chr.2: 219553412-219553412 on Build GRCh38
rs901769816 Chr.2: 219553295-219553295 on Build GRCh38
rs1019687198 Chr.2: 219553468-219553468 on Build GRCh38
rs763068097 Chr.2: 219553679-219553679 on Build GRCh38
rs752791867 Chr.2: 219553580-219553580 on Build GRCh38
rs531461092 Chr.2: 219553316-219553316 on Build GRCh38
rs754476402 Chr.2: 219553724-219553724 on Build GRCh38
rs191093745 Chr.2: 219553666-219553666 on Build GRCh38
rs367657651 Chr.2: 219553647-219553647 on Build GRCh38
rs753356436 Chr.2: 219553654-219553654 on Build GRCh38
rs1006614221 Chr.2: 219553423-219553423 on Build GRCh38
rs372856431 Chr.2: 219553661-219553661 on Build GRCh38
rs981061219 Chr.2: 219553587-219553587 on Build GRCh38
rs375870998 Chr.2: 219553622-219553622 on Build GRCh38
rs771909036 Chr.2: 219553623-219553623 on Build GRCh38
rs1040159118 Chr.2: 219553288-219553288 on Build GRCh38
rs762255736 Chr.2: 219553639-219553639 on Build GRCh38
rs192760 Chr.2: 219553502-219553502 on Build GRCh38
rs999037251 Chr.2: 219553458-219553458 on Build GRCh38
rs554007132 Chr.2: 219553519-219553519 on Build GRCh38
rs760511999 Chr.2: 219553574-219553576 on Build GRCh38
rs952116587 Chr.2: 219553417-219553417 on Build GRCh38
rs934923106 Chr.2: 219553718-219553718 on Build GRCh38
rs368068900 Chr.2: 219553615-219553615 on Build GRCh38
rs774710450 Chr.2: 219553701-219553701 on Build GRCh38
rs768061606 Chr.2: 219553640-219553640 on Build GRCh38
rs758657495 Chr.2: 219553595-219553595 on Build GRCh38
rs770935107 Chr.2: 219553670-219553670 on Build GRCh38
rs764382772 Chr.2: 219553634-219553634 on Build GRCh38
rs750829684 Chr.2: 219553702-219553702 on Build GRCh38

View Full Product Details