Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222918939-222919489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL3
Entrez Gene ID
2181
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
ACS3, FACL3, PRO2194
UniGene
Hs.655772
Gene Chromosome Location
Chr.2: 222860984-222944624 on Build GRCh38
Transcripts
NM_004457.3 NM_203372.1 XM_017003584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1163236 c.674T>C p.V225A Chr.2: 222919071-222919071 on Build GRCh38
1565037 c.705C>A p.I235I Chr.2: 222919102-222919102 on Build GRCh38
1017047 c.688C>T p.R230C Chr.2: 222919085-222919085 on Build GRCh38
4091910 c.672A>C p.I224I Chr.2: 222919069-222919069 on Build GRCh38
1017051 c.748G>A p.G250S Chr.2: 222919145-222919145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755506459 Chr.2: 222919206-222919206 on Build GRCh38
rs755869725 Chr.2: 222919088-222919088 on Build GRCh38
rs750603066 Chr.2: 222919169-222919169 on Build GRCh38
rs769587657 Chr.2: 222919228-222919228 on Build GRCh38
rs974996311 Chr.2: 222919339-222919339 on Build GRCh38
rs145194965 Chr.2: 222919122-222919122 on Build GRCh38
rs775355400 Chr.2: 222919229-222919229 on Build GRCh38
rs776335717 Chr.2: 222919142-222919142 on Build GRCh38
rs570587981 Chr.2: 222919224-222919224 on Build GRCh38
rs746055510 Chr.2: 222919222-222919222 on Build GRCh38
rs546599601 Chr.2: 222919348-222919348 on Build GRCh38
rs990015987 Chr.2: 222919292-222919292 on Build GRCh38
rs146239364 Chr.2: 222919163-222919163 on Build GRCh38
rs964473623 Chr.2: 222919330-222919330 on Build GRCh38
rs761822696 Chr.2: 222919161-222919161 on Build GRCh38
rs760270998 Chr.2: 222919130-222919130 on Build GRCh38
rs2119157 Chr.2: 222919350-222919350 on Build GRCh38
rs933383705 Chr.2: 222919353-222919353 on Build GRCh38
rs776478456 Chr.2: 222919219-222919219 on Build GRCh38
rs981531534 Chr.2: 222918990-222918990 on Build GRCh38
rs760482853 Chr.2: 222919056-222919056 on Build GRCh38
rs777467919 Chr.2: 222919117-222919117 on Build GRCh38
rs371178618 Chr.2: 222919158-222919158 on Build GRCh38
rs143351672 Chr.2: 222919091-222919091 on Build GRCh38
rs139163050 Chr.2: 222919199-222919199 on Build GRCh38
rs779091229 Chr.2: 222919032-222919035 on Build GRCh38
rs773774619 Chr.2: 222919038-222919038 on Build GRCh38
rs372421754 Chr.2: 222919047-222919047 on Build GRCh38
rs13000358 Chr.2: 222919123-222919123 on Build GRCh38
rs771663123 Chr.2: 222919216-222919216 on Build GRCh38
rs561778631 Chr.2: 222919197-222919197 on Build GRCh38
rs914401418 Chr.2: 222919067-222919067 on Build GRCh38
rs911776768 Chr.2: 222919035-222919035 on Build GRCh38
rs914495867 Chr.2: 222919325-222919325 on Build GRCh38
rs776631918 Chr.2: 222919039-222919039 on Build GRCh38
rs772411586 Chr.2: 222919290-222919290 on Build GRCh38
rs564645237 Chr.2: 222919226-222919226 on Build GRCh38
rs57570971 Chr.2: 222919423-222919423 on Build GRCh38
rs768172715 Chr.2: 222919036-222919036 on Build GRCh38
rs934564815 Chr.2: 222919043-222919043 on Build GRCh38
rs748604442 Chr.2: 222919086-222919086 on Build GRCh38
rs534705466 Chr.2: 222919406-222919406 on Build GRCh38
rs763347033 Chr.2: 222919394-222919394 on Build GRCh38
rs776007640 Chr.2: 222919013-222919013 on Build GRCh38
rs181406206 Chr.2: 222919262-222919262 on Build GRCh38
rs946044170 Chr.2: 222919002-222919002 on Build GRCh38
rs200213683 Chr.2: 222919213-222919213 on Build GRCh38
rs267599218 Chr.2: 222919194-222919194 on Build GRCh38
rs376738660 Chr.2: 222919092-222919092 on Build GRCh38
rs375926923 Chr.2: 222919232-222919232 on Build GRCh38
rs967789077 Chr.2: 222919291-222919291 on Build GRCh38
rs990048926 Chr.2: 222919044-222919044 on Build GRCh38
rs764207762 Chr.2: 222919085-222919085 on Build GRCh38
rs749780783 Chr.2: 222919028-222919028 on Build GRCh38
rs759428756 Chr.2: 222919081-222919081 on Build GRCh38
rs886465714 Chr.2: 222919112-222919112 on Build GRCh38
rs558677329 Chr.2: 222919285-222919285 on Build GRCh38
rs144556995 Chr.2: 222919191-222919191 on Build GRCh38
rs759501803 Chr.2: 222919143-222919143 on Build GRCh38
rs770724586 Chr.2: 222919133-222919133 on Build GRCh38
rs767553888 Chr.2: 222919237-222919237 on Build GRCh38
rs750655474 Chr.2: 222919089-222919089 on Build GRCh38
rs149904851 Chr.2: 222919446-222919446 on Build GRCh38
rs370080501 Chr.2: 222919031-222919031 on Build GRCh38
rs769094747 Chr.2: 222919105-222919105 on Build GRCh38
rs907426877 Chr.2: 222919165-222919165 on Build GRCh38
rs879082398 Chr.2: 222919002-222919007 on Build GRCh38
rs771910418 Chr.2: 222919045-222919045 on Build GRCh38
rs765152453 Chr.2: 222919154-222919154 on Build GRCh38
rs766104754 Chr.2: 222919068-222919068 on Build GRCh38
rs746647106 Chr.2: 222919218-222919218 on Build GRCh38
rs777461277 Chr.2: 222919217-222919217 on Build GRCh38
rs753763561 Chr.2: 222919071-222919071 on Build GRCh38
rs1038881783 Chr.2: 222919079-222919079 on Build GRCh38
rs199621546 Chr.2: 222919113-222919113 on Build GRCh38
rs1047847187 Chr.2: 222919376-222919376 on Build GRCh38
rs546213707 Chr.2: 222919093-222919093 on Build GRCh38
rs778554171 Chr.2: 222919107-222919107 on Build GRCh38
rs761536442 Chr.2: 222919042-222919042 on Build GRCh38
rs986779508 Chr.2: 222919305-222919305 on Build GRCh38
rs752633642 Chr.2: 222919156-222919156 on Build GRCh38

View Full Product Details