Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231004061-231004601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA3
Entrez Gene ID
130560
Gene Name
spermatogenesis associated 3
Gene Aliases
TSARG1
UniGene
Hs.745178
Gene Chromosome Location
Chr.2: 230996124-231019939 on Build GRCh38
Transcripts
NM_139073.3 XM_005246303.2 XM_017003363.1

SNP Detail

SNP ID Chromosome Location
rs145142097 Chr.2: 231004487-231004487 on Build GRCh38
rs555636409 Chr.2: 231004365-231004365 on Build GRCh38
rs541013212 Chr.2: 231004484-231004484 on Build GRCh38
rs1032068696 Chr.2: 231004443-231004443 on Build GRCh38
rs147352327 Chr.2: 231004325-231004325 on Build GRCh38
rs998213318 Chr.2: 231004381-231004381 on Build GRCh38
rs917087752 Chr.2: 231004357-231004357 on Build GRCh38
rs11690743 Chr.2: 231004580-231004580 on Build GRCh38
rs527324604 Chr.2: 231004451-231004451 on Build GRCh38
rs1025101616 Chr.2: 231004464-231004464 on Build GRCh38
rs926499210 Chr.2: 231004520-231004520 on Build GRCh38
rs953542317 Chr.2: 231004191-231004191 on Build GRCh38
rs931101076 Chr.2: 231004299-231004299 on Build GRCh38
rs917571669 Chr.2: 231004221-231004221 on Build GRCh38
rs556943956 Chr.2: 231004454-231004455 on Build GRCh38
rs762412792 Chr.2: 231004522-231004522 on Build GRCh38
rs979152479 Chr.2: 231004198-231004198 on Build GRCh38
rs901837759 Chr.2: 231004346-231004346 on Build GRCh38
rs931153767 Chr.2: 231004579-231004579 on Build GRCh38
rs1005625780 Chr.2: 231004119-231004120 on Build GRCh38
rs3731784 Chr.2: 231004214-231004214 on Build GRCh38
rs1029607872 Chr.2: 231004178-231004178 on Build GRCh38
rs368907281 Chr.2: 231004102-231004102 on Build GRCh38
rs1017190526 Chr.2: 231004280-231004280 on Build GRCh38
rs997767529 Chr.2: 231004163-231004163 on Build GRCh38
rs776200374 Chr.2: 231004323-231004323 on Build GRCh38
rs3821002 Chr.2: 231004326-231004326 on Build GRCh38
rs1016134769 Chr.2: 231004157-231004157 on Build GRCh38
rs1000241669 Chr.2: 231004558-231004558 on Build GRCh38
rs139405615 Chr.2: 231004344-231004344 on Build GRCh38
rs949356905 Chr.2: 231004228-231004228 on Build GRCh38
rs748496807 Chr.2: 231004202-231004202 on Build GRCh38
rs1000957776 Chr.2: 231004427-231004427 on Build GRCh38
rs11679650 Chr.2: 231004417-231004417 on Build GRCh38
rs879648769 Chr.2: 231004201-231004201 on Build GRCh38
rs987895992 Chr.2: 231004459-231004459 on Build GRCh38
rs981104514 Chr.2: 231004503-231004503 on Build GRCh38
rs866747411 Chr.2: 231004488-231004488 on Build GRCh38
rs889289108 Chr.2: 231004420-231004420 on Build GRCh38
rs751778577 Chr.2: 231004496-231004496 on Build GRCh38
rs926445759 Chr.2: 231004287-231004287 on Build GRCh38
rs1022526501 Chr.2: 231004537-231004537 on Build GRCh38
rs1031353998 Chr.2: 231004577-231004577 on Build GRCh38
rs897304101 Chr.2: 231004193-231004193 on Build GRCh38
rs968824781 Chr.2: 231004288-231004288 on Build GRCh38
rs1051167177 Chr.2: 231004486-231004486 on Build GRCh38
rs373202453 Chr.2: 231004206-231004206 on Build GRCh38
rs968398472 Chr.2: 231004556-231004556 on Build GRCh38
rs376173293 Chr.2: 231004229-231004229 on Build GRCh38

View Full Product Details