Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231455928-231456433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCL
Entrez Gene ID
4691
Gene Name
nucleolin
Gene Aliases
C23
UniGene
Hs.79110
Gene Chromosome Location
Chr.2: 231454748-231464494 on Build GRCh38
Transcripts
NM_005381.2 NR_002921.1

Gene Symbol
SNORA75
Entrez Gene ID
654321
Gene Name
small nucleolar RNA, H/ACA box 75
Gene Aliases
SNORA75A, U23
UniGene
Hs.674388
Gene Chromosome Location
Chr.2: 231455800-231455936 on Build GRCh38
Transcripts
NM_005381.2 NR_002921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3909824 c.1713C>T p.S571S Chr.2: 231456129-231456129 on Build GRCh38
5973652 c.1786G>A p.V596I Chr.2: 231456056-231456056 on Build GRCh38
3579021 c.1712C>T p.S571F Chr.2: 231456130-231456130 on Build GRCh38
1482857 c.1789C>T p.R597W Chr.2: 231456053-231456053 on Build GRCh38
327970 c.1706-10_1706-9insT p.? Chr.2: 231456145-231456146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372849199 Chr.2: 231456405-231456405 on Build GRCh38
rs911994826 Chr.2: 231456404-231456404 on Build GRCh38
rs941099221 Chr.2: 231456272-231456272 on Build GRCh38
rs558008297 Chr.2: 231456025-231456025 on Build GRCh38
rs765578645 Chr.2: 231456212-231456212 on Build GRCh38
rs960511340 Chr.2: 231456342-231456342 on Build GRCh38
rs142532822 Chr.2: 231456108-231456108 on Build GRCh38
rs769736556 Chr.2: 231455994-231455994 on Build GRCh38
rs1010741139 Chr.2: 231456230-231456230 on Build GRCh38
rs896859104 Chr.2: 231455955-231455955 on Build GRCh38
rs560390229 Chr.2: 231456195-231456195 on Build GRCh38
rs774450051 Chr.2: 231456224-231456224 on Build GRCh38
rs1022536173 Chr.2: 231456394-231456394 on Build GRCh38
rs747845005 Chr.2: 231455971-231455971 on Build GRCh38
rs200390674 Chr.2: 231456144-231456144 on Build GRCh38
rs550005492 Chr.2: 231456337-231456337 on Build GRCh38
rs145953863 Chr.2: 231456379-231456379 on Build GRCh38
rs572406712 Chr.2: 231455961-231455961 on Build GRCh38
rs1020587575 Chr.2: 231456309-231456309 on Build GRCh38
rs368724845 Chr.2: 231456159-231456159 on Build GRCh38
rs919219827 Chr.2: 231456141-231456141 on Build GRCh38
rs546001499 Chr.2: 231456000-231456000 on Build GRCh38
rs902171401 Chr.2: 231456313-231456313 on Build GRCh38
rs765628776 Chr.2: 231456075-231456075 on Build GRCh38
rs748310037 Chr.2: 231456157-231456157 on Build GRCh38
rs189865329 Chr.2: 231456063-231456063 on Build GRCh38
rs766823824 Chr.2: 231456158-231456158 on Build GRCh38
rs374655131 Chr.2: 231456217-231456217 on Build GRCh38
rs756859655 Chr.2: 231456130-231456130 on Build GRCh38
rs757814072 Chr.2: 231455972-231455972 on Build GRCh38
rs1049672792 Chr.2: 231456237-231456237 on Build GRCh38
rs773333006 Chr.2: 231456168-231456168 on Build GRCh38
rs753039666 Chr.2: 231456400-231456400 on Build GRCh38
rs760095525 Chr.2: 231456397-231456397 on Build GRCh38
rs991667277 Chr.2: 231456351-231456351 on Build GRCh38
rs767494004 Chr.2: 231456004-231456004 on Build GRCh38
rs777407354 Chr.2: 231455975-231455975 on Build GRCh38
rs112840566 Chr.2: 231456137-231456137 on Build GRCh38
rs963624491 Chr.2: 231456369-231456369 on Build GRCh38
rs987989148 Chr.2: 231456403-231456403 on Build GRCh38
rs745852480 Chr.2: 231456149-231456149 on Build GRCh38
rs919284533 Chr.2: 231456372-231456372 on Build GRCh38
rs973617670 Chr.2: 231456370-231456370 on Build GRCh38
rs766488719 Chr.2: 231456180-231456180 on Build GRCh38
rs181415034 Chr.2: 231456239-231456239 on Build GRCh38
rs370933084 Chr.2: 231455985-231455985 on Build GRCh38
rs769684387 Chr.2: 231456155-231456155 on Build GRCh38
rs770844868 Chr.2: 231455980-231455980 on Build GRCh38
rs760771676 Chr.2: 231456179-231456179 on Build GRCh38
rs561881971 Chr.2: 231456362-231456362 on Build GRCh38
rs79523334 Chr.2: 231456233-231456233 on Build GRCh38
rs909410229 Chr.2: 231456260-231456260 on Build GRCh38
rs1038815678 Chr.2: 231456210-231456210 on Build GRCh38
rs374299439 Chr.2: 231456035-231456035 on Build GRCh38
rs568582393 Chr.2: 231456355-231456355 on Build GRCh38
rs923932492 Chr.2: 231456161-231456161 on Build GRCh38
rs59377947 Chr.2: 231456154-231456155 on Build GRCh38
rs527999919 Chr.2: 231456232-231456232 on Build GRCh38
rs916785208 Chr.2: 231456215-231456215 on Build GRCh38
rs960229822 Chr.2: 231455990-231455990 on Build GRCh38
rs1003741182 Chr.2: 231456316-231456316 on Build GRCh38
rs535467322 Chr.2: 231456365-231456365 on Build GRCh38
rs774633476 Chr.2: 231455981-231455981 on Build GRCh38
rs548339356 Chr.2: 231456306-231456306 on Build GRCh38
rs896912420 Chr.2: 231456320-231456320 on Build GRCh38
rs375337610 Chr.2: 231456143-231456143 on Build GRCh38
rs760671544 Chr.2: 231456033-231456033 on Build GRCh38
rs369902237 Chr.2: 231456145-231456145 on Build GRCh38
rs766828166 Chr.2: 231456056-231456056 on Build GRCh38
rs16828079 Chr.2: 231456087-231456087 on Build GRCh38
rs777303754 Chr.2: 231456101-231456101 on Build GRCh38
rs2595518 Chr.2: 231456171-231456171 on Build GRCh38
rs746907631 Chr.2: 231455978-231455978 on Build GRCh38
rs1034754641 Chr.2: 231456322-231456322 on Build GRCh38
rs760778155 Chr.2: 231456124-231456124 on Build GRCh38
rs769308610 Chr.2: 231456146-231456146 on Build GRCh38
rs755477918 Chr.2: 231455994-231455994 on Build GRCh38
rs149759354 Chr.2: 231456078-231456078 on Build GRCh38
rs755364611 Chr.2: 231455953-231455953 on Build GRCh38
rs113883398 Chr.2: 231456083-231456083 on Build GRCh38
rs142355913 Chr.2: 231456299-231456299 on Build GRCh38
rs985667401 Chr.2: 231455995-231455995 on Build GRCh38
rs556504062 Chr.2: 231456280-231456280 on Build GRCh38
rs150234462 Chr.2: 231456145-231456146 on Build GRCh38
rs201866212 Chr.2: 231456181-231456181 on Build GRCh38
rs774695825 Chr.2: 231455996-231455996 on Build GRCh38
rs868722182 Chr.2: 231456058-231456058 on Build GRCh38
rs139786193 Chr.2: 231456057-231456057 on Build GRCh38
rs765581969 Chr.2: 231456399-231456399 on Build GRCh38
rs775495867 Chr.2: 231456156-231456156 on Build GRCh38
rs753117391 Chr.2: 231456249-231456249 on Build GRCh38

View Full Product Details