Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237487019-237487511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLPH
Entrez Gene ID
79083
Gene Name
melanophilin
Gene Aliases
SLAC2-A
UniGene
Hs.102406
Gene Chromosome Location
Chr.2: 237486410-237555318 on Build GRCh38
Transcripts
NM_001042467.2 NM_001281473.1 NM_001281474.1 NM_024101.6 NR_104019.1 XM_006712737.1 XM_006712739.1 XM_006712740.1 XM_017004893.1 XM_017004894.1

SNP Detail

SNP ID Chromosome Location
rs188472124 Chr.2: 237487357-237487357 on Build GRCh38
rs995385803 Chr.2: 237487101-237487101 on Build GRCh38
rs1016696692 Chr.2: 237487180-237487180 on Build GRCh38
rs1003041050 Chr.2: 237487144-237487144 on Build GRCh38
rs386656742 Chr.2: 237487170-237487172 on Build GRCh38
rs1036701995 Chr.2: 237487440-237487440 on Build GRCh38
rs923745419 Chr.2: 237487326-237487326 on Build GRCh38
rs954156933 Chr.2: 237487138-237487138 on Build GRCh38
rs542087696 Chr.2: 237487125-237487125 on Build GRCh38
rs1053672481 Chr.2: 237487353-237487353 on Build GRCh38
rs977880891 Chr.2: 237487279-237487279 on Build GRCh38
rs1024690656 Chr.2: 237487131-237487131 on Build GRCh38
rs886583615 Chr.2: 237487417-237487417 on Build GRCh38
rs1044513533 Chr.2: 237487221-237487221 on Build GRCh38
rs1000517894 Chr.2: 237487081-237487081 on Build GRCh38
rs775850998 Chr.2: 237487491-237487491 on Build GRCh38
rs3049924 Chr.2: 237487146-237487147 on Build GRCh38
rs923377477 Chr.2: 237487252-237487252 on Build GRCh38
rs971592483 Chr.2: 237487143-237487143 on Build GRCh38
rs562296064 Chr.2: 237487159-237487159 on Build GRCh38
rs894790052 Chr.2: 237487112-237487112 on Build GRCh38
rs933147545 Chr.2: 237487041-237487041 on Build GRCh38
rs1053963249 Chr.2: 237487188-237487188 on Build GRCh38
rs969615274 Chr.2: 237487284-237487284 on Build GRCh38
rs1016507942 Chr.2: 237487437-237487437 on Build GRCh38
rs1012409304 Chr.2: 237487126-237487126 on Build GRCh38
rs948242169 Chr.2: 237487426-237487426 on Build GRCh38
rs1574530 Chr.2: 237487065-237487065 on Build GRCh38
rs896789450 Chr.2: 237487473-237487473 on Build GRCh38
rs199528173 Chr.2: 237487361-237487361 on Build GRCh38
rs974751449 Chr.2: 237487489-237487489 on Build GRCh38
rs368975429 Chr.2: 237487360-237487372 on Build GRCh38
rs138821909 Chr.2: 237487406-237487406 on Build GRCh38
rs535415862 Chr.2: 237487432-237487432 on Build GRCh38
rs772591534 Chr.2: 237487098-237487098 on Build GRCh38
rs531777402 Chr.2: 237487053-237487053 on Build GRCh38
rs17524101 Chr.2: 237487170-237487170 on Build GRCh38
rs146072497 Chr.2: 237487211-237487211 on Build GRCh38
rs35934917 Chr.2: 237487189-237487190 on Build GRCh38
rs916703467 Chr.2: 237487196-237487196 on Build GRCh38
rs965112062 Chr.2: 237487442-237487442 on Build GRCh38
rs947740583 Chr.2: 237487359-237487359 on Build GRCh38
rs141732125 Chr.2: 237487348-237487360 on Build GRCh38
rs1002566199 Chr.2: 237487254-237487254 on Build GRCh38
rs948512782 Chr.2: 237487219-237487219 on Build GRCh38
rs1036689115 Chr.2: 237487262-237487262 on Build GRCh38
rs776634542 Chr.2: 237487404-237487404 on Build GRCh38
rs1026592037 Chr.2: 237487152-237487152 on Build GRCh38
rs1026826358 Chr.2: 237487341-237487341 on Build GRCh38
rs936607702 Chr.2: 237487329-237487329 on Build GRCh38
rs906265523 Chr.2: 237487380-237487380 on Build GRCh38
rs1027231188 Chr.2: 237487127-237487127 on Build GRCh38
rs1574529 Chr.2: 237487228-237487228 on Build GRCh38
rs576900759 Chr.2: 237487435-237487435 on Build GRCh38
rs868149791 Chr.2: 237487273-237487273 on Build GRCh38
rs758649974 Chr.2: 237487449-237487451 on Build GRCh38
rs374420381 Chr.2: 237487172-237487172 on Build GRCh38
rs1046612406 Chr.2: 237487360-237487360 on Build GRCh38
rs916771414 Chr.2: 237487421-237487421 on Build GRCh38
rs936924533 Chr.2: 237487175-237487175 on Build GRCh38
rs928401308 Chr.2: 237487434-237487434 on Build GRCh38
rs995790468 Chr.2: 237487285-237487285 on Build GRCh38
rs897114056 Chr.2: 237487270-237487270 on Build GRCh38

View Full Product Details