Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238188049-238188547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILKAP
Entrez Gene ID
80895
Gene Name
ILK associated serine/threonine phosphatase
Gene Aliases
ILKAP2, ILKAP3, PP2C-DELTA, PPM1O
UniGene
Hs.92033
Gene Chromosome Location
Chr.2: 238170401-238203702 on Build GRCh38
Transcripts
NM_030768.2 XM_005246106.1 XM_006712784.1 XM_011511946.2 XM_017005056.1 XM_017005057.1 XM_017005058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1406722 c.335C>G p.A112G Chr.2: 238188221-238188221 on Build GRCh38
6285469 c.396C>T p.T132T Chr.2: 238188160-238188160 on Build GRCh38
3364649 c.367G>A p.D123N Chr.2: 238188189-238188189 on Build GRCh38
1018535 c.375C>T p.H125H Chr.2: 238188181-238188181 on Build GRCh38
3673659 c.371C>T p.A124V Chr.2: 238188185-238188185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933572786 Chr.2: 238188401-238188401 on Build GRCh38
rs561872208 Chr.2: 238188400-238188400 on Build GRCh38
rs752999087 Chr.2: 238188288-238188288 on Build GRCh38
rs765203449 Chr.2: 238188092-238188092 on Build GRCh38
rs759389108 Chr.2: 238188351-238188351 on Build GRCh38
rs952024310 Chr.2: 238188315-238188315 on Build GRCh38
rs148464553 Chr.2: 238188160-238188160 on Build GRCh38
rs76719170 Chr.2: 238188374-238188374 on Build GRCh38
rs768970282 Chr.2: 238188109-238188109 on Build GRCh38
rs998895241 Chr.2: 238188459-238188459 on Build GRCh38
rs771209602 Chr.2: 238188321-238188321 on Build GRCh38
rs369165716 Chr.2: 238188348-238188348 on Build GRCh38
rs969955836 Chr.2: 238188240-238188240 on Build GRCh38
rs779463100 Chr.2: 238188122-238188122 on Build GRCh38
rs199781260 Chr.2: 238188114-238188114 on Build GRCh38
rs775463832 Chr.2: 238188373-238188373 on Build GRCh38
rs770999294 Chr.2: 238188493-238188493 on Build GRCh38
rs2305171 Chr.2: 238188499-238188499 on Build GRCh38
rs769371150 Chr.2: 238188199-238188199 on Build GRCh38
rs762250296 Chr.2: 238188108-238188108 on Build GRCh38
rs1011450222 Chr.2: 238188519-238188519 on Build GRCh38
rs772537000 Chr.2: 238188216-238188216 on Build GRCh38
rs775271626 Chr.2: 238188203-238188203 on Build GRCh38
rs369251195 Chr.2: 238188128-238188128 on Build GRCh38
rs764324427 Chr.2: 238188295-238188295 on Build GRCh38
rs765301079 Chr.2: 238188278-238188278 on Build GRCh38
rs760989167 Chr.2: 238188263-238188263 on Build GRCh38
rs376087694 Chr.2: 238188106-238188106 on Build GRCh38
rs754986679 Chr.2: 238188152-238188152 on Build GRCh38
rs747013380 Chr.2: 238188168-238188168 on Build GRCh38
rs779124631 Chr.2: 238188155-238188155 on Build GRCh38
rs759674402 Chr.2: 238188274-238188274 on Build GRCh38
rs962461189 Chr.2: 238188285-238188285 on Build GRCh38
rs758284655 Chr.2: 238188159-238188159 on Build GRCh38
rs982721524 Chr.2: 238188171-238188171 on Build GRCh38
rs60823094 Chr.2: 238188358-238188358 on Build GRCh38
rs769962569 Chr.2: 238188181-238188181 on Build GRCh38
rs1052425506 Chr.2: 238188425-238188425 on Build GRCh38
rs200272443 Chr.2: 238188273-238188273 on Build GRCh38
rs761065499 Chr.2: 238188133-238188133 on Build GRCh38
rs776256430 Chr.2: 238188092-238188092 on Build GRCh38
rs1044744666 Chr.2: 238188497-238188497 on Build GRCh38
rs771011333 Chr.2: 238188169-238188169 on Build GRCh38
rs781233976 Chr.2: 238188178-238188178 on Build GRCh38
rs1052159454 Chr.2: 238188405-238188405 on Build GRCh38
rs202009900 Chr.2: 238188292-238188292 on Build GRCh38
rs573004265 Chr.2: 238188371-238188371 on Build GRCh38
rs773473922 Chr.2: 238188262-238188262 on Build GRCh38
rs748653246 Chr.2: 238188135-238188135 on Build GRCh38
rs182100230 Chr.2: 238188105-238188105 on Build GRCh38
rs972161326 Chr.2: 238188303-238188303 on Build GRCh38
rs1052877047 Chr.2: 238188469-238188469 on Build GRCh38
rs762482133 Chr.2: 238188204-238188204 on Build GRCh38
rs72991006 Chr.2: 238188314-238188314 on Build GRCh38
rs935086581 Chr.2: 238188377-238188377 on Build GRCh38
rs950752961 Chr.2: 238188326-238188326 on Build GRCh38
rs766823995 Chr.2: 238188134-238188134 on Build GRCh38
rs890811918 Chr.2: 238188433-238188433 on Build GRCh38
rs759065163 Chr.2: 238188101-238188101 on Build GRCh38
rs752853235 Chr.2: 238188156-238188156 on Build GRCh38
rs1003779541 Chr.2: 238188506-238188506 on Build GRCh38
rs900955044 Chr.2: 238188078-238188078 on Build GRCh38
rs138619010 Chr.2: 238188139-238188139 on Build GRCh38
rs773377401 Chr.2: 238188251-238188251 on Build GRCh38
rs777058767 Chr.2: 238188345-238188345 on Build GRCh38
rs909265828 Chr.2: 238188353-238188353 on Build GRCh38

View Full Product Details