Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241233729-241234239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO7
Entrez Gene ID
50636
Gene Name
anoctamin 7
Gene Aliases
D-TMPP, DTMPP, IPCA-5, IPCA5, NGEP, PCANAP5, PCANAP5L, TMEM16G
UniGene
Hs.163909
Gene Chromosome Location
Chr.2: 241188509-241239602 on Build GRCh38
Transcripts
XM_011511267.2 XM_017004229.1 NM_001243900.2 NM_001320965.1 NM_001320966.1 NM_001320967.1 NM_005336.5 NM_203346.4 XM_005247002.3 XM_005247003.4 XM_006712475.3 XM_011511058.2 XM_011511060.2 XM_017003940.1 XM_017003941.1 XM_017003942.1

Gene Symbol
HDLBP
Entrez Gene ID
3069
Gene Name
high density lipoprotein binding protein
Gene Aliases
HBP, PRO2900, VGL
UniGene
Hs.471851
Gene Chromosome Location
Chr.2: 241227264-241315842 on Build GRCh38
Transcripts
XM_011511267.2 XM_017004229.1 NM_001243900.2 NM_001320965.1 NM_001320966.1 NM_001320967.1 NM_005336.5 NM_203346.4 XM_005247002.3 XM_005247003.4 XM_006712475.3 XM_011511058.2 XM_011511060.2 XM_017003940.1 XM_017003941.1 XM_017003942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5960013 c.3142G>A p.E1048K Chr.2: 241233867-241233867 on Build GRCh38
6375734 c.3170A>G p.D1057G Chr.2: 241233839-241233839 on Build GRCh38
6489946 c.3180C>A p.D1060E Chr.2: 241233928-241233928 on Build GRCh38
7194986 c.3173A>G p.K1058R Chr.2: 241233836-241233836 on Build GRCh38
7192217 c.3223A>C p.I1075L Chr.2: 241233885-241233885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769530644 Chr.2: 241233791-241233791 on Build GRCh38
rs776440975 Chr.2: 241233853-241233853 on Build GRCh38
rs768105559 Chr.2: 241233877-241233877 on Build GRCh38
rs745607429 Chr.2: 241233840-241233840 on Build GRCh38
rs544201184 Chr.2: 241234110-241234110 on Build GRCh38
rs577729148 Chr.2: 241234214-241234214 on Build GRCh38
rs367712272 Chr.2: 241233826-241233826 on Build GRCh38
rs549861739 Chr.2: 241233800-241233800 on Build GRCh38
rs111961607 Chr.2: 241233819-241233819 on Build GRCh38
rs6760946 Chr.2: 241234216-241234216 on Build GRCh38
rs920986135 Chr.2: 241234138-241234138 on Build GRCh38
rs200137065 Chr.2: 241233799-241233799 on Build GRCh38
rs763957868 Chr.2: 241233782-241233782 on Build GRCh38
rs968194911 Chr.2: 241234150-241234150 on Build GRCh38
rs769686898 Chr.2: 241233859-241233859 on Build GRCh38
rs773492192 Chr.2: 241234171-241234171 on Build GRCh38
rs138093297 Chr.2: 241233945-241233945 on Build GRCh38
rs953630043 Chr.2: 241234193-241234193 on Build GRCh38
rs922211298 Chr.2: 241234177-241234177 on Build GRCh38
rs758477087 Chr.2: 241233823-241233823 on Build GRCh38
rs932454376 Chr.2: 241233768-241233768 on Build GRCh38
rs778450261 Chr.2: 241233756-241233756 on Build GRCh38
rs1054397878 Chr.2: 241234209-241234209 on Build GRCh38
rs1031003487 Chr.2: 241234160-241234160 on Build GRCh38
rs779978284 Chr.2: 241233990-241233990 on Build GRCh38
rs73013896 Chr.2: 241233983-241233983 on Build GRCh38
rs768460666 Chr.2: 241233847-241233847 on Build GRCh38
rs747746237 Chr.2: 241234011-241234011 on Build GRCh38
rs1012107939 Chr.2: 241234085-241234085 on Build GRCh38
rs763095106 Chr.2: 241234175-241234175 on Build GRCh38
rs987017849 Chr.2: 241234161-241234161 on Build GRCh38
rs556209656 Chr.2: 241234203-241234203 on Build GRCh38
rs866105821 Chr.2: 241233963-241233963 on Build GRCh38
rs775160190 Chr.2: 241233794-241233794 on Build GRCh38
rs751619751 Chr.2: 241233976-241233976 on Build GRCh38
rs777103021 Chr.2: 241233927-241233927 on Build GRCh38
rs375987018 Chr.2: 241233865-241233865 on Build GRCh38
rs752975081 Chr.2: 241233943-241233943 on Build GRCh38
rs746934199 Chr.2: 241233829-241233829 on Build GRCh38
rs761088184 Chr.2: 241233904-241233904 on Build GRCh38
rs763627429 Chr.2: 241233795-241233795 on Build GRCh38
rs941124184 Chr.2: 241234215-241234215 on Build GRCh38
rs748902590 Chr.2: 241233997-241233997 on Build GRCh38
rs1034904756 Chr.2: 241234039-241234039 on Build GRCh38
rs2230359 Chr.2: 241233832-241233832 on Build GRCh38
rs111899060 Chr.2: 241233965-241233965 on Build GRCh38
rs768285031 Chr.2: 241234001-241234001 on Build GRCh38
rs972055083 Chr.2: 241234037-241234038 on Build GRCh38
rs765585751 Chr.2: 241233929-241233929 on Build GRCh38
rs146188468 Chr.2: 241233913-241233913 on Build GRCh38
rs567299024 Chr.2: 241233773-241233773 on Build GRCh38
rs373547434 Chr.2: 241234003-241234003 on Build GRCh38
rs367684621 Chr.2: 241233797-241233797 on Build GRCh38
rs757162618 Chr.2: 241233979-241233979 on Build GRCh38
rs536580027 Chr.2: 241234046-241234046 on Build GRCh38
rs748965030 Chr.2: 241233872-241233872 on Build GRCh38
rs138579779 Chr.2: 241234077-241234077 on Build GRCh38
rs752889146 Chr.2: 241233814-241233814 on Build GRCh38
rs568985466 Chr.2: 241233835-241233835 on Build GRCh38
rs571243212 Chr.2: 241233941-241233941 on Build GRCh38
rs186719852 Chr.2: 241234033-241234033 on Build GRCh38
rs756039030 Chr.2: 241233987-241233987 on Build GRCh38
rs764087244 Chr.2: 241233975-241233975 on Build GRCh38
rs962335173 Chr.2: 241234086-241234086 on Build GRCh38
rs773848988 Chr.2: 241233903-241233903 on Build GRCh38
rs765253871 Chr.2: 241234078-241234078 on Build GRCh38
rs574021603 Chr.2: 241234099-241234099 on Build GRCh38
rs759268352 Chr.2: 241233784-241233784 on Build GRCh38
rs1003478245 Chr.2: 241234013-241234013 on Build GRCh38
rs755217872 Chr.2: 241233808-241233808 on Build GRCh38
rs776431776 Chr.2: 241233780-241233780 on Build GRCh38
rs759936329 Chr.2: 241233928-241233928 on Build GRCh38
rs537896293 Chr.2: 241234143-241234143 on Build GRCh38
rs978546580 Chr.2: 241234153-241234153 on Build GRCh38
rs201229744 Chr.2: 241233778-241233778 on Build GRCh38
rs945275616 Chr.2: 241234172-241234172 on Build GRCh38
rs557782392 Chr.2: 241234199-241234199 on Build GRCh38
rs11555550 Chr.2: 241233894-241233894 on Build GRCh38
rs1033016444 Chr.2: 241233841-241233841 on Build GRCh38
rs1022545739 Chr.2: 241234117-241234117 on Build GRCh38
rs903782757 Chr.2: 241234197-241234197 on Build GRCh38
rs111666660 Chr.2: 241233966-241233966 on Build GRCh38

View Full Product Details