Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 661887-662389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRT2
Entrez Gene ID
85508
Gene Name
scratch family transcriptional repressor 2
Gene Aliases
ZNF898B
UniGene
Hs.355284
Gene Chromosome Location
Chr.20: 661596-676179 on Build GRCh38
Transcripts
NM_033129.3

SNP Detail

SNP ID Chromosome Location
rs953467703 Chr.20: 661941-661941 on Build GRCh38
rs77181298 Chr.20: 662149-662149 on Build GRCh38
rs557859912 Chr.20: 662065-662065 on Build GRCh38
rs752026007 Chr.20: 662231-662231 on Build GRCh38
rs930419562 Chr.20: 661969-661969 on Build GRCh38
rs986940743 Chr.20: 662242-662242 on Build GRCh38
rs76627181 Chr.20: 662006-662006 on Build GRCh38
rs1012605946 Chr.20: 662209-662209 on Build GRCh38
rs1042861559 Chr.20: 662267-662267 on Build GRCh38
rs768030787 Chr.20: 661971-661971 on Build GRCh38
rs938923417 Chr.20: 662216-662216 on Build GRCh38
rs986972043 Chr.20: 662082-662082 on Build GRCh38
rs1031056601 Chr.20: 662226-662226 on Build GRCh38
rs1031170975 Chr.20: 662071-662072 on Build GRCh38
rs984803586 Chr.20: 662155-662155 on Build GRCh38
rs1056380078 Chr.20: 662225-662225 on Build GRCh38
rs181992370 Chr.20: 662208-662208 on Build GRCh38
rs545095921 Chr.20: 662276-662276 on Build GRCh38
rs199957229 Chr.20: 662343-662343 on Build GRCh38
rs886198236 Chr.20: 662154-662154 on Build GRCh38
rs1040689026 Chr.20: 662303-662303 on Build GRCh38
rs371343653 Chr.20: 662240-662240 on Build GRCh38
rs951769883 Chr.20: 662306-662306 on Build GRCh38
rs879053140 Chr.20: 662275-662275 on Build GRCh38
rs75740470 Chr.20: 661998-661998 on Build GRCh38
rs1029802805 Chr.20: 662090-662090 on Build GRCh38
rs764740282 Chr.20: 662083-662083 on Build GRCh38
rs898888143 Chr.20: 661966-661966 on Build GRCh38
rs974424848 Chr.20: 662135-662135 on Build GRCh38
rs1012226694 Chr.20: 662056-662056 on Build GRCh38
rs557182625 Chr.20: 662162-662162 on Build GRCh38
rs1018444759 Chr.20: 662357-662357 on Build GRCh38
rs974771851 Chr.20: 662287-662287 on Build GRCh38
rs1039121620 Chr.20: 661956-661956 on Build GRCh38
rs939160124 Chr.20: 662158-662158 on Build GRCh38
rs143737403 Chr.20: 662286-662286 on Build GRCh38
rs1036610152 Chr.20: 661979-661979 on Build GRCh38
rs1047897708 Chr.20: 662152-662152 on Build GRCh38
rs987088866 Chr.20: 661935-661936 on Build GRCh38
rs1034867417 Chr.20: 662041-662041 on Build GRCh38
rs573856239 Chr.20: 662191-662191 on Build GRCh38
rs1011297031 Chr.20: 662044-662044 on Build GRCh38
rs553195919 Chr.20: 662215-662215 on Build GRCh38
rs878952928 Chr.20: 662239-662240 on Build GRCh38
rs1022287029 Chr.20: 662212-662212 on Build GRCh38
rs568390043 Chr.20: 662087-662087 on Build GRCh38
rs996744133 Chr.20: 662331-662331 on Build GRCh38
rs1018475800 Chr.20: 662272-662272 on Build GRCh38
rs909181218 Chr.20: 661962-661962 on Build GRCh38
rs907608540 Chr.20: 662339-662339 on Build GRCh38
rs901095545 Chr.20: 662320-662320 on Build GRCh38
rs962911321 Chr.20: 662362-662362 on Build GRCh38
rs962055408 Chr.20: 662200-662200 on Build GRCh38
rs920194699 Chr.20: 662301-662301 on Build GRCh38
rs370114399 Chr.20: 661942-661942 on Build GRCh38
rs955484157 Chr.20: 662072-662072 on Build GRCh38
rs909300220 Chr.20: 662197-662197 on Build GRCh38
rs1042746459 Chr.20: 662059-662059 on Build GRCh38
rs975262870 Chr.20: 661923-661923 on Build GRCh38
rs997100330 Chr.20: 662094-662094 on Build GRCh38
rs1007028186 Chr.20: 662347-662347 on Build GRCh38
rs895080974 Chr.20: 662045-662045 on Build GRCh38
rs934337193 Chr.20: 662302-662302 on Build GRCh38
rs138924618 Chr.20: 662060-662060 on Build GRCh38
rs894234508 Chr.20: 662011-662011 on Build GRCh38
rs529439061 Chr.20: 661995-661995 on Build GRCh38
rs1028579467 Chr.20: 662345-662345 on Build GRCh38
rs547624197 Chr.20: 662167-662167 on Build GRCh38
rs571590313 Chr.20: 662071-662071 on Build GRCh38
rs1028125848 Chr.20: 662113-662113 on Build GRCh38
rs6116988 Chr.20: 662111-662111 on Build GRCh38
rs538624015 Chr.20: 662277-662279 on Build GRCh38
rs6116987 Chr.20: 662106-662106 on Build GRCh38
rs922036486 Chr.20: 661940-661940 on Build GRCh38

View Full Product Details