Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 675667-676209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRT2
Entrez Gene ID
85508
Gene Name
scratch family transcriptional repressor 2
Gene Aliases
ZNF898B
UniGene
Hs.355284
Gene Chromosome Location
Chr.20: 661596-676179 on Build GRCh38
Transcripts
NM_033129.3

SNP Detail

SNP ID Chromosome Location
rs993110052 Chr.20: 676086-676086 on Build GRCh38
rs901297066 Chr.20: 676033-676033 on Build GRCh38
rs940354045 Chr.20: 675866-675866 on Build GRCh38
rs557213104 Chr.20: 675798-675798 on Build GRCh38
rs910609900 Chr.20: 675781-675781 on Build GRCh38
rs926448003 Chr.20: 676160-676160 on Build GRCh38
rs150411471 Chr.20: 675726-675726 on Build GRCh38
rs61589065 Chr.20: 675776-675777 on Build GRCh38
rs929286586 Chr.20: 676073-676073 on Build GRCh38
rs942026770 Chr.20: 675785-675785 on Build GRCh38
rs534584224 Chr.20: 675862-675862 on Build GRCh38
rs193225234 Chr.20: 676017-676017 on Build GRCh38
rs1044509796 Chr.20: 676157-676157 on Build GRCh38
rs1012694072 Chr.20: 675695-675695 on Build GRCh38
rs146589074 Chr.20: 675763-675764 on Build GRCh38
rs189312619 Chr.20: 675977-675977 on Build GRCh38
rs948893175 Chr.20: 676122-676122 on Build GRCh38
rs963420426 Chr.20: 676035-676035 on Build GRCh38
rs552123816 Chr.20: 675843-675844 on Build GRCh38
rs949228676 Chr.20: 675920-675920 on Build GRCh38
rs980932994 Chr.20: 676148-676148 on Build GRCh38
rs896293929 Chr.20: 675728-675729 on Build GRCh38
rs905138752 Chr.20: 675745-675745 on Build GRCh38
rs984361401 Chr.20: 676066-676069 on Build GRCh38
rs557240710 Chr.20: 676100-676100 on Build GRCh38
rs13045130 Chr.20: 675755-675755 on Build GRCh38
rs949204272 Chr.20: 676109-676109 on Build GRCh38
rs1009576915 Chr.20: 675784-675785 on Build GRCh38
rs1022658554 Chr.20: 675704-675704 on Build GRCh38
rs573863983 Chr.20: 675806-675806 on Build GRCh38
rs537246596 Chr.20: 675989-675989 on Build GRCh38
rs940385112 Chr.20: 675719-675719 on Build GRCh38
rs778405943 Chr.20: 675819-675819 on Build GRCh38
rs971785754 Chr.20: 675874-675874 on Build GRCh38
rs533367287 Chr.20: 675715-675715 on Build GRCh38
rs973360708 Chr.20: 675794-675794 on Build GRCh38
rs1029537120 Chr.20: 675751-675751 on Build GRCh38
rs61546405 Chr.20: 676166-676166 on Build GRCh38
rs1029979716 Chr.20: 676013-676013 on Build GRCh38
rs984784737 Chr.20: 675855-675855 on Build GRCh38
rs1001266415 Chr.20: 675746-675746 on Build GRCh38
rs986242810 Chr.20: 675759-675759 on Build GRCh38
rs997419974 Chr.20: 676039-676039 on Build GRCh38
rs1019642279 Chr.20: 676019-676019 on Build GRCh38
rs752432690 Chr.20: 675831-675831 on Build GRCh38
rs904210900 Chr.20: 675705-675705 on Build GRCh38
rs917467200 Chr.20: 676094-676094 on Build GRCh38
rs1028449926 Chr.20: 676059-676059 on Build GRCh38
rs1017702767 Chr.20: 676021-676021 on Build GRCh38
rs956712121 Chr.20: 675765-675765 on Build GRCh38
rs1057500715 Chr.20: 675810-675810 on Build GRCh38
rs777559940 Chr.20: 676149-676149 on Build GRCh38
rs535591466 Chr.20: 675732-675732 on Build GRCh38
rs953915171 Chr.20: 676014-676014 on Build GRCh38
rs904243321 Chr.20: 675876-675876 on Build GRCh38
rs971752943 Chr.20: 676091-676091 on Build GRCh38
rs999794554 Chr.20: 675708-675708 on Build GRCh38
rs1044941307 Chr.20: 675929-675929 on Build GRCh38
rs371185461 Chr.20: 675847-675848 on Build GRCh38
rs961689470 Chr.20: 676086-676087 on Build GRCh38
rs953993932 Chr.20: 675753-675753 on Build GRCh38
rs1032384272 Chr.20: 675754-675754 on Build GRCh38
rs917665187 Chr.20: 676097-676097 on Build GRCh38
rs919290613 Chr.20: 676060-676060 on Build GRCh38
rs753272566 Chr.20: 675959-675959 on Build GRCh38
rs549813204 Chr.20: 675720-675720 on Build GRCh38
rs1044144973 Chr.20: 675858-675858 on Build GRCh38
rs558047387 Chr.20: 675911-675911 on Build GRCh38
rs998136030 Chr.20: 675999-675999 on Build GRCh38
rs973484966 Chr.20: 676038-676038 on Build GRCh38
rs758178703 Chr.20: 675938-675938 on Build GRCh38
rs905107285 Chr.20: 675986-675986 on Build GRCh38

View Full Product Details