Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 759693-760238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC52A3
Entrez Gene ID
113278
Gene Name
solute carrier family 52 member 3
Gene Aliases
BVVLS, BVVLS1, C20orf54, RFT2, RFVT3, bA371L19.1, hRFT2
UniGene
Hs.283865
Gene Chromosome Location
Chr.20: 760080-775985 on Build GRCh38
Transcripts
NM_033409.3 XM_005260655.3 XM_011529148.1

SNP Detail

SNP ID Chromosome Location
rs927716649 Chr.20: 759860-759860 on Build GRCh38
rs901263882 Chr.20: 760052-760052 on Build GRCh38
rs1055785607 Chr.20: 759816-759816 on Build GRCh38
rs760576964 Chr.20: 759871-759872 on Build GRCh38
rs143023974 Chr.20: 759819-759819 on Build GRCh38
rs908738210 Chr.20: 759822-759822 on Build GRCh38
rs546799769 Chr.20: 760202-760202 on Build GRCh38
rs558569470 Chr.20: 759740-759740 on Build GRCh38
rs1028368818 Chr.20: 760071-760071 on Build GRCh38
rs530785789 Chr.20: 760129-760129 on Build GRCh38
rs369016916 Chr.20: 759973-759973 on Build GRCh38
rs915893921 Chr.20: 760039-760039 on Build GRCh38
rs996903635 Chr.20: 760064-760064 on Build GRCh38
rs576794221 Chr.20: 759868-759868 on Build GRCh38
rs573916659 Chr.20: 759898-759898 on Build GRCh38
rs1035155243 Chr.20: 760140-760140 on Build GRCh38
rs982695674 Chr.20: 760120-760120 on Build GRCh38
rs769763062 Chr.20: 759900-759900 on Build GRCh38
rs530089121 Chr.20: 759834-759834 on Build GRCh38
rs919776670 Chr.20: 759924-759924 on Build GRCh38
rs35871248 Chr.20: 759855-759855 on Build GRCh38
rs537354456 Chr.20: 759843-759843 on Build GRCh38
rs954940986 Chr.20: 759863-759863 on Build GRCh38
rs1053018 Chr.20: 760189-760189 on Build GRCh38
rs1007879202 Chr.20: 759876-759876 on Build GRCh38
rs946671677 Chr.20: 759928-759928 on Build GRCh38
rs1015870831 Chr.20: 760087-760087 on Build GRCh38
rs140593591 Chr.20: 760156-760156 on Build GRCh38
rs780783308 Chr.20: 759996-759996 on Build GRCh38
rs1043507213 Chr.20: 760069-760069 on Build GRCh38
rs919753540 Chr.20: 759732-759732 on Build GRCh38
rs573456324 Chr.20: 759995-759995 on Build GRCh38
rs1051868483 Chr.20: 760136-760136 on Build GRCh38
rs914904960 Chr.20: 759919-759919 on Build GRCh38
rs779896892 Chr.20: 760075-760075 on Build GRCh38
rs542676349 Chr.20: 759903-759903 on Build GRCh38
rs1027124865 Chr.20: 760201-760201 on Build GRCh38
rs559579630 Chr.20: 759980-759980 on Build GRCh38
rs569392053 Chr.20: 760209-760209 on Build GRCh38
rs961518063 Chr.20: 759824-759824 on Build GRCh38
rs565321657 Chr.20: 760031-760031 on Build GRCh38
rs903370120 Chr.20: 760088-760088 on Build GRCh38
rs928443745 Chr.20: 760214-760214 on Build GRCh38
rs1046980330 Chr.20: 759747-759747 on Build GRCh38
rs965467938 Chr.20: 759757-759757 on Build GRCh38
rs1028296676 Chr.20: 759803-759803 on Build GRCh38
rs186211467 Chr.20: 760180-760180 on Build GRCh38
rs751019431 Chr.20: 759979-759979 on Build GRCh38
rs765663155 Chr.20: 759908-759908 on Build GRCh38
rs903316723 Chr.20: 759829-759829 on Build GRCh38
rs142926065 Chr.20: 760011-760012 on Build GRCh38
rs998979488 Chr.20: 759731-759731 on Build GRCh38
rs45481905 Chr.20: 760018-760018 on Build GRCh38
rs1005828167 Chr.20: 760078-760078 on Build GRCh38
rs961618899 Chr.20: 760112-760112 on Build GRCh38
rs190022755 Chr.20: 759818-759818 on Build GRCh38
rs180778100 Chr.20: 760074-760074 on Build GRCh38
rs567672725 Chr.20: 759747-759749 on Build GRCh38
rs377548107 Chr.20: 759914-759915 on Build GRCh38
rs951258916 Chr.20: 759950-759950 on Build GRCh38
rs750044987 Chr.20: 759998-759998 on Build GRCh38
rs778738972 Chr.20: 760178-760178 on Build GRCh38
rs997270296 Chr.20: 759798-759798 on Build GRCh38
rs944607784 Chr.20: 760208-760208 on Build GRCh38
rs959421403 Chr.20: 760145-760145 on Build GRCh38
rs947503559 Chr.20: 760045-760045 on Build GRCh38

View Full Product Details