Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1228203-1228721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21L1
Entrez Gene ID
642636
Gene Name
RAD21 cohesin complex component like 1
Gene Aliases
RAD21L, dJ545L17.2
UniGene
Hs.713451
Gene Chromosome Location
Chr.20: 1226043-1255876 on Build GRCh38
Transcripts
NM_001136566.2 XM_006723602.2 XM_006723603.2 XM_006723604.2 XM_006723605.3 XM_011529317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024523 c.47A>G p.K16R Chr.20: 1228500-1228500 on Build GRCh38
1024525 c.133C>A p.L45I Chr.20: 1228586-1228586 on Build GRCh38
1483430 c.40T>C p.L14L Chr.20: 1228493-1228493 on Build GRCh38
1024526 c.133C>A p.L45I Chr.20: 1228586-1228586 on Build GRCh38
1615324 c.100G>T p.E34* Chr.20: 1228553-1228553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972593757 Chr.20: 1228492-1228492 on Build GRCh38
rs994867760 Chr.20: 1228407-1228407 on Build GRCh38
rs891880021 Chr.20: 1228288-1228288 on Build GRCh38
rs267605837 Chr.20: 1228577-1228577 on Build GRCh38
rs1035598264 Chr.20: 1228253-1228253 on Build GRCh38
rs943182880 Chr.20: 1228384-1228384 on Build GRCh38
rs924558280 Chr.20: 1228363-1228363 on Build GRCh38
rs576509634 Chr.20: 1228432-1228432 on Build GRCh38
rs985322592 Chr.20: 1228453-1228453 on Build GRCh38
rs562930022 Chr.20: 1228377-1228377 on Build GRCh38
rs768027328 Chr.20: 1228572-1228572 on Build GRCh38
rs6078241 Chr.20: 1228283-1228283 on Build GRCh38
rs899231376 Chr.20: 1228403-1228403 on Build GRCh38
rs139003524 Chr.20: 1228285-1228285 on Build GRCh38
rs1012391850 Chr.20: 1228651-1228651 on Build GRCh38
rs1051642959 Chr.20: 1228373-1228375 on Build GRCh38
rs918453025 Chr.20: 1228566-1228566 on Build GRCh38
rs752110056 Chr.20: 1228511-1228511 on Build GRCh38
rs959821964 Chr.20: 1228650-1228650 on Build GRCh38
rs769346469 Chr.20: 1228465-1228465 on Build GRCh38
rs991935540 Chr.20: 1228662-1228662 on Build GRCh38
rs183738538 Chr.20: 1228648-1228648 on Build GRCh38
rs775142447 Chr.20: 1228479-1228479 on Build GRCh38
rs886732438 Chr.20: 1228471-1228471 on Build GRCh38
rs978377422 Chr.20: 1228356-1228356 on Build GRCh38
rs950932133 Chr.20: 1228234-1228234 on Build GRCh38
rs947071142 Chr.20: 1228356-1228357 on Build GRCh38
rs762275074 Chr.20: 1228512-1228512 on Build GRCh38
rs998260920 Chr.20: 1228317-1228317 on Build GRCh38
rs762518164 Chr.20: 1228484-1228484 on Build GRCh38
rs982338660 Chr.20: 1228235-1228235 on Build GRCh38
rs385856 Chr.20: 1228523-1228523 on Build GRCh38
rs915633193 Chr.20: 1228353-1228353 on Build GRCh38
rs763580583 Chr.20: 1228489-1228489 on Build GRCh38
rs1039547973 Chr.20: 1228402-1228402 on Build GRCh38
rs954069670 Chr.20: 1228395-1228395 on Build GRCh38
rs541981249 Chr.20: 1228630-1228630 on Build GRCh38
rs1000380676 Chr.20: 1228231-1228231 on Build GRCh38
rs939074520 Chr.20: 1228586-1228586 on Build GRCh38
rs934571018 Chr.20: 1228366-1228366 on Build GRCh38
rs959530318 Chr.20: 1228278-1228278 on Build GRCh38

View Full Product Details