Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1242944-1243475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21L1
Entrez Gene ID
642636
Gene Name
RAD21 cohesin complex component like 1
Gene Aliases
RAD21L, dJ545L17.2
UniGene
Hs.713451
Gene Chromosome Location
Chr.20: 1226043-1255876 on Build GRCh38
Transcripts
NM_001136566.2 XM_006723602.2 XM_006723603.2 XM_006723604.2 XM_006723605.3 XM_011529317.2 XM_011529319.1 XM_011529320.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5447355 c.1119A>C p.K373N Chr.20: 1243132-1243132 on Build GRCh38
1024535 c.1107C>A p.S369S Chr.20: 1243120-1243120 on Build GRCh38
6533030 c.1176T>C p.N392N Chr.20: 1243189-1243189 on Build GRCh38
4977503 c.1099T>G p.F367V Chr.20: 1243112-1243112 on Build GRCh38
6190125 c.1138C>A p.Q380K Chr.20: 1243151-1243151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960886279 Chr.20: 1243388-1243388 on Build GRCh38
rs1016980777 Chr.20: 1243095-1243095 on Build GRCh38
rs995458549 Chr.20: 1243220-1243220 on Build GRCh38
rs761977281 Chr.20: 1243076-1243076 on Build GRCh38
rs562847784 Chr.20: 1243283-1243283 on Build GRCh38
rs998009087 Chr.20: 1243036-1243036 on Build GRCh38
rs6134334 Chr.20: 1243452-1243452 on Build GRCh38
rs763873759 Chr.20: 1243108-1243108 on Build GRCh38
rs545912970 Chr.20: 1243255-1243255 on Build GRCh38
rs537683864 Chr.20: 1243069-1243069 on Build GRCh38
rs753153239 Chr.20: 1243075-1243075 on Build GRCh38
rs758065578 Chr.20: 1243076-1243076 on Build GRCh38
rs1009416970 Chr.20: 1243072-1243072 on Build GRCh38
rs776344877 Chr.20: 1243245-1243246 on Build GRCh38
rs974711570 Chr.20: 1243240-1243240 on Build GRCh38
rs1044088866 Chr.20: 1243430-1243430 on Build GRCh38
rs936020460 Chr.20: 1243419-1243419 on Build GRCh38
rs925577533 Chr.20: 1243435-1243435 on Build GRCh38
rs752064898 Chr.20: 1243429-1243429 on Build GRCh38
rs200764637 Chr.20: 1243263-1243263 on Build GRCh38
rs575778340 Chr.20: 1243402-1243402 on Build GRCh38
rs774584064 Chr.20: 1243075-1243076 on Build GRCh38
rs555963660 Chr.20: 1243071-1243071 on Build GRCh38
rs531711259 Chr.20: 1243357-1243357 on Build GRCh38
rs373402621 Chr.20: 1243227-1243227 on Build GRCh38
rs1031518710 Chr.20: 1243027-1243027 on Build GRCh38
rs970094128 Chr.20: 1243360-1243360 on Build GRCh38
rs574703429 Chr.20: 1242997-1242997 on Build GRCh38
rs535073597 Chr.20: 1243403-1243403 on Build GRCh38
rs917440182 Chr.20: 1243303-1243303 on Build GRCh38
rs111583951 Chr.20: 1242971-1242971 on Build GRCh38
rs540775998 Chr.20: 1243016-1243016 on Build GRCh38
rs756878073 Chr.20: 1243191-1243191 on Build GRCh38
rs952303987 Chr.20: 1243320-1243320 on Build GRCh38
rs532178503 Chr.20: 1243213-1243215 on Build GRCh38
rs542162571 Chr.20: 1243377-1243377 on Build GRCh38
rs527540346 Chr.20: 1243448-1243448 on Build GRCh38
rs369975355 Chr.20: 1243054-1243054 on Build GRCh38
rs577260951 Chr.20: 1243148-1243148 on Build GRCh38
rs1044368052 Chr.20: 1242970-1242970 on Build GRCh38
rs891599781 Chr.20: 1243034-1243034 on Build GRCh38
rs780819010 Chr.20: 1243192-1243192 on Build GRCh38
rs913137426 Chr.20: 1243436-1243436 on Build GRCh38
rs1026521486 Chr.20: 1243228-1243228 on Build GRCh38
rs992944620 Chr.20: 1243280-1243280 on Build GRCh38

View Full Product Details