Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1304788-1305278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNPH
Entrez Gene ID
9751
Gene Name
syntaphilin
Gene Aliases
-
UniGene
Hs.323833
Gene Chromosome Location
Chr.20: 1266291-1309327 on Build GRCh38
Transcripts
NM_001318234.1 NM_014723.3 XM_005260889.3 XM_011529402.2 XM_011529403.2 XM_011529404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5365916 c.696C>T p.P232P Chr.20: 1305265-1305265 on Build GRCh38
4289606 c.693C>A p.A231A Chr.20: 1305130-1305130 on Build GRCh38
5365917 c.828C>T p.P276P Chr.20: 1305265-1305265 on Build GRCh38
1659837 c.645C>A p.Y215* Chr.20: 1305214-1305214 on Build GRCh38
4134235 c.468G>A p.T156T Chr.20: 1304905-1304905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932863084 Chr.20: 1304993-1304993 on Build GRCh38
rs145538484 Chr.20: 1304944-1304944 on Build GRCh38
rs776133925 Chr.20: 1305231-1305231 on Build GRCh38
rs751832991 Chr.20: 1305255-1305255 on Build GRCh38
rs777161828 Chr.20: 1305156-1305156 on Build GRCh38
rs761460938 Chr.20: 1304896-1304896 on Build GRCh38
rs772913959 Chr.20: 1305135-1305135 on Build GRCh38
rs112041885 Chr.20: 1304950-1304950 on Build GRCh38
rs748293041 Chr.20: 1304841-1304841 on Build GRCh38
rs761585417 Chr.20: 1305016-1305016 on Build GRCh38
rs141020147 Chr.20: 1304997-1304997 on Build GRCh38
rs370884038 Chr.20: 1304868-1304868 on Build GRCh38
rs372565382 Chr.20: 1305053-1305053 on Build GRCh38
rs772787492 Chr.20: 1305220-1305220 on Build GRCh38
rs749955763 Chr.20: 1305128-1305128 on Build GRCh38
rs770443739 Chr.20: 1305223-1305223 on Build GRCh38
rs749330341 Chr.20: 1304874-1304875 on Build GRCh38
rs746226956 Chr.20: 1304832-1304833 on Build GRCh38
rs370939722 Chr.20: 1305067-1305067 on Build GRCh38
rs140499293 Chr.20: 1305256-1305256 on Build GRCh38
rs754354340 Chr.20: 1305046-1305046 on Build GRCh38
rs751246300 Chr.20: 1304889-1304889 on Build GRCh38
rs191018281 Chr.20: 1305249-1305249 on Build GRCh38
rs768687253 Chr.20: 1305038-1305039 on Build GRCh38
rs757349234 Chr.20: 1305088-1305088 on Build GRCh38
rs144660706 Chr.20: 1305105-1305105 on Build GRCh38
rs771623503 Chr.20: 1305127-1305127 on Build GRCh38
rs772244014 Chr.20: 1304959-1304959 on Build GRCh38
rs747128780 Chr.20: 1305071-1305071 on Build GRCh38
rs760160542 Chr.20: 1305141-1305141 on Build GRCh38
rs767102450 Chr.20: 1304904-1304904 on Build GRCh38
rs202106741 Chr.20: 1304849-1304849 on Build GRCh38
rs1026244147 Chr.20: 1304869-1304869 on Build GRCh38
rs760060824 Chr.20: 1305161-1305161 on Build GRCh38
rs544910092 Chr.20: 1304874-1304874 on Build GRCh38
rs376543479 Chr.20: 1305172-1305172 on Build GRCh38
rs147752801 Chr.20: 1304953-1304953 on Build GRCh38
rs779907799 Chr.20: 1305089-1305089 on Build GRCh38
rs747890865 Chr.20: 1305115-1305115 on Build GRCh38
rs756597097 Chr.20: 1304915-1304915 on Build GRCh38
rs769627020 Chr.20: 1304966-1304966 on Build GRCh38
rs374611015 Chr.20: 1304852-1304852 on Build GRCh38
rs370636304 Chr.20: 1305199-1305199 on Build GRCh38
rs746640965 Chr.20: 1305222-1305222 on Build GRCh38
rs757323118 Chr.20: 1305197-1305197 on Build GRCh38
rs373501992 Chr.20: 1305124-1305124 on Build GRCh38
rs978277873 Chr.20: 1305047-1305047 on Build GRCh38
rs767145769 Chr.20: 1305017-1305017 on Build GRCh38
rs748022382 Chr.20: 1305206-1305206 on Build GRCh38
rs866586809 Chr.20: 1305238-1305238 on Build GRCh38
rs758979785 Chr.20: 1305232-1305232 on Build GRCh38
rs766862169 Chr.20: 1305035-1305035 on Build GRCh38
rs774307573 Chr.20: 1305106-1305106 on Build GRCh38
rs573421544 Chr.20: 1305215-1305215 on Build GRCh38
rs779162332 Chr.20: 1304954-1304954 on Build GRCh38
rs753011927 Chr.20: 1305058-1305058 on Build GRCh38
rs35467629 Chr.20: 1304905-1304905 on Build GRCh38
rs142687427 Chr.20: 1305033-1305033 on Build GRCh38
rs762901766 Chr.20: 1304984-1304984 on Build GRCh38
rs748346139 Chr.20: 1304956-1304956 on Build GRCh38
rs879326174 Chr.20: 1304951-1304951 on Build GRCh38
rs775496762 Chr.20: 1304980-1304980 on Build GRCh38
rs777738406 Chr.20: 1304962-1304962 on Build GRCh38
rs147549046 Chr.20: 1305248-1305248 on Build GRCh38
rs923933905 Chr.20: 1304812-1304812 on Build GRCh38
rs747052614 Chr.20: 1304965-1304965 on Build GRCh38
rs778904561 Chr.20: 1305041-1305042 on Build GRCh38
rs754975513 Chr.20: 1305201-1305201 on Build GRCh38
rs201621065 Chr.20: 1304873-1304873 on Build GRCh38
rs768551730 Chr.20: 1304996-1304996 on Build GRCh38
rs765889460 Chr.20: 1305153-1305153 on Build GRCh38
rs748386557 Chr.20: 1305092-1305094 on Build GRCh38
rs756558469 Chr.20: 1304853-1304854 on Build GRCh38
rs760163859 Chr.20: 1305034-1305034 on Build GRCh38
rs889883533 Chr.20: 1304823-1304823 on Build GRCh38
rs780564967 Chr.20: 1304863-1304865 on Build GRCh38
rs769723000 Chr.20: 1304863-1304863 on Build GRCh38
rs764283957 Chr.20: 1305189-1305189 on Build GRCh38
rs376424530 Chr.20: 1304853-1304853 on Build GRCh38
rs780548052 Chr.20: 1304926-1304926 on Build GRCh38
rs753064810 Chr.20: 1305173-1305173 on Build GRCh38
rs187274269 Chr.20: 1305070-1305070 on Build GRCh38
rs749210160 Chr.20: 1305100-1305100 on Build GRCh38
rs776672080 Chr.20: 1304851-1304851 on Build GRCh38
rs981917640 Chr.20: 1304818-1304818 on Build GRCh38
rs151237144 Chr.20: 1305188-1305188 on Build GRCh38
rs911251364 Chr.20: 1305217-1305217 on Build GRCh38
rs1034831176 Chr.20: 1304822-1304822 on Build GRCh38
rs933990243 Chr.20: 1304820-1304820 on Build GRCh38
rs982308031 Chr.20: 1304991-1304999 on Build GRCh38
rs770353515 Chr.20: 1305247-1305247 on Build GRCh38
rs765622837 Chr.20: 1305055-1305055 on Build GRCh38
rs566454103 Chr.20: 1304834-1304834 on Build GRCh38
rs751787189 Chr.20: 1305192-1305192 on Build GRCh38

View Full Product Details