Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1895360-1895853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRPA
Entrez Gene ID
140885
Gene Name
signal regulatory protein alpha
Gene Aliases
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP
UniGene
Hs.581021
Gene Chromosome Location
Chr.20: 1894117-1939895 on Build GRCh38
Transcripts
NM_001040022.1 NM_001040023.1 NM_080792.2 XM_005260669.3 XM_005260670.3 XM_006723545.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319689 c.45C>T p.L15L Chr.20: 1895492-1895492 on Build GRCh38
5449399 c.58G>A p.A20T Chr.20: 1895505-1895505 on Build GRCh38
5513600 c.79+8delC p.? Chr.20: 1895534-1895534 on Build GRCh38
4512343 c.8C>T p.P3L Chr.20: 1895455-1895455 on Build GRCh38
6410093 c.37C>A p.P13T Chr.20: 1895484-1895484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989697705 Chr.20: 1895539-1895539 on Build GRCh38
rs777620920 Chr.20: 1895665-1895665 on Build GRCh38
rs560741229 Chr.20: 1895718-1895718 on Build GRCh38
rs750995601 Chr.20: 1895433-1895433 on Build GRCh38
rs931948826 Chr.20: 1895540-1895540 on Build GRCh38
rs1025165674 Chr.20: 1895443-1895443 on Build GRCh38
rs1003000885 Chr.20: 1895600-1895600 on Build GRCh38
rs554244933 Chr.20: 1895618-1895618 on Build GRCh38
rs1033320320 Chr.20: 1895492-1895492 on Build GRCh38
rs941914854 Chr.20: 1895389-1895389 on Build GRCh38
rs550210998 Chr.20: 1895826-1895826 on Build GRCh38
rs990899951 Chr.20: 1895427-1895427 on Build GRCh38
rs941360210 Chr.20: 1895635-1895635 on Build GRCh38
rs959120098 Chr.20: 1895701-1895701 on Build GRCh38
rs112674043 Chr.20: 1895439-1895439 on Build GRCh38
rs1010557760 Chr.20: 1895799-1895799 on Build GRCh38
rs962215784 Chr.20: 1895397-1895397 on Build GRCh38
rs780722105 Chr.20: 1895728-1895728 on Build GRCh38
rs935169449 Chr.20: 1895507-1895507 on Build GRCh38
rs1036372220 Chr.20: 1895403-1895403 on Build GRCh38
rs536740410 Chr.20: 1895785-1895785 on Build GRCh38
rs923187821 Chr.20: 1895525-1895525 on Build GRCh38
rs567465683 Chr.20: 1895776-1895776 on Build GRCh38
rs929158749 Chr.20: 1895736-1895736 on Build GRCh38
rs755781590 Chr.20: 1895467-1895467 on Build GRCh38
rs897721778 Chr.20: 1895736-1895744 on Build GRCh38
rs181584999 Chr.20: 1895798-1895798 on Build GRCh38
rs1038059925 Chr.20: 1895650-1895650 on Build GRCh38
rs547472341 Chr.20: 1895766-1895766 on Build GRCh38
rs909854870 Chr.20: 1895626-1895626 on Build GRCh38
rs1013191888 Chr.20: 1895703-1895703 on Build GRCh38
rs981548852 Chr.20: 1895398-1895398 on Build GRCh38
rs779679392 Chr.20: 1895827-1895827 on Build GRCh38
rs967304709 Chr.20: 1895557-1895557 on Build GRCh38
rs757862537 Chr.20: 1895486-1895486 on Build GRCh38
rs562529545 Chr.20: 1895567-1895567 on Build GRCh38
rs968099700 Chr.20: 1895462-1895462 on Build GRCh38
rs967717533 Chr.20: 1895825-1895825 on Build GRCh38
rs923654765 Chr.20: 1895498-1895498 on Build GRCh38
rs545145662 Chr.20: 1895655-1895655 on Build GRCh38
rs577733641 Chr.20: 1895643-1895643 on Build GRCh38
rs1036420696 Chr.20: 1895594-1895594 on Build GRCh38
rs1001882194 Chr.20: 1895470-1895470 on Build GRCh38
rs897890916 Chr.20: 1895597-1895597 on Build GRCh38
rs759597616 Chr.20: 1895533-1895534 on Build GRCh38
rs571281078 Chr.20: 1895586-1895586 on Build GRCh38
rs958947685 Chr.20: 1895426-1895426 on Build GRCh38
rs1035829385 Chr.20: 1895638-1895638 on Build GRCh38
rs1040190802 Chr.20: 1895516-1895516 on Build GRCh38
rs932716089 Chr.20: 1895592-1895600 on Build GRCh38
rs887810198 Chr.20: 1895582-1895582 on Build GRCh38
rs1037506845 Chr.20: 1895395-1895395 on Build GRCh38
rs922692567 Chr.20: 1895575-1895575 on Build GRCh38
rs976934372 Chr.20: 1895566-1895566 on Build GRCh38
rs916282843 Chr.20: 1895440-1895440 on Build GRCh38
rs1001913596 Chr.20: 1895767-1895767 on Build GRCh38
rs781759027 Chr.20: 1895548-1895548 on Build GRCh38

View Full Product Details