Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2325771-2326265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM3
Entrez Gene ID
7053
Gene Name
transglutaminase 3
Gene Aliases
TGE
UniGene
Hs.2022
Gene Chromosome Location
Chr.20: 2295967-2341079 on Build GRCh38
Transcripts
NM_003245.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
315894 c.1038G>T p.S346S Chr.20: 2325903-2325903 on Build GRCh38
4520777 c.1075G>C p.E359Q Chr.20: 2325940-2325940 on Build GRCh38
1713080 c.984G>A p.W328* Chr.20: 2325849-2325849 on Build GRCh38
1686799 c.984-1G>A p.? Chr.20: 2325848-2325848 on Build GRCh38
477903 c.1042G>A p.G348S Chr.20: 2325907-2325907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752966586 Chr.20: 2326081-2326081 on Build GRCh38
rs958431228 Chr.20: 2326151-2326151 on Build GRCh38
rs553733247 Chr.20: 2325859-2325859 on Build GRCh38
rs969207126 Chr.20: 2326094-2326094 on Build GRCh38
rs765477797 Chr.20: 2325926-2325926 on Build GRCh38
rs760627673 Chr.20: 2325876-2325876 on Build GRCh38
rs764985984 Chr.20: 2325895-2325895 on Build GRCh38
rs758724894 Chr.20: 2325980-2325980 on Build GRCh38
rs1009712406 Chr.20: 2326205-2326205 on Build GRCh38
rs1023307162 Chr.20: 2326084-2326084 on Build GRCh38
rs986434016 Chr.20: 2325918-2325918 on Build GRCh38
rs776553784 Chr.20: 2325881-2325881 on Build GRCh38
rs141727243 Chr.20: 2325948-2325948 on Build GRCh38
rs574756177 Chr.20: 2326115-2326115 on Build GRCh38
rs148025061 Chr.20: 2325906-2325906 on Build GRCh38
rs1005531513 Chr.20: 2325966-2325966 on Build GRCh38
rs375694905 Chr.20: 2325903-2325903 on Build GRCh38
rs957179457 Chr.20: 2325869-2325869 on Build GRCh38
rs979211624 Chr.20: 2326097-2326097 on Build GRCh38
rs750786507 Chr.20: 2325911-2325911 on Build GRCh38
rs139599978 Chr.20: 2325907-2325907 on Build GRCh38
rs763581598 Chr.20: 2325992-2325992 on Build GRCh38
rs372653108 Chr.20: 2325887-2325887 on Build GRCh38
rs780274543 Chr.20: 2325924-2325924 on Build GRCh38
rs776643989 Chr.20: 2325957-2325957 on Build GRCh38
rs757614003 Chr.20: 2325822-2325822 on Build GRCh38
rs771005194 Chr.20: 2325952-2325952 on Build GRCh38
rs531002610 Chr.20: 2326127-2326127 on Build GRCh38
rs766342169 Chr.20: 2325880-2325880 on Build GRCh38
rs180916668 Chr.20: 2325996-2325996 on Build GRCh38
rs201826691 Chr.20: 2325981-2325981 on Build GRCh38
rs111244888 Chr.20: 2325900-2325900 on Build GRCh38
rs781508499 Chr.20: 2325823-2325823 on Build GRCh38
rs191199076 Chr.20: 2325936-2325936 on Build GRCh38
rs542098302 Chr.20: 2326118-2326118 on Build GRCh38
rs1013548298 Chr.20: 2326098-2326098 on Build GRCh38
rs45603442 Chr.20: 2326119-2326119 on Build GRCh38
rs1049256832 Chr.20: 2326221-2326221 on Build GRCh38
rs779644639 Chr.20: 2325989-2325990 on Build GRCh38
rs559716785 Chr.20: 2326040-2326040 on Build GRCh38
rs756427588 Chr.20: 2325919-2325919 on Build GRCh38
rs759436174 Chr.20: 2325965-2325965 on Build GRCh38
rs900677685 Chr.20: 2325922-2325922 on Build GRCh38
rs763641265 Chr.20: 2325902-2325902 on Build GRCh38
rs752355035 Chr.20: 2325896-2325896 on Build GRCh38
rs552365780 Chr.20: 2326209-2326209 on Build GRCh38
rs749501993 Chr.20: 2325934-2325934 on Build GRCh38
rs565808355 Chr.20: 2325870-2325870 on Build GRCh38
rs887170736 Chr.20: 2326044-2326044 on Build GRCh38
rs887934800 Chr.20: 2326235-2326235 on Build GRCh38
rs775618808 Chr.20: 2325838-2325838 on Build GRCh38
rs773069124 Chr.20: 2325873-2325873 on Build GRCh38
rs200497601 Chr.20: 2325935-2325935 on Build GRCh38
rs75608655 Chr.20: 2325979-2325979 on Build GRCh38
rs932033493 Chr.20: 2326175-2326175 on Build GRCh38
rs538219176 Chr.20: 2325829-2325829 on Build GRCh38
rs764401329 Chr.20: 2325812-2325812 on Build GRCh38
rs369189179 Chr.20: 2325830-2325830 on Build GRCh38
rs942554527 Chr.20: 2325943-2325943 on Build GRCh38
rs1032503082 Chr.20: 2326137-2326137 on Build GRCh38

View Full Product Details