Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2508793-2509284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF343
Entrez Gene ID
79175
Gene Name
zinc finger protein 343
Gene Aliases
dJ734P14.5
UniGene
Hs.516846
Gene Chromosome Location
Chr.20: 2481817-2524522 on Build GRCh38
Transcripts
NM_001282495.1 NM_001282496.1 NM_001282497.1 NM_001282498.1 NM_001282499.1 NM_001321800.1 NM_001321801.1 NM_001321802.1 NM_001321803.1 NM_001321805.1 NM_024325.5 XM_017028062.1

SNP Detail

SNP ID Chromosome Location
rs1037211178 Chr.20: 2509105-2509105 on Build GRCh38
rs895139808 Chr.20: 2509167-2509167 on Build GRCh38
rs186611839 Chr.20: 2509231-2509231 on Build GRCh38
rs942497534 Chr.20: 2509258-2509258 on Build GRCh38
rs1026317322 Chr.20: 2509095-2509095 on Build GRCh38
rs902602954 Chr.20: 2509230-2509230 on Build GRCh38
rs914986769 Chr.20: 2509180-2509181 on Build GRCh38
rs920912770 Chr.20: 2508981-2508981 on Build GRCh38
rs939640336 Chr.20: 2509109-2509109 on Build GRCh38
rs138860462 Chr.20: 2508869-2508869 on Build GRCh38
rs953720727 Chr.20: 2509061-2509061 on Build GRCh38
rs1010927883 Chr.20: 2508899-2508899 on Build GRCh38
rs369538080 Chr.20: 2508861-2508861 on Build GRCh38
rs879889197 Chr.20: 2509118-2509119 on Build GRCh38
rs889635673 Chr.20: 2508998-2508998 on Build GRCh38
rs551329556 Chr.20: 2509247-2509247 on Build GRCh38
rs369315705 Chr.20: 2508939-2508939 on Build GRCh38
rs976388718 Chr.20: 2508980-2508980 on Build GRCh38
rs982649929 Chr.20: 2508881-2508881 on Build GRCh38
rs1055330310 Chr.20: 2509138-2509138 on Build GRCh38
rs765431284 Chr.20: 2508817-2508817 on Build GRCh38
rs959515927 Chr.20: 2509123-2509123 on Build GRCh38
rs950999871 Chr.20: 2509101-2509101 on Build GRCh38
rs909672687 Chr.20: 2509074-2509074 on Build GRCh38
rs753216703 Chr.20: 2508821-2508821 on Build GRCh38
rs755786001 Chr.20: 2508888-2508888 on Build GRCh38
rs968950446 Chr.20: 2508916-2508916 on Build GRCh38
rs573511157 Chr.20: 2508839-2508839 on Build GRCh38
rs946524498 Chr.20: 2509217-2509217 on Build GRCh38
rs573897255 Chr.20: 2509252-2509252 on Build GRCh38
rs995244266 Chr.20: 2509090-2509090 on Build GRCh38
rs951036635 Chr.20: 2508875-2508875 on Build GRCh38
rs527852786 Chr.20: 2509081-2509081 on Build GRCh38
rs879132056 Chr.20: 2509129-2509129 on Build GRCh38
rs531987260 Chr.20: 2509198-2509198 on Build GRCh38
rs1037215722 Chr.20: 2509135-2509135 on Build GRCh38
rs895211739 Chr.20: 2508889-2508889 on Build GRCh38
rs551635344 Chr.20: 2508852-2508852 on Build GRCh38
rs560303450 Chr.20: 2508955-2508955 on Build GRCh38
rs6138281 Chr.20: 2508885-2508885 on Build GRCh38
rs919490352 Chr.20: 2508872-2508872 on Build GRCh38
rs1007624988 Chr.20: 2509059-2509059 on Build GRCh38
rs1031519364 Chr.20: 2509232-2509232 on Build GRCh38
rs140169124 Chr.20: 2509069-2509069 on Build GRCh38
rs561455527 Chr.20: 2509108-2509108 on Build GRCh38
rs149408215 Chr.20: 2508919-2508919 on Build GRCh38
rs902423912 Chr.20: 2508958-2508958 on Build GRCh38
rs1010896909 Chr.20: 2509210-2509210 on Build GRCh38
rs1035129777 Chr.20: 2509119-2509119 on Build GRCh38
rs181121203 Chr.20: 2508854-2508854 on Build GRCh38
rs532900460 Chr.20: 2509242-2509242 on Build GRCh38
rs928178378 Chr.20: 2509106-2509106 on Build GRCh38
rs76165531 Chr.20: 2508975-2508975 on Build GRCh38
rs758870667 Chr.20: 2508844-2508844 on Build GRCh38
rs978119873 Chr.20: 2509227-2509227 on Build GRCh38
rs748951284 Chr.20: 2508959-2508959 on Build GRCh38
rs1022365278 Chr.20: 2508903-2508903 on Build GRCh38
rs771964806 Chr.20: 2509228-2509228 on Build GRCh38
rs748043714 Chr.20: 2509190-2509190 on Build GRCh38
rs746717794 Chr.20: 2509251-2509251 on Build GRCh38
rs778423610 Chr.20: 2509089-2509089 on Build GRCh38
rs6138282 Chr.20: 2508949-2508949 on Build GRCh38
rs1007654663 Chr.20: 2508828-2508828 on Build GRCh38

View Full Product Details