Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2592219-2592718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC2
Entrez Gene ID
117532
Gene Name
transmembrane channel like 2
Gene Aliases
C20orf145
UniGene
Hs.352626
Gene Chromosome Location
Chr.20: 2536607-2642185 on Build GRCh38
Transcripts
NM_080751.2 XM_005260660.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5365965 c.835-1G>T p.? Chr.20: 2592309-2592309 on Build GRCh38
3544915 c.918C>T p.V306V Chr.20: 2592393-2592393 on Build GRCh38
6438542 c.906_907GG>AA p.M302_D303>IN Chr.20: 2592381-2592382 on Build GRCh38
1229452 c.900G>T p.K300N Chr.20: 2592375-2592375 on Build GRCh38
1229453 c.912T>G p.F304L Chr.20: 2592387-2592387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569849494 Chr.20: 2592320-2592320 on Build GRCh38
rs780278819 Chr.20: 2592414-2592414 on Build GRCh38
rs939725398 Chr.20: 2592474-2592474 on Build GRCh38
rs773873202 Chr.20: 2592264-2592264 on Build GRCh38
rs765161437 Chr.20: 2592393-2592393 on Build GRCh38
rs774736293 Chr.20: 2592355-2592355 on Build GRCh38
rs868253071 Chr.20: 2592383-2592383 on Build GRCh38
rs772680064 Chr.20: 2592452-2592452 on Build GRCh38
rs748594078 Chr.20: 2592433-2592433 on Build GRCh38
rs946700333 Chr.20: 2592534-2592534 on Build GRCh38
rs370293293 Chr.20: 2592457-2592457 on Build GRCh38
rs972516855 Chr.20: 2592475-2592475 on Build GRCh38
rs750053287 Chr.20: 2592279-2592279 on Build GRCh38
rs997814971 Chr.20: 2592256-2592257 on Build GRCh38
rs548042845 Chr.20: 2592272-2592272 on Build GRCh38
rs1016418490 Chr.20: 2592558-2592558 on Build GRCh38
rs767473458 Chr.20: 2592407-2592407 on Build GRCh38
rs757609571 Chr.20: 2592408-2592408 on Build GRCh38
rs868391384 Chr.20: 2592489-2592489 on Build GRCh38
rs779404903 Chr.20: 2592432-2592432 on Build GRCh38
rs766039682 Chr.20: 2592412-2592412 on Build GRCh38
rs902075176 Chr.20: 2592251-2592251 on Build GRCh38
rs202220836 Chr.20: 2592415-2592415 on Build GRCh38
rs141669275 Chr.20: 2592344-2592344 on Build GRCh38
rs758426455 Chr.20: 2592406-2592406 on Build GRCh38
rs141680917 Chr.20: 2592322-2592322 on Build GRCh38
rs6083778 Chr.20: 2592309-2592309 on Build GRCh38
rs946176356 Chr.20: 2592478-2592478 on Build GRCh38
rs993300686 Chr.20: 2592578-2592578 on Build GRCh38
rs552668921 Chr.20: 2592368-2592368 on Build GRCh38
rs771482195 Chr.20: 2592362-2592362 on Build GRCh38
rs753012129 Chr.20: 2592280-2592280 on Build GRCh38
rs980719080 Chr.20: 2592425-2592425 on Build GRCh38
rs112491668 Chr.20: 2592329-2592329 on Build GRCh38
rs58180900 Chr.20: 2592505-2592506 on Build GRCh38
rs184347079 Chr.20: 2592269-2592269 on Build GRCh38
rs760445896 Chr.20: 2592289-2592289 on Build GRCh38
rs766165467 Chr.20: 2592378-2592378 on Build GRCh38
rs373534707 Chr.20: 2592412-2592412 on Build GRCh38
rs139309364 Chr.20: 2592588-2592588 on Build GRCh38
rs777613688 Chr.20: 2592644-2592644 on Build GRCh38
rs780366532 Chr.20: 2592342-2592342 on Build GRCh38
rs543133587 Chr.20: 2592682-2592682 on Build GRCh38
rs752579525 Chr.20: 2592400-2592400 on Build GRCh38
rs776523499 Chr.20: 2592384-2592384 on Build GRCh38
rs149529222 Chr.20: 2592683-2592683 on Build GRCh38
rs752278399 Chr.20: 2592575-2592575 on Build GRCh38
rs760376857 Chr.20: 2592371-2592371 on Build GRCh38
rs531374596 Chr.20: 2592597-2592597 on Build GRCh38
rs753732094 Chr.20: 2592302-2592302 on Build GRCh38
rs895436416 Chr.20: 2592507-2592507 on Build GRCh38
rs1019048159 Chr.20: 2592589-2592589 on Build GRCh38
rs181141326 Chr.20: 2592678-2592678 on Build GRCh38
rs983784572 Chr.20: 2592443-2592443 on Build GRCh38
rs759423889 Chr.20: 2592387-2592387 on Build GRCh38
rs753610525 Chr.20: 2592451-2592451 on Build GRCh38
rs928091413 Chr.20: 2592476-2592476 on Build GRCh38
rs138588376 Chr.20: 2592244-2592244 on Build GRCh38
rs779022296 Chr.20: 2592310-2592310 on Build GRCh38
rs1050597052 Chr.20: 2592505-2592505 on Build GRCh38
rs900983974 Chr.20: 2592593-2592593 on Build GRCh38
rs377153411 Chr.20: 2592358-2592358 on Build GRCh38
rs190017588 Chr.20: 2592610-2592610 on Build GRCh38
rs1042105250 Chr.20: 2592485-2592485 on Build GRCh38
rs766260252 Chr.20: 2592296-2592296 on Build GRCh38
rs570845091 Chr.20: 2592418-2592418 on Build GRCh38
rs751607324 Chr.20: 2592317-2592317 on Build GRCh38
rs146231540 Chr.20: 2592542-2592542 on Build GRCh38
rs7353668 Chr.20: 2592642-2592642 on Build GRCh38
rs939516517 Chr.20: 2592488-2592488 on Build GRCh38
rs756376854 Chr.20: 2592333-2592333 on Build GRCh38
rs769104197 Chr.20: 2592349-2592349 on Build GRCh38
rs750556547 Chr.20: 2592409-2592409 on Build GRCh38
rs1015945273 Chr.20: 2592359-2592359 on Build GRCh38
rs775597370 Chr.20: 2592382-2592382 on Build GRCh38
rs754932424 Chr.20: 2592303-2592303 on Build GRCh38

View Full Product Details