Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3213166-3213664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPA
Entrez Gene ID
3704
Gene Name
inosine triphosphatase
Gene Aliases
C20orf37, HLC14-06-P, ITPase, My049, NTPase, dJ794I6.3
UniGene
Hs.415299
Gene Chromosome Location
Chr.20: 3208868-3227449 on Build GRCh38
Transcripts
NM_001267623.1 NM_001324236.1 NM_001324237.1 NM_001324238.1 NM_001324240.1 NM_033453.3 NM_181493.3 NR_052000.1 NR_052001.2 NR_052002.1 XM_006723564.3 XM_006723565.3 XM_011529234.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1411196 c.128C>T p.P43L Chr.20: 3213322-3213322 on Build GRCh38
4136827 c.92T>G p.F31C Chr.20: 3213194-3213194 on Build GRCh38
149130 c.94C>A p.P32T Chr.20: 3213196-3213196 on Build GRCh38
236456 c.73C>A p.Q25K Chr.20: 3213175-3213175 on Build GRCh38
4514026 c.95C>T p.P32L Chr.20: 3213197-3213197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369545461 Chr.20: 3213270-3213270 on Build GRCh38
rs773069391 Chr.20: 3213212-3213212 on Build GRCh38
rs138760860 Chr.20: 3213364-3213364 on Build GRCh38
rs924079029 Chr.20: 3213620-3213620 on Build GRCh38
rs376142053 Chr.20: 3213227-3213227 on Build GRCh38
rs1127354 Chr.20: 3213196-3213196 on Build GRCh38
rs965761182 Chr.20: 3213248-3213248 on Build GRCh38
rs780117445 Chr.20: 3213529-3213529 on Build GRCh38
rs560426260 Chr.20: 3213445-3213445 on Build GRCh38
rs749907162 Chr.20: 3213487-3213487 on Build GRCh38
rs772877398 Chr.20: 3213280-3213280 on Build GRCh38
rs1006005169 Chr.20: 3213498-3213498 on Build GRCh38
rs778925537 Chr.20: 3213548-3213548 on Build GRCh38
rs780431801 Chr.20: 3213319-3213319 on Build GRCh38
rs567367339 Chr.20: 3213473-3213473 on Build GRCh38
rs555480105 Chr.20: 3213195-3213195 on Build GRCh38
rs201934888 Chr.20: 3213292-3213292 on Build GRCh38
rs766279705 Chr.20: 3213229-3213230 on Build GRCh38
rs201119006 Chr.20: 3213378-3213378 on Build GRCh38
rs199769023 Chr.20: 3213202-3213202 on Build GRCh38
rs779332687 Chr.20: 3213630-3213630 on Build GRCh38
rs754257362 Chr.20: 3213407-3213407 on Build GRCh38
rs754811147 Chr.20: 3213603-3213603 on Build GRCh38
rs761938281 Chr.20: 3213302-3213302 on Build GRCh38
rs902707295 Chr.20: 3213435-3213435 on Build GRCh38
rs751721755 Chr.20: 3213301-3213301 on Build GRCh38
rs773947334 Chr.20: 3213203-3213203 on Build GRCh38
rs756551860 Chr.20: 3213257-3213257 on Build GRCh38
rs746662660 Chr.20: 3213345-3213345 on Build GRCh38
rs780589450 Chr.20: 3213260-3213260 on Build GRCh38
rs370769779 Chr.20: 3213276-3213276 on Build GRCh38
rs939381685 Chr.20: 3213353-3213353 on Build GRCh38
rs996018119 Chr.20: 3213621-3213621 on Build GRCh38
rs551410988 Chr.20: 3213421-3213421 on Build GRCh38
rs951769271 Chr.20: 3213495-3213495 on Build GRCh38
rs747694061 Chr.20: 3213277-3213277 on Build GRCh38
rs750870460 Chr.20: 3213255-3213255 on Build GRCh38
rs760558176 Chr.20: 3213282-3213282 on Build GRCh38
rs767824484 Chr.20: 3213303-3213303 on Build GRCh38
rs549041787 Chr.20: 3213462-3213462 on Build GRCh38
rs774267237 Chr.20: 3213376-3213376 on Build GRCh38
rs767520628 Chr.20: 3213379-3213379 on Build GRCh38
rs1035620403 Chr.20: 3213547-3213547 on Build GRCh38
rs768112087 Chr.20: 3213267-3213267 on Build GRCh38
rs764213076 Chr.20: 3213300-3213300 on Build GRCh38
rs199623719 Chr.20: 3213279-3213279 on Build GRCh38
rs777528511 Chr.20: 3213341-3213341 on Build GRCh38
rs756425674 Chr.20: 3213318-3213318 on Build GRCh38
rs550362633 Chr.20: 3213525-3213525 on Build GRCh38
rs765811885 Chr.20: 3213433-3213433 on Build GRCh38
rs1000166523 Chr.20: 3213459-3213459 on Build GRCh38
rs946787934 Chr.20: 3213614-3213614 on Build GRCh38
rs761555605 Chr.20: 3213205-3213205 on Build GRCh38
rs990777489 Chr.20: 3213602-3213602 on Build GRCh38
rs1053551133 Chr.20: 3213584-3213584 on Build GRCh38
rs757955055 Chr.20: 3213340-3213340 on Build GRCh38
rs368434084 Chr.20: 3213231-3213231 on Build GRCh38
rs777029172 Chr.20: 3213236-3213240 on Build GRCh38
rs142979373 Chr.20: 3213322-3213322 on Build GRCh38
rs35991941 Chr.20: 3213591-3213591 on Build GRCh38
rs995847790 Chr.20: 3213489-3213489 on Build GRCh38
rs200947572 Chr.20: 3213239-3213239 on Build GRCh38
rs776582353 Chr.20: 3213355-3213355 on Build GRCh38
rs200506201 Chr.20: 3213294-3213294 on Build GRCh38
rs537947721 Chr.20: 3213494-3213494 on Build GRCh38
rs369860957 Chr.20: 3213394-3213394 on Build GRCh38
rs759552310 Chr.20: 3213295-3213295 on Build GRCh38
rs773447998 Chr.20: 3213387-3213387 on Build GRCh38
rs7270101 Chr.20: 3213247-3213247 on Build GRCh38
rs936240807 Chr.20: 3213578-3213578 on Build GRCh38
rs752125365 Chr.20: 3213434-3213434 on Build GRCh38
rs146112146 Chr.20: 3213323-3213323 on Build GRCh38
rs979562541 Chr.20: 3213616-3213616 on Build GRCh38
rs538432614 Chr.20: 3213219-3213219 on Build GRCh38
rs770808787 Chr.20: 3213354-3213354 on Build GRCh38
rs980202804 Chr.20: 3213538-3213538 on Build GRCh38
rs8362 Chr.20: 3213332-3213332 on Build GRCh38
rs771694985 Chr.20: 3213209-3213209 on Build GRCh38
rs1015154000 Chr.20: 3213503-3213503 on Build GRCh38
rs113777460 Chr.20: 3213233-3213233 on Build GRCh38
rs749819594 Chr.20: 3213261-3213261 on Build GRCh38
rs67002563 Chr.20: 3213527-3213527 on Build GRCh38
rs1043864593 Chr.20: 3213429-3213429 on Build GRCh38
rs111757940 Chr.20: 3213452-3213452 on Build GRCh38
rs374874902 Chr.20: 3213307-3213307 on Build GRCh38
rs769920670 Chr.20: 3213368-3213368 on Build GRCh38
rs766668623 Chr.20: 3213397-3213397 on Build GRCh38
rs142574480 Chr.20: 3213226-3213226 on Build GRCh38
rs144682597 Chr.20: 3213198-3213198 on Build GRCh38
rs545393698 Chr.20: 3213237-3213237 on Build GRCh38

View Full Product Details