Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74199535-74200046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5039815 c.100G>A p.G34R Chr.1: 74199681-74199681 on Build GRCh38
6991168 c.215A>G p.Y72C Chr.1: 74199796-74199796 on Build GRCh38
5930546 c.206G>A p.G69E Chr.1: 74199787-74199787 on Build GRCh38
912065 c.97C>T p.R33C Chr.1: 74199678-74199678 on Build GRCh38
682195 c.99T>C p.R33R Chr.1: 74199680-74199680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199556922 Chr.1: 74199837-74199837 on Build GRCh38
rs540870571 Chr.1: 74199946-74199946 on Build GRCh38
rs201158336 Chr.1: 74199713-74199713 on Build GRCh38
rs200281954 Chr.1: 74199722-74199722 on Build GRCh38
rs779095739 Chr.1: 74199859-74199859 on Build GRCh38
rs944123337 Chr.1: 74199933-74199933 on Build GRCh38
rs35676124 Chr.1: 74199655-74199655 on Build GRCh38
rs757805088 Chr.1: 74199743-74199743 on Build GRCh38
rs998594096 Chr.1: 74199797-74199797 on Build GRCh38
rs760487337 Chr.1: 74199705-74199705 on Build GRCh38
rs757200610 Chr.1: 74199872-74199872 on Build GRCh38
rs186399715 Chr.1: 74199686-74199686 on Build GRCh38
rs202243065 Chr.1: 74199824-74199824 on Build GRCh38
rs200359521 Chr.1: 74199784-74199784 on Build GRCh38
rs2069302 Chr.1: 74199923-74199923 on Build GRCh38
rs553179730 Chr.1: 74199959-74199959 on Build GRCh38
rs774998036 Chr.1: 74199829-74199829 on Build GRCh38
rs922642773 Chr.1: 74199998-74199998 on Build GRCh38
rs749592241 Chr.1: 74199774-74199774 on Build GRCh38
rs984711300 Chr.1: 74199907-74199907 on Build GRCh38
rs752130584 Chr.1: 74199622-74199622 on Build GRCh38
rs200157650 Chr.1: 74199718-74199718 on Build GRCh38
rs573738199 Chr.1: 74199931-74199931 on Build GRCh38
rs748855968 Chr.1: 74199889-74199889 on Build GRCh38
rs780733130 Chr.1: 74199974-74199974 on Build GRCh38
rs769419272 Chr.1: 74199813-74199813 on Build GRCh38
rs959016712 Chr.1: 74199751-74199751 on Build GRCh38
rs1053943460 Chr.1: 74199976-74199976 on Build GRCh38
rs948901627 Chr.1: 74199659-74199659 on Build GRCh38
rs745974687 Chr.1: 74199668-74199668 on Build GRCh38
rs138015421 Chr.1: 74199832-74199832 on Build GRCh38
rs748259400 Chr.1: 74199863-74199863 on Build GRCh38
rs746912279 Chr.1: 74199792-74199792 on Build GRCh38
rs977103552 Chr.1: 74199594-74199594 on Build GRCh38
rs200882722 Chr.1: 74199712-74199712 on Build GRCh38
rs750779015 Chr.1: 74199747-74199747 on Build GRCh38
rs780370068 Chr.1: 74199679-74199679 on Build GRCh38
rs377237678 Chr.1: 74199788-74199788 on Build GRCh38
rs537311556 Chr.1: 74199881-74199881 on Build GRCh38
rs753986181 Chr.1: 74199843-74199843 on Build GRCh38
rs902700368 Chr.1: 74199952-74199952 on Build GRCh38
rs202113178 Chr.1: 74199702-74199702 on Build GRCh38
rs754999716 Chr.1: 74199783-74199783 on Build GRCh38
rs792310 Chr.1: 74199809-74199809 on Build GRCh38
rs780145871 Chr.1: 74199772-74199772 on Build GRCh38
rs201001853 Chr.1: 74199719-74199719 on Build GRCh38
rs933271866 Chr.1: 74199768-74199768 on Build GRCh38
rs936869212 Chr.1: 74199968-74199968 on Build GRCh38
rs200074343 Chr.1: 74199630-74199630 on Build GRCh38
rs370987974 Chr.1: 74199868-74199868 on Build GRCh38
rs368872427 Chr.1: 74199802-74199802 on Build GRCh38
rs757769988 Chr.1: 74199628-74199628 on Build GRCh38
rs573630971 Chr.1: 74199902-74199902 on Build GRCh38
rs777988597 Chr.1: 74199613-74199613 on Build GRCh38
rs750735574 Chr.1: 74199838-74199838 on Build GRCh38
rs776517136 Chr.1: 74199800-74199800 on Build GRCh38
rs909967771 Chr.1: 74199696-74199696 on Build GRCh38
rs373139291 Chr.1: 74199835-74199835 on Build GRCh38
rs866488007 Chr.1: 74199966-74199966 on Build GRCh38
rs200510819 Chr.1: 74199839-74199839 on Build GRCh38
rs924258137 Chr.1: 74199603-74199603 on Build GRCh38
rs763672829 Chr.1: 74199733-74199733 on Build GRCh38
rs769458921 Chr.1: 74199883-74199883 on Build GRCh38
rs146451796 Chr.1: 74199795-74199795 on Build GRCh38
rs150277198 Chr.1: 74199729-74199729 on Build GRCh38
rs200086357 Chr.1: 74199689-74199689 on Build GRCh38
rs202078895 Chr.1: 74199776-74199776 on Build GRCh38
rs775400521 Chr.1: 74199884-74199884 on Build GRCh38
rs771967355 Chr.1: 74199693-74199693 on Build GRCh38
rs1021434754 Chr.1: 74199789-74199789 on Build GRCh38
rs377443940 Chr.1: 74199949-74199949 on Build GRCh38
rs763823114 Chr.1: 74199825-74199825 on Build GRCh38
rs370760053 Chr.1: 74199749-74199749 on Build GRCh38
rs1049823471 Chr.1: 74199593-74199593 on Build GRCh38
rs377042126 Chr.1: 74199678-74199678 on Build GRCh38
rs201351675 Chr.1: 74199836-74199836 on Build GRCh38
rs932695763 Chr.1: 74199575-74199575 on Build GRCh38
rs190966349 Chr.1: 74199846-74199846 on Build GRCh38
rs375572507 Chr.1: 74199612-74199612 on Build GRCh38
rs752044745 Chr.1: 74199735-74199735 on Build GRCh38
rs763798519 Chr.1: 74199813-74199813 on Build GRCh38
rs1001088562 Chr.1: 74199744-74199744 on Build GRCh38
rs373335896 Chr.1: 74199894-74199894 on Build GRCh38

View Full Product Details