Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74205720-74206263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202686 c.1734delT p.Y579fs*7 Chr.1: 74205781-74205781 on Build GRCh38
4009840 c.1721A>G p.E574G Chr.1: 74205768-74205768 on Build GRCh38
1185487 c.1739G>T p.R580M Chr.1: 74205786-74205786 on Build GRCh38
1665476 c.1690delA p.I564fs*10 Chr.1: 74205737-74205737 on Build GRCh38
4652867 c.1762A>G p.S588G Chr.1: 74205809-74205809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777890640 Chr.1: 74206000-74206000 on Build GRCh38
rs774315921 Chr.1: 74205790-74205790 on Build GRCh38
rs994657067 Chr.1: 74206100-74206100 on Build GRCh38
rs895471268 Chr.1: 74205945-74205945 on Build GRCh38
rs768537296 Chr.1: 74205789-74205789 on Build GRCh38
rs776562252 Chr.1: 74206123-74206123 on Build GRCh38
rs964603151 Chr.1: 74206138-74206138 on Build GRCh38
rs754270559 Chr.1: 74205828-74205828 on Build GRCh38
rs897271392 Chr.1: 74206079-74206079 on Build GRCh38
rs955856549 Chr.1: 74206216-74206216 on Build GRCh38
rs779945629 Chr.1: 74206007-74206007 on Build GRCh38
rs879131143 Chr.1: 74205881-74205881 on Build GRCh38
rs1038483519 Chr.1: 74206069-74206069 on Build GRCh38
rs988971326 Chr.1: 74206218-74206218 on Build GRCh38
rs985561787 Chr.1: 74205858-74205858 on Build GRCh38
rs947799693 Chr.1: 74205988-74205988 on Build GRCh38
rs781369221 Chr.1: 74205854-74205854 on Build GRCh38
rs879216185 Chr.1: 74205884-74205884 on Build GRCh38
rs775272386 Chr.1: 74205784-74205784 on Build GRCh38
rs1044637431 Chr.1: 74205971-74205971 on Build GRCh38
rs778100815 Chr.1: 74205792-74205793 on Build GRCh38
rs767286333 Chr.1: 74205809-74205809 on Build GRCh38
rs553778822 Chr.1: 74205968-74205968 on Build GRCh38
rs755511609 Chr.1: 74205829-74205829 on Build GRCh38
rs760095046 Chr.1: 74205825-74205825 on Build GRCh38
rs755892661 Chr.1: 74205872-74205872 on Build GRCh38
rs539707389 Chr.1: 74206161-74206161 on Build GRCh38
rs944231942 Chr.1: 74205888-74205889 on Build GRCh38
rs113617272 Chr.1: 74205979-74205979 on Build GRCh38
rs749292769 Chr.1: 74205787-74205787 on Build GRCh38
rs943195384 Chr.1: 74205778-74205778 on Build GRCh38
rs906347091 Chr.1: 74206060-74206060 on Build GRCh38
rs533409144 Chr.1: 74206030-74206030 on Build GRCh38
rs749136899 Chr.1: 74205868-74205868 on Build GRCh38
rs562800576 Chr.1: 74205772-74205772 on Build GRCh38
rs757258443 Chr.1: 74205851-74205851 on Build GRCh38
rs780042474 Chr.1: 74205807-74205807 on Build GRCh38
rs777770818 Chr.1: 74205747-74205747 on Build GRCh38
rs770863988 Chr.1: 74206085-74206085 on Build GRCh38
rs571848 Chr.1: 74205835-74205835 on Build GRCh38
rs906097253 Chr.1: 74206024-74206024 on Build GRCh38
rs954071395 Chr.1: 74206157-74206157 on Build GRCh38
rs916973246 Chr.1: 74206227-74206227 on Build GRCh38
rs535570007 Chr.1: 74205837-74205837 on Build GRCh38
rs955564598 Chr.1: 74206115-74206115 on Build GRCh38
rs371947111 Chr.1: 74205821-74205821 on Build GRCh38
rs949684533 Chr.1: 74205949-74205949 on Build GRCh38
rs934390746 Chr.1: 74205913-74205913 on Build GRCh38
rs978566745 Chr.1: 74205805-74205805 on Build GRCh38
rs372918657 Chr.1: 74205799-74205800 on Build GRCh38
rs565699819 Chr.1: 74206128-74206128 on Build GRCh38
rs144922146 Chr.1: 74206207-74206207 on Build GRCh38
rs768733792 Chr.1: 74205883-74205883 on Build GRCh38
rs768571602 Chr.1: 74205962-74205962 on Build GRCh38
rs1017550193 Chr.1: 74206129-74206129 on Build GRCh38
rs545430654 Chr.1: 74206126-74206127 on Build GRCh38
rs771108793 Chr.1: 74205774-74205774 on Build GRCh38
rs777967231 Chr.1: 74205838-74205838 on Build GRCh38
rs747313963 Chr.1: 74205848-74205848 on Build GRCh38
rs936298998 Chr.1: 74205967-74205967 on Build GRCh38
rs576141929 Chr.1: 74206197-74206197 on Build GRCh38
rs1052825834 Chr.1: 74205941-74205941 on Build GRCh38
rs879634177 Chr.1: 74205993-74205994 on Build GRCh38
rs949735005 Chr.1: 74206241-74206241 on Build GRCh38
rs747063121 Chr.1: 74205773-74205773 on Build GRCh38
rs1029706011 Chr.1: 74206105-74206105 on Build GRCh38
rs149826787 Chr.1: 74206189-74206189 on Build GRCh38
rs978617817 Chr.1: 74206140-74206140 on Build GRCh38
rs765963504 Chr.1: 74205826-74205826 on Build GRCh38

View Full Product Details