Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 4726255-4726767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRND
Entrez Gene ID
23627
Gene Name
prion protein 2 (dublet)
Gene Aliases
DOPPEL, DPL, PrPLP, dJ1068H6.4
UniGene
Hs.406696
Gene Chromosome Location
Chr.20: 4721854-4728462 on Build GRCh38
Transcripts
NM_012409.3

SNP Detail

SNP ID Chromosome Location
rs748401047 Chr.20: 4726688-4726688 on Build GRCh38
rs968646646 Chr.20: 4726707-4726707 on Build GRCh38
rs924196626 Chr.20: 4726519-4726519 on Build GRCh38
rs184698913 Chr.20: 4726624-4726624 on Build GRCh38
rs901045770 Chr.20: 4726353-4726353 on Build GRCh38
rs543675665 Chr.20: 4726589-4726589 on Build GRCh38
rs561401491 Chr.20: 4726535-4726535 on Build GRCh38
rs879073066 Chr.20: 4726645-4726645 on Build GRCh38
rs558923874 Chr.20: 4726629-4726629 on Build GRCh38
rs980132914 Chr.20: 4726514-4726514 on Build GRCh38
rs1051638044 Chr.20: 4726318-4726318 on Build GRCh38
rs781588014 Chr.20: 4726473-4726473 on Build GRCh38
rs757911737 Chr.20: 4726309-4726309 on Build GRCh38
rs889544931 Chr.20: 4726391-4726391 on Build GRCh38
rs188805797 Chr.20: 4726495-4726495 on Build GRCh38
rs993010843 Chr.20: 4726488-4726488 on Build GRCh38
rs569495029 Chr.20: 4726315-4726315 on Build GRCh38
rs184448336 Chr.20: 4726393-4726393 on Build GRCh38
rs896856823 Chr.20: 4726627-4726627 on Build GRCh38
rs554217942 Chr.20: 4726453-4726453 on Build GRCh38
rs538977298 Chr.20: 4726439-4726439 on Build GRCh38
rs1027563944 Chr.20: 4726401-4726401 on Build GRCh38
rs2735675 Chr.20: 4726742-4726742 on Build GRCh38
rs534115152 Chr.20: 4726696-4726696 on Build GRCh38
rs934604539 Chr.20: 4726350-4726350 on Build GRCh38
rs1049495815 Chr.20: 4726368-4726368 on Build GRCh38
rs529671517 Chr.20: 4726695-4726695 on Build GRCh38
rs968180910 Chr.20: 4726504-4726504 on Build GRCh38
rs889575921 Chr.20: 4726610-4726610 on Build GRCh38
rs941121698 Chr.20: 4726623-4726623 on Build GRCh38
rs376598924 Chr.20: 4726478-4726478 on Build GRCh38
rs1053439095 Chr.20: 4726352-4726352 on Build GRCh38
rs547867321 Chr.20: 4726725-4726725 on Build GRCh38
rs191833035 Chr.20: 4726600-4726600 on Build GRCh38
rs569669264 Chr.20: 4726733-4726733 on Build GRCh38
rs997957462 Chr.20: 4726367-4726367 on Build GRCh38
rs1016602551 Chr.20: 4726400-4726400 on Build GRCh38
rs537688363 Chr.20: 4726601-4726601 on Build GRCh38
rs1004540729 Chr.20: 4726447-4726447 on Build GRCh38
rs192334960 Chr.20: 4726282-4726282 on Build GRCh38
rs896123699 Chr.20: 4726629-4726633 on Build GRCh38
rs577086482 Chr.20: 4726421-4726421 on Build GRCh38
rs1023118691 Chr.20: 4726518-4726518 on Build GRCh38
rs1005184650 Chr.20: 4726620-4726620 on Build GRCh38
rs923974197 Chr.20: 4726306-4726306 on Build GRCh38
rs922498067 Chr.20: 4726570-4726570 on Build GRCh38
rs143548564 Chr.20: 4726655-4726656 on Build GRCh38
rs537917440 Chr.20: 4726525-4726525 on Build GRCh38
rs1014478283 Chr.20: 4726722-4726722 on Build GRCh38
rs925909283 Chr.20: 4726333-4726333 on Build GRCh38
rs6139531 Chr.20: 4726420-4726420 on Build GRCh38
rs4815731 Chr.20: 4726466-4726466 on Build GRCh38
rs948757263 Chr.20: 4726307-4726307 on Build GRCh38
rs1011987564 Chr.20: 4726630-4726630 on Build GRCh38
rs6139532 Chr.20: 4726546-4726546 on Build GRCh38
rs900298028 Chr.20: 4726392-4726392 on Build GRCh38
rs926011523 Chr.20: 4726540-4726540 on Build GRCh38
rs942731040 Chr.20: 4726381-4726381 on Build GRCh38
rs147589194 Chr.20: 4726561-4726561 on Build GRCh38
rs1026594180 Chr.20: 4726692-4726692 on Build GRCh38
rs1038903781 Chr.20: 4726384-4726384 on Build GRCh38
rs548498557 Chr.20: 4726594-4726594 on Build GRCh38
rs896091155 Chr.20: 4726719-4726719 on Build GRCh38

View Full Product Details