Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 13110034-13110536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTLC3
Entrez Gene ID
55304
Gene Name
serine palmitoyltransferase long chain base subunit 3
Gene Aliases
C20orf38, LCB2B, LCB3, SPT3, SPTLC2L, dJ718P11, dJ718P11.1, hLCB2b
UniGene
Hs.425023
Gene Chromosome Location
Chr.20: 13008979-13169103 on Build GRCh38
Transcripts
NM_018327.2 XM_011529279.1 XM_011529280.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842439 c.841G>C p.E281Q Chr.20: 13110126-13110126 on Build GRCh38
6046881 c.911T>A p.I304N Chr.20: 13110196-13110196 on Build GRCh38
3544137 c.853A>G p.R285G Chr.20: 13110138-13110138 on Build GRCh38
3544138 c.881G>A p.R294Q Chr.20: 13110166-13110166 on Build GRCh38
3840426 c.852G>T p.L284L Chr.20: 13110137-13110137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772648914 Chr.20: 13110255-13110255 on Build GRCh38
rs779454871 Chr.20: 13110173-13110173 on Build GRCh38
rs755263203 Chr.20: 13110099-13110099 on Build GRCh38
rs530438541 Chr.20: 13110378-13110378 on Build GRCh38
rs757313209 Chr.20: 13110166-13110166 on Build GRCh38
rs920209879 Chr.20: 13110225-13110225 on Build GRCh38
rs1008812619 Chr.20: 13110483-13110483 on Build GRCh38
rs111709563 Chr.20: 13110507-13110507 on Build GRCh38
rs781771097 Chr.20: 13110141-13110141 on Build GRCh38
rs6109712 Chr.20: 13110081-13110081 on Build GRCh38
rs61746220 Chr.20: 13110172-13110172 on Build GRCh38
rs367557160 Chr.20: 13110197-13110197 on Build GRCh38
rs752285773 Chr.20: 13110212-13110212 on Build GRCh38
rs763450452 Chr.20: 13110087-13110088 on Build GRCh38
rs576470827 Chr.20: 13110086-13110087 on Build GRCh38
rs201594440 Chr.20: 13110166-13110166 on Build GRCh38
rs151049361 Chr.20: 13110403-13110403 on Build GRCh38
rs994781569 Chr.20: 13110240-13110240 on Build GRCh38
rs375575863 Chr.20: 13110087-13110087 on Build GRCh38
rs912701196 Chr.20: 13110432-13110432 on Build GRCh38
rs777507010 Chr.20: 13110152-13110152 on Build GRCh38
rs545282580 Chr.20: 13110433-13110433 on Build GRCh38
rs900466803 Chr.20: 13110510-13110510 on Build GRCh38
rs1024743208 Chr.20: 13110347-13110347 on Build GRCh38
rs549696363 Chr.20: 13110244-13110244 on Build GRCh38
rs921038575 Chr.20: 13110488-13110488 on Build GRCh38
rs751573042 Chr.20: 13110109-13110110 on Build GRCh38
rs778511213 Chr.20: 13110286-13110286 on Build GRCh38
rs538456455 Chr.20: 13110314-13110314 on Build GRCh38
rs547828339 Chr.20: 13110171-13110171 on Build GRCh38
rs958965089 Chr.20: 13110183-13110183 on Build GRCh38
rs773733355 Chr.20: 13110106-13110106 on Build GRCh38
rs199832772 Chr.20: 13110158-13110158 on Build GRCh38
rs140202361 Chr.20: 13110065-13110065 on Build GRCh38
rs537592072 Chr.20: 13110249-13110249 on Build GRCh38
rs764700149 Chr.20: 13110204-13110204 on Build GRCh38
rs190409577 Chr.20: 13110382-13110382 on Build GRCh38
rs189490985 Chr.20: 13110132-13110132 on Build GRCh38
rs1032708658 Chr.20: 13110117-13110117 on Build GRCh38
rs924218127 Chr.20: 13110374-13110374 on Build GRCh38
rs372257304 Chr.20: 13110245-13110245 on Build GRCh38
rs1039480175 Chr.20: 13110271-13110271 on Build GRCh38
rs181963687 Chr.20: 13110155-13110155 on Build GRCh38
rs576534250 Chr.20: 13110421-13110421 on Build GRCh38
rs745357595 Chr.20: 13110305-13110305 on Build GRCh38
rs386812643 Chr.20: 13110097-13110098 on Build GRCh38
rs552098864 Chr.20: 13110147-13110147 on Build GRCh38
rs372366943 Chr.20: 13110267-13110267 on Build GRCh38
rs749343340 Chr.20: 13110090-13110090 on Build GRCh38
rs776581063 Chr.20: 13110470-13110470 on Build GRCh38
rs759053565 Chr.20: 13110202-13110202 on Build GRCh38
rs760364228 Chr.20: 13110107-13110107 on Build GRCh38
rs761133600 Chr.20: 13110198-13110198 on Build GRCh38
rs951688068 Chr.20: 13110410-13110410 on Build GRCh38
rs780292355 Chr.20: 13110231-13110231 on Build GRCh38
rs751345059 Chr.20: 13110216-13110216 on Build GRCh38
rs536775172 Chr.20: 13110215-13110215 on Build GRCh38
rs746044374 Chr.20: 13110072-13110072 on Build GRCh38
rs200466273 Chr.20: 13110086-13110086 on Build GRCh38
rs186355763 Chr.20: 13110250-13110250 on Build GRCh38
rs1052737000 Chr.20: 13110259-13110259 on Build GRCh38
rs369662555 Chr.20: 13110262-13110262 on Build GRCh38
rs983035893 Chr.20: 13110426-13110426 on Build GRCh38
rs770690336 Chr.20: 13110201-13110201 on Build GRCh38
rs944222762 Chr.20: 13110434-13110434 on Build GRCh38
rs558773880 Chr.20: 13110343-13110343 on Build GRCh38
rs534288493 Chr.20: 13110375-13110375 on Build GRCh38
rs142788845 Chr.20: 13110353-13110353 on Build GRCh38
rs964825458 Chr.20: 13110277-13110277 on Build GRCh38
rs1000979457 Chr.20: 13110302-13110302 on Build GRCh38
rs776295961 Chr.20: 13110071-13110071 on Build GRCh38
rs762813556 Chr.20: 13110213-13110213 on Build GRCh38
rs752614160 Chr.20: 13110143-13110143 on Build GRCh38
rs974792686 Chr.20: 13110298-13110298 on Build GRCh38
rs2236126 Chr.20: 13110098-13110098 on Build GRCh38
rs772735010 Chr.20: 13110194-13110194 on Build GRCh38
rs899490596 Chr.20: 13110295-13110295 on Build GRCh38
rs748553288 Chr.20: 13110192-13110192 on Build GRCh38
rs182994919 Chr.20: 13110404-13110404 on Build GRCh38
rs771826224 Chr.20: 13110345-13110345 on Build GRCh38
rs766132581 Chr.20: 13110125-13110125 on Build GRCh38
rs768285608 Chr.20: 13110444-13110444 on Build GRCh38
rs757111865 Chr.20: 13110219-13110219 on Build GRCh38

View Full Product Details