Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17226025-17226514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCSK2
Entrez Gene ID
5126
Gene Name
proprotein convertase subtilisin/kexin type 2
Gene Aliases
NEC 2, NEC-2, NEC2, PC2, SPC2
UniGene
Hs.315186
Gene Chromosome Location
Chr.20: 17226107-17484578 on Build GRCh38
Transcripts
NM_001201528.1

SNP Detail

SNP ID Chromosome Location
rs142608132 Chr.20: 17226430-17226430 on Build GRCh38
rs933581960 Chr.20: 17226383-17226383 on Build GRCh38
rs934680175 Chr.20: 17226255-17226255 on Build GRCh38
rs768042635 Chr.20: 17226217-17226217 on Build GRCh38
rs983904660 Chr.20: 17226270-17226270 on Build GRCh38
rs987656285 Chr.20: 17226384-17226384 on Build GRCh38
rs73256197 Chr.20: 17226428-17226428 on Build GRCh38
rs961147180 Chr.20: 17226302-17226302 on Build GRCh38
rs895772770 Chr.20: 17226185-17226185 on Build GRCh38
rs1028457876 Chr.20: 17226485-17226485 on Build GRCh38
rs910741262 Chr.20: 17226397-17226397 on Build GRCh38
rs538109210 Chr.20: 17226171-17226171 on Build GRCh38
rs376646084 Chr.20: 17226356-17226356 on Build GRCh38
rs1031667701 Chr.20: 17226182-17226182 on Build GRCh38
rs905059622 Chr.20: 17226292-17226292 on Build GRCh38
rs1015870989 Chr.20: 17226156-17226156 on Build GRCh38
rs1002402040 Chr.20: 17226055-17226055 on Build GRCh38
rs888087569 Chr.20: 17226401-17226401 on Build GRCh38
rs147772251 Chr.20: 17226267-17226267 on Build GRCh38
rs118031845 Chr.20: 17226056-17226056 on Build GRCh38
rs1046226411 Chr.20: 17226276-17226276 on Build GRCh38
rs376127651 Chr.20: 17226181-17226181 on Build GRCh38
rs925917133 Chr.20: 17226231-17226235 on Build GRCh38
rs952363859 Chr.20: 17226099-17226099 on Build GRCh38
rs1004427167 Chr.20: 17226100-17226100 on Build GRCh38
rs998880571 Chr.20: 17226438-17226438 on Build GRCh38
rs13037451 Chr.20: 17226325-17226325 on Build GRCh38
rs564319589 Chr.20: 17226453-17226453 on Build GRCh38
rs199826195 Chr.20: 17226355-17226356 on Build GRCh38
rs994659425 Chr.20: 17226261-17226261 on Build GRCh38
rs900307635 Chr.20: 17226161-17226161 on Build GRCh38
rs570437984 Chr.20: 17226140-17226140 on Build GRCh38
rs1052276778 Chr.20: 17226266-17226266 on Build GRCh38
rs965657046 Chr.20: 17226434-17226434 on Build GRCh38
rs1017116501 Chr.20: 17226295-17226295 on Build GRCh38
rs764716914 Chr.20: 17226169-17226169 on Build GRCh38
rs922232467 Chr.20: 17226340-17226340 on Build GRCh38
rs762399913 Chr.20: 17226179-17226179 on Build GRCh38
rs531832404 Chr.20: 17226463-17226463 on Build GRCh38
rs943482405 Chr.20: 17226120-17226120 on Build GRCh38
rs904638730 Chr.20: 17226442-17226442 on Build GRCh38
rs368197460 Chr.20: 17226250-17226250 on Build GRCh38
rs868492802 Chr.20: 17226200-17226200 on Build GRCh38
rs921473728 Chr.20: 17226186-17226186 on Build GRCh38

View Full Product Details