Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20035043-20035540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
UniGene
Hs.171342
Gene Chromosome Location
Chr.20: 20034361-20056046 on Build GRCh38
Transcripts
NM_001278625.1 NM_001278626.1 NM_001278627.1 NM_001278628.1 NM_016652.5

SNP Detail

SNP ID Chromosome Location
rs78877184 Chr.20: 20035327-20035327 on Build GRCh38
rs565376063 Chr.20: 20035171-20035171 on Build GRCh38
rs934903447 Chr.20: 20035084-20035084 on Build GRCh38
rs999385814 Chr.20: 20035284-20035284 on Build GRCh38
rs918632244 Chr.20: 20035452-20035452 on Build GRCh38
rs918568623 Chr.20: 20035134-20035134 on Build GRCh38
rs928876951 Chr.20: 20035096-20035096 on Build GRCh38
rs528716844 Chr.20: 20035168-20035168 on Build GRCh38
rs1056700742 Chr.20: 20035173-20035173 on Build GRCh38
rs566935125 Chr.20: 20035412-20035412 on Build GRCh38
rs796447486 Chr.20: 20035482-20035483 on Build GRCh38
rs955308624 Chr.20: 20035263-20035263 on Build GRCh38
rs970625237 Chr.20: 20035066-20035066 on Build GRCh38
rs548267952 Chr.20: 20035349-20035349 on Build GRCh38
rs565488911 Chr.20: 20035238-20035238 on Build GRCh38
rs980235067 Chr.20: 20035073-20035073 on Build GRCh38
rs776101486 Chr.20: 20035212-20035213 on Build GRCh38
rs969127715 Chr.20: 20035393-20035393 on Build GRCh38
rs753432900 Chr.20: 20035361-20035361 on Build GRCh38
rs999060492 Chr.20: 20035402-20035402 on Build GRCh38
rs996997634 Chr.20: 20035246-20035246 on Build GRCh38
rs187766794 Chr.20: 20035127-20035127 on Build GRCh38
rs75503129 Chr.20: 20035451-20035451 on Build GRCh38
rs1031832535 Chr.20: 20035502-20035502 on Build GRCh38
rs931436659 Chr.20: 20035170-20035170 on Build GRCh38
rs938781888 Chr.20: 20035390-20035390 on Build GRCh38
rs192741116 Chr.20: 20035395-20035395 on Build GRCh38
rs750722170 Chr.20: 20035442-20035442 on Build GRCh38
rs77985149 Chr.20: 20035219-20035219 on Build GRCh38
rs942764523 Chr.20: 20035493-20035493 on Build GRCh38
rs905025267 Chr.20: 20035131-20035131 on Build GRCh38
rs889659796 Chr.20: 20035235-20035235 on Build GRCh38
rs373746422 Chr.20: 20035249-20035249 on Build GRCh38
rs1004097668 Chr.20: 20035217-20035217 on Build GRCh38
rs13044732 Chr.20: 20035270-20035270 on Build GRCh38
rs372848212 Chr.20: 20035400-20035400 on Build GRCh38
rs778892600 Chr.20: 20035464-20035464 on Build GRCh38
rs111720765 Chr.20: 20035115-20035115 on Build GRCh38
rs34473409 Chr.20: 20035511-20035512 on Build GRCh38
rs1030649641 Chr.20: 20035329-20035329 on Build GRCh38
rs924792486 Chr.20: 20035283-20035284 on Build GRCh38
rs952814489 Chr.20: 20035465-20035465 on Build GRCh38
rs928695094 Chr.20: 20035382-20035382 on Build GRCh38
rs3186300 Chr.20: 20035485-20035485 on Build GRCh38
rs139689547 Chr.20: 20035211-20035212 on Build GRCh38
rs956249083 Chr.20: 20035293-20035293 on Build GRCh38
rs1044797079 Chr.20: 20035364-20035364 on Build GRCh38
rs984103268 Chr.20: 20035484-20035484 on Build GRCh38
rs533792996 Chr.20: 20035413-20035413 on Build GRCh38
rs551246970 Chr.20: 20035394-20035394 on Build GRCh38
rs1044512649 Chr.20: 20035126-20035126 on Build GRCh38
rs990431532 Chr.20: 20035337-20035337 on Build GRCh38
rs530078857 Chr.20: 20035399-20035399 on Build GRCh38
rs1023814578 Chr.20: 20035383-20035383 on Build GRCh38
rs867360055 Chr.20: 20035444-20035444 on Build GRCh38
rs761945075 Chr.20: 20035253-20035253 on Build GRCh38
rs201793234 Chr.20: 20035405-20035405 on Build GRCh38
rs1003341769 Chr.20: 20035139-20035140 on Build GRCh38
rs548733406 Chr.20: 20035411-20035411 on Build GRCh38
rs914560765 Chr.20: 20035349-20035349 on Build GRCh38
rs893461056 Chr.20: 20035358-20035358 on Build GRCh38
rs1054436501 Chr.20: 20035087-20035087 on Build GRCh38

View Full Product Details