Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21326201-21326714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRN2
Entrez Gene ID
22803
Gene Name
5'-3' exoribonuclease 2
Gene Aliases
-
UniGene
Hs.255932
Gene Chromosome Location
Chr.20: 21303284-21389825 on Build GRCh38
Transcripts
NM_001317960.1 NM_012255.4 XM_017027722.1 XM_017027723.1 XM_017027724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7240486 c.204-8C>G p.? Chr.20: 21326482-21326482 on Build GRCh38
1025088 c.116G>A p.S39N Chr.20: 21326319-21326319 on Build GRCh38
6824189 c.276A>G p.V92V Chr.20: 21326562-21326562 on Build GRCh38
1025089 c.200A>G p.D67G Chr.20: 21326403-21326403 on Build GRCh38
4741760 c.164T>C p.M55T Chr.20: 21326367-21326367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991023733 Chr.20: 21326235-21326235 on Build GRCh38
rs78434796 Chr.20: 21326589-21326589 on Build GRCh38
rs745512832 Chr.20: 21326382-21326382 on Build GRCh38
rs751898750 Chr.20: 21326639-21326639 on Build GRCh38
rs759752650 Chr.20: 21326454-21326454 on Build GRCh38
rs773357284 Chr.20: 21326242-21326242 on Build GRCh38
rs369080895 Chr.20: 21326257-21326257 on Build GRCh38
rs778233192 Chr.20: 21326636-21326636 on Build GRCh38
rs766050712 Chr.20: 21326626-21326626 on Build GRCh38
rs768422695 Chr.20: 21326590-21326590 on Build GRCh38
rs779863575 Chr.20: 21326486-21326486 on Build GRCh38
rs551757662 Chr.20: 21326468-21326468 on Build GRCh38
rs925951940 Chr.20: 21326684-21326684 on Build GRCh38
rs925954912 Chr.20: 21326410-21326410 on Build GRCh38
rs759714778 Chr.20: 21326265-21326265 on Build GRCh38
rs756791504 Chr.20: 21326326-21326326 on Build GRCh38
rs747378978 Chr.20: 21326635-21326635 on Build GRCh38
rs1008116389 Chr.20: 21326505-21326505 on Build GRCh38
rs769920395 Chr.20: 21326572-21326572 on Build GRCh38
rs537561401 Chr.20: 21326487-21326487 on Build GRCh38
rs113292985 Chr.20: 21326536-21326536 on Build GRCh38
rs534077630 Chr.20: 21326613-21326613 on Build GRCh38
rs370308584 Chr.20: 21326462-21326462 on Build GRCh38
rs775686209 Chr.20: 21326578-21326578 on Build GRCh38
rs780608516 Chr.20: 21326375-21326375 on Build GRCh38
rs765385322 Chr.20: 21326456-21326456 on Build GRCh38
rs773841644 Chr.20: 21326430-21326430 on Build GRCh38
rs745949862 Chr.20: 21326562-21326562 on Build GRCh38
rs775211380 Chr.20: 21326457-21326457 on Build GRCh38
rs774157429 Chr.20: 21326607-21326607 on Build GRCh38
rs374750927 Chr.20: 21326418-21326418 on Build GRCh38
rs751559463 Chr.20: 21326295-21326295 on Build GRCh38
rs749935157 Chr.20: 21326475-21326475 on Build GRCh38
rs374380499 Chr.20: 21326629-21326629 on Build GRCh38
rs778322865 Chr.20: 21326501-21326501 on Build GRCh38
rs759547235 Chr.20: 21326477-21326477 on Build GRCh38
rs760850691 Chr.20: 21326253-21326253 on Build GRCh38
rs146110428 Chr.20: 21326524-21326524 on Build GRCh38
rs758778363 Chr.20: 21326504-21326509 on Build GRCh38
rs189255798 Chr.20: 21326428-21326428 on Build GRCh38
rs753353121 Chr.20: 21326249-21326249 on Build GRCh38
rs752508946 Chr.20: 21326281-21326281 on Build GRCh38
rs200799558 Chr.20: 21326534-21326534 on Build GRCh38
rs950220040 Chr.20: 21326446-21326446 on Build GRCh38
rs764036284 Chr.20: 21326292-21326292 on Build GRCh38
rs373315919 Chr.20: 21326289-21326289 on Build GRCh38
rs776903908 Chr.20: 21326258-21326258 on Build GRCh38
rs781289560 Chr.20: 21326556-21326556 on Build GRCh38
rs752360814 Chr.20: 21326452-21326453 on Build GRCh38
rs767704118 Chr.20: 21326511-21326511 on Build GRCh38
rs377077576 Chr.20: 21326609-21326609 on Build GRCh38
rs2296728 Chr.20: 21326469-21326469 on Build GRCh38
rs899281284 Chr.20: 21326466-21326466 on Build GRCh38
rs781680127 Chr.20: 21326636-21326636 on Build GRCh38
rs776620358 Chr.20: 21326435-21326435 on Build GRCh38
rs754726948 Chr.20: 21326635-21326635 on Build GRCh38
rs754499506 Chr.20: 21326267-21326267 on Build GRCh38
rs760087097 Chr.20: 21326617-21326617 on Build GRCh38
rs200920750 Chr.20: 21326635-21326636 on Build GRCh38
rs778055599 Chr.20: 21326623-21326628 on Build GRCh38
rs765235941 Chr.20: 21326661-21326661 on Build GRCh38
rs547728016 Chr.20: 21326434-21326434 on Build GRCh38
rs779383651 Chr.20: 21326417-21326417 on Build GRCh38
rs202127993 Chr.20: 21326463-21326463 on Build GRCh38
rs949647991 Chr.20: 21326333-21326333 on Build GRCh38
rs368037588 Chr.20: 21326483-21326483 on Build GRCh38
rs765056802 Chr.20: 21326266-21326266 on Build GRCh38
rs147260116 Chr.20: 21326537-21326537 on Build GRCh38
rs376549093 Chr.20: 21326416-21326416 on Build GRCh38
rs371800917 Chr.20: 21326495-21326495 on Build GRCh38
rs764915313 Chr.20: 21326447-21326451 on Build GRCh38
rs755722673 Chr.20: 21326476-21326476 on Build GRCh38
rs940733192 Chr.20: 21326451-21326451 on Build GRCh38
rs376927991 Chr.20: 21326433-21326433 on Build GRCh38
rs1023825028 Chr.20: 21326677-21326677 on Build GRCh38

View Full Product Details