Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25612874-25613367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NANP
Entrez Gene ID
140838
Gene Name
N-acetylneuraminic acid phosphatase
Gene Aliases
C20orf147, HDHD4, dJ694B14.3
UniGene
Hs.143137
Gene Chromosome Location
Chr.20: 25612935-25624012 on Build GRCh38
Transcripts
NM_152667.2

SNP Detail

SNP ID Chromosome Location
rs148569433 Chr.20: 25613137-25613137 on Build GRCh38
rs1051012043 Chr.20: 25613227-25613227 on Build GRCh38
rs760195244 Chr.20: 25613148-25613149 on Build GRCh38
rs537906972 Chr.20: 25612943-25612943 on Build GRCh38
rs925756110 Chr.20: 25613145-25613145 on Build GRCh38
rs980000294 Chr.20: 25613133-25613133 on Build GRCh38
rs952784893 Chr.20: 25612941-25612941 on Build GRCh38
rs1044152678 Chr.20: 25612911-25612911 on Build GRCh38
rs992936404 Chr.20: 25613041-25613041 on Build GRCh38
rs1050974240 Chr.20: 25613020-25613020 on Build GRCh38
rs925689710 Chr.20: 25612933-25612933 on Build GRCh38
rs912915214 Chr.20: 25613175-25613175 on Build GRCh38
rs1028418487 Chr.20: 25612928-25612928 on Build GRCh38
rs1015570269 Chr.20: 25613242-25613242 on Build GRCh38
rs896491171 Chr.20: 25613325-25613325 on Build GRCh38
rs961634115 Chr.20: 25612997-25612997 on Build GRCh38
rs146649401 Chr.20: 25613307-25613307 on Build GRCh38
rs865863128 Chr.20: 25613023-25613023 on Build GRCh38
rs141431249 Chr.20: 25613345-25613345 on Build GRCh38
rs1007097902 Chr.20: 25613045-25613045 on Build GRCh38
rs544646039 Chr.20: 25613285-25613285 on Build GRCh38
rs992375687 Chr.20: 25613104-25613106 on Build GRCh38
rs6115228 Chr.20: 25613288-25613288 on Build GRCh38
rs367887833 Chr.20: 25613100-25613100 on Build GRCh38
rs916789215 Chr.20: 25613112-25613112 on Build GRCh38
rs998123551 Chr.20: 25612974-25612974 on Build GRCh38
rs917391158 Chr.20: 25613042-25613042 on Build GRCh38
rs142919517 Chr.20: 25613203-25613203 on Build GRCh38
rs1052818214 Chr.20: 25613094-25613094 on Build GRCh38
rs183916398 Chr.20: 25612908-25612908 on Build GRCh38
rs948901402 Chr.20: 25613047-25613047 on Build GRCh38
rs988556766 Chr.20: 25613170-25613170 on Build GRCh38
rs771541136 Chr.20: 25612952-25612955 on Build GRCh38
rs191266028 Chr.20: 25613077-25613077 on Build GRCh38
rs996029201 Chr.20: 25613194-25613194 on Build GRCh38
rs1025647725 Chr.20: 25613293-25613293 on Build GRCh38
rs567518468 Chr.20: 25613048-25613048 on Build GRCh38
rs984176615 Chr.20: 25612948-25612948 on Build GRCh38
rs186752242 Chr.20: 25613004-25613004 on Build GRCh38
rs898304522 Chr.20: 25613270-25613270 on Build GRCh38
rs935732468 Chr.20: 25613149-25613150 on Build GRCh38
rs900304768 Chr.20: 25613161-25613161 on Build GRCh38
rs781408343 Chr.20: 25612913-25612913 on Build GRCh38
rs12481037 Chr.20: 25613052-25613052 on Build GRCh38
rs574675024 Chr.20: 25613340-25613340 on Build GRCh38
rs879033898 Chr.20: 25613058-25613058 on Build GRCh38
rs942576675 Chr.20: 25613241-25613241 on Build GRCh38
rs763498485 Chr.20: 25613308-25613308 on Build GRCh38
rs908776846 Chr.20: 25612982-25612982 on Build GRCh38
rs1008796232 Chr.20: 25613113-25613113 on Build GRCh38
rs960665075 Chr.20: 25613097-25613097 on Build GRCh38
rs952858386 Chr.20: 25613212-25613212 on Build GRCh38
rs242131 Chr.20: 25613235-25613235 on Build GRCh38

View Full Product Details