Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31819264-31819788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLK2
Entrez Gene ID
85366
Gene Name
myosin light chain kinase 2
Gene Aliases
KMLC, MLCK, MLCK2, skMLCK
UniGene
Hs.86092
Gene Chromosome Location
Chr.20: 31819375-31834697 on Build GRCh38
Transcripts
NM_033118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7378414 c.4G>A p.A2T Chr.20: 31819584-31819584 on Build GRCh38
4136807 c.38A>C p.Q13P Chr.20: 31819618-31819618 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113279119 Chr.20: 31819429-31819429 on Build GRCh38
rs1056552527 Chr.20: 31819386-31819386 on Build GRCh38
rs113893002 Chr.20: 31819529-31819529 on Build GRCh38
rs974287692 Chr.20: 31819400-31819400 on Build GRCh38
rs773366735 Chr.20: 31819557-31819557 on Build GRCh38
rs727503302 Chr.20: 31819596-31819596 on Build GRCh38
rs1053639936 Chr.20: 31819525-31819525 on Build GRCh38
rs1030904630 Chr.20: 31819464-31819464 on Build GRCh38
rs1887731 Chr.20: 31819410-31819410 on Build GRCh38
rs541898459 Chr.20: 31819385-31819385 on Build GRCh38
rs1047303973 Chr.20: 31819328-31819328 on Build GRCh38
rs569261012 Chr.20: 31819700-31819700 on Build GRCh38
rs777578125 Chr.20: 31819645-31819645 on Build GRCh38
rs187513506 Chr.20: 31819295-31819295 on Build GRCh38
rs551005458 Chr.20: 31819674-31819674 on Build GRCh38
rs575357063 Chr.20: 31819536-31819536 on Build GRCh38
rs150707461 Chr.20: 31819505-31819505 on Build GRCh38
rs397517470 Chr.20: 31819568-31819568 on Build GRCh38
rs534288463 Chr.20: 31819318-31819318 on Build GRCh38
rs143463992 Chr.20: 31819567-31819567 on Build GRCh38
rs896209135 Chr.20: 31819399-31819399 on Build GRCh38
rs917591418 Chr.20: 31819758-31819758 on Build GRCh38
rs779225909 Chr.20: 31819656-31819656 on Build GRCh38
rs573216739 Chr.20: 31819305-31819305 on Build GRCh38
rs930134365 Chr.20: 31819440-31819440 on Build GRCh38
rs113684992 Chr.20: 31819539-31819539 on Build GRCh38
rs201983158 Chr.20: 31819586-31819586 on Build GRCh38
rs552527808 Chr.20: 31819329-31819329 on Build GRCh38
rs534202603 Chr.20: 31819749-31819749 on Build GRCh38
rs551580125 Chr.20: 31819746-31819746 on Build GRCh38
rs746589686 Chr.20: 31819665-31819665 on Build GRCh38
rs903685181 Chr.20: 31819418-31819418 on Build GRCh38
rs1044879859 Chr.20: 31819416-31819416 on Build GRCh38
rs987232167 Chr.20: 31819289-31819289 on Build GRCh38
rs1046822373 Chr.20: 31819441-31819441 on Build GRCh38
rs1045093501 Chr.20: 31819489-31819490 on Build GRCh38
rs566747747 Chr.20: 31819748-31819748 on Build GRCh38
rs556265822 Chr.20: 31819318-31819318 on Build GRCh38
rs183666766 Chr.20: 31819650-31819650 on Build GRCh38
rs940350124 Chr.20: 31819342-31819342 on Build GRCh38
rs986795179 Chr.20: 31819344-31819344 on Build GRCh38
rs545611943 Chr.20: 31819494-31819494 on Build GRCh38
rs554142647 Chr.20: 31819767-31819767 on Build GRCh38
rs949091366 Chr.20: 31819485-31819485 on Build GRCh38
rs1001140463 Chr.20: 31819459-31819459 on Build GRCh38
rs117502839 Chr.20: 31819584-31819584 on Build GRCh38
rs931522494 Chr.20: 31819324-31819324 on Build GRCh38
rs975522458 Chr.20: 31819309-31819309 on Build GRCh38
rs879798562 Chr.20: 31819304-31819304 on Build GRCh38
rs754853086 Chr.20: 31819601-31819601 on Build GRCh38
rs1056100654 Chr.20: 31819755-31819755 on Build GRCh38
rs956858345 Chr.20: 31819469-31819469 on Build GRCh38
rs964340998 Chr.20: 31819740-31819740 on Build GRCh38
rs920047085 Chr.20: 31819633-31819633 on Build GRCh38
rs983379812 Chr.20: 31819675-31819675 on Build GRCh38
rs753139766 Chr.20: 31819676-31819676 on Build GRCh38
rs917664217 Chr.20: 31819484-31819484 on Build GRCh38
rs142318926 Chr.20: 31819632-31819632 on Build GRCh38
rs920097834 Chr.20: 31819312-31819312 on Build GRCh38
rs574381754 Chr.20: 31819363-31819363 on Build GRCh38
rs964113223 Chr.20: 31819296-31819296 on Build GRCh38
rs373406874 Chr.20: 31819585-31819585 on Build GRCh38
rs192056427 Chr.20: 31819630-31819630 on Build GRCh38
rs779632451 Chr.20: 31819426-31819426 on Build GRCh38
rs539609319 Chr.20: 31819708-31819708 on Build GRCh38

View Full Product Details