Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32145171-32145665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM9SF4
Entrez Gene ID
9777
Gene Name
transmembrane 9 superfamily member 4
Gene Aliases
dJ836N17.2
UniGene
Hs.654665
Gene Chromosome Location
Chr.20: 32109506-32167258 on Build GRCh38
Transcripts
NM_014742.3 XM_005260622.4 XM_017028154.1 XM_017028155.1 XM_017028156.1 XM_017028157.1 XM_017028158.1 XM_017028159.1 XM_017028160.1 XM_017028161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7333360 c.819G>T p.M273I Chr.20: 32145359-32145359 on Build GRCh38
1288477 c.798C>A p.I266I Chr.20: 32145389-32145389 on Build GRCh38
3707691 c.772-5G>T p.? Chr.20: 32145307-32145307 on Build GRCh38
4291106 c.796C>T p.R266C Chr.20: 32145336-32145336 on Build GRCh38
3840714 c.720+1G>A p.? Chr.20: 32145210-32145210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901093954 Chr.20: 32145482-32145482 on Build GRCh38
rs746491372 Chr.20: 32145307-32145307 on Build GRCh38
rs754766880 Chr.20: 32145278-32145278 on Build GRCh38
rs961851621 Chr.20: 32145539-32145539 on Build GRCh38
rs745519189 Chr.20: 32145460-32145460 on Build GRCh38
rs189316771 Chr.20: 32145365-32145365 on Build GRCh38
rs1012317254 Chr.20: 32145446-32145446 on Build GRCh38
rs780237593 Chr.20: 32145276-32145276 on Build GRCh38
rs754049102 Chr.20: 32145413-32145413 on Build GRCh38
rs1037300581 Chr.20: 32145227-32145227 on Build GRCh38
rs1022428248 Chr.20: 32145456-32145456 on Build GRCh38
rs772678265 Chr.20: 32145465-32145465 on Build GRCh38
rs370898272 Chr.20: 32145216-32145216 on Build GRCh38
rs773708119 Chr.20: 32145475-32145475 on Build GRCh38
rs565782088 Chr.20: 32145563-32145563 on Build GRCh38
rs763378782 Chr.20: 32145222-32145222 on Build GRCh38
rs949037070 Chr.20: 32145240-32145240 on Build GRCh38
rs778775933 Chr.20: 32145279-32145279 on Build GRCh38
rs771599565 Chr.20: 32145202-32145202 on Build GRCh38
rs760718030 Chr.20: 32145399-32145399 on Build GRCh38
rs767642400 Chr.20: 32145238-32145238 on Build GRCh38
rs6121363 Chr.20: 32145545-32145545 on Build GRCh38
rs374554961 Chr.20: 32145235-32145235 on Build GRCh38
rs965967062 Chr.20: 32145518-32145518 on Build GRCh38
rs753677792 Chr.20: 32145277-32145277 on Build GRCh38
rs777818970 Chr.20: 32145434-32145434 on Build GRCh38
rs542425078 Chr.20: 32145443-32145443 on Build GRCh38
rs541011990 Chr.20: 32145289-32145289 on Build GRCh38
rs373781383 Chr.20: 32145431-32145431 on Build GRCh38
rs371250976 Chr.20: 32145430-32145430 on Build GRCh38
rs908931259 Chr.20: 32145593-32145593 on Build GRCh38
rs746585133 Chr.20: 32145439-32145439 on Build GRCh38
rs148713328 Chr.20: 32145398-32145398 on Build GRCh38
rs770655350 Chr.20: 32145220-32145220 on Build GRCh38
rs6121362 Chr.20: 32145514-32145514 on Build GRCh38
rs776417265 Chr.20: 32145357-32145357 on Build GRCh38
rs967899117 Chr.20: 32145199-32145199 on Build GRCh38
rs1048826979 Chr.20: 32145300-32145300 on Build GRCh38
rs770747344 Chr.20: 32145334-32145334 on Build GRCh38
rs749726953 Chr.20: 32145362-32145362 on Build GRCh38
rs984632817 Chr.20: 32145527-32145527 on Build GRCh38
rs910050393 Chr.20: 32145305-32145305 on Build GRCh38
rs972857746 Chr.20: 32145541-32145541 on Build GRCh38
rs367614289 Chr.20: 32145302-32145302 on Build GRCh38
rs532171470 Chr.20: 32145544-32145544 on Build GRCh38
rs764575877 Chr.20: 32145228-32145228 on Build GRCh38
rs931705642 Chr.20: 32145285-32145285 on Build GRCh38
rs199660977 Chr.20: 32145255-32145255 on Build GRCh38
rs755099817 Chr.20: 32145418-32145418 on Build GRCh38
rs1022122565 Chr.20: 32145197-32145198 on Build GRCh38
rs117226364 Chr.20: 32145492-32145492 on Build GRCh38
rs763801526 Chr.20: 32145528-32145528 on Build GRCh38
rs774513801 Chr.20: 32145366-32145366 on Build GRCh38
rs950265893 Chr.20: 32145633-32145633 on Build GRCh38
rs142472310 Chr.20: 32145458-32145458 on Build GRCh38
rs535924187 Chr.20: 32145590-32145590 on Build GRCh38
rs767864256 Chr.20: 32145375-32145375 on Build GRCh38
rs762415505 Chr.20: 32145371-32145371 on Build GRCh38
rs1028730042 Chr.20: 32145493-32145493 on Build GRCh38
rs765109885 Chr.20: 32145419-32145419 on Build GRCh38
rs774803112 Chr.20: 32145464-32145464 on Build GRCh38
rs748953351 Chr.20: 32145206-32145206 on Build GRCh38
rs771276038 Chr.20: 32145459-32145459 on Build GRCh38
rs576865921 Chr.20: 32145231-32145231 on Build GRCh38
rs564762895 Chr.20: 32145536-32145536 on Build GRCh38
rs766658424 Chr.20: 32145408-32145408 on Build GRCh38
rs747683805 Chr.20: 32145196-32145196 on Build GRCh38
rs1020828053 Chr.20: 32145473-32145473 on Build GRCh38

View Full Product Details