Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32987368-32987897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUN5
Entrez Gene ID
140732
Gene Name
Sad1 and UNC84 domain containing 5
Gene Aliases
SPAG4L, TSARG4, dJ726C3.1
UniGene
Hs.375186
Gene Chromosome Location
Chr.20: 32983773-33004449 on Build GRCh38
Transcripts
NM_080675.3 XM_011528573.1 XM_011528574.1 XM_011528575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400221 c.677G>A p.W226* Chr.20: 32987712-32987712 on Build GRCh38
6793213 c.711C>T p.P237P Chr.20: 32987678-32987678 on Build GRCh38
6882162 c.710C>T p.P237L Chr.20: 32987679-32987679 on Build GRCh38
1411149 c.638C>T p.T213M Chr.20: 32987751-32987751 on Build GRCh38
107409 c.696G>A p.W232* Chr.20: 32987693-32987693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977329700 Chr.20: 32987545-32987545 on Build GRCh38
rs1013618811 Chr.20: 32987623-32987623 on Build GRCh38
rs368868700 Chr.20: 32987809-32987809 on Build GRCh38
rs930142984 Chr.20: 32987411-32987411 on Build GRCh38
rs767417262 Chr.20: 32987685-32987686 on Build GRCh38
rs758521577 Chr.20: 32987662-32987662 on Build GRCh38
rs995617281 Chr.20: 32987864-32987864 on Build GRCh38
rs898390986 Chr.20: 32987542-32987542 on Build GRCh38
rs903291955 Chr.20: 32987790-32987790 on Build GRCh38
rs374115193 Chr.20: 32987615-32987615 on Build GRCh38
rs912912419 Chr.20: 32987443-32987443 on Build GRCh38
rs893159452 Chr.20: 32987449-32987449 on Build GRCh38
rs202057899 Chr.20: 32987753-32987753 on Build GRCh38
rs754351766 Chr.20: 32987655-32987655 on Build GRCh38
rs201188412 Chr.20: 32987687-32987687 on Build GRCh38
rs114181968 Chr.20: 32987843-32987843 on Build GRCh38
rs749023318 Chr.20: 32987785-32987785 on Build GRCh38
rs144675342 Chr.20: 32987744-32987744 on Build GRCh38
rs1011539786 Chr.20: 32987498-32987498 on Build GRCh38
rs540597982 Chr.20: 32987750-32987750 on Build GRCh38
rs868667084 Chr.20: 32987546-32987546 on Build GRCh38
rs745991744 Chr.20: 32987779-32987779 on Build GRCh38
rs766058513 Chr.20: 32987626-32987626 on Build GRCh38
rs942476421 Chr.20: 32987483-32987483 on Build GRCh38
rs546317329 Chr.20: 32987605-32987605 on Build GRCh38
rs775762179 Chr.20: 32987651-32987651 on Build GRCh38
rs770773234 Chr.20: 32987611-32987611 on Build GRCh38
rs747050839 Chr.20: 32987671-32987671 on Build GRCh38
rs371039244 Chr.20: 32987652-32987652 on Build GRCh38
rs778128145 Chr.20: 32987668-32987668 on Build GRCh38
rs777080340 Chr.20: 32987729-32987729 on Build GRCh38
rs751811907 Chr.20: 32987760-32987760 on Build GRCh38
rs781209729 Chr.20: 32987680-32987680 on Build GRCh38
rs1006824998 Chr.20: 32987630-32987630 on Build GRCh38
rs763144762 Chr.20: 32987650-32987651 on Build GRCh38
rs575762673 Chr.20: 32987589-32987589 on Build GRCh38
rs768506775 Chr.20: 32987789-32987789 on Build GRCh38
rs918303058 Chr.20: 32987399-32987399 on Build GRCh38
rs947404749 Chr.20: 32987463-32987463 on Build GRCh38
rs199809874 Chr.20: 32987766-32987766 on Build GRCh38
rs999805574 Chr.20: 32987602-32987602 on Build GRCh38
rs137876184 Chr.20: 32987693-32987693 on Build GRCh38
rs768189397 Chr.20: 32987691-32987691 on Build GRCh38
rs753156524 Chr.20: 32987660-32987660 on Build GRCh38
rs762533196 Chr.20: 32987638-32987638 on Build GRCh38
rs974248506 Chr.20: 32987392-32987392 on Build GRCh38
rs933229480 Chr.20: 32987557-32987557 on Build GRCh38
rs368880440 Chr.20: 32987801-32987801 on Build GRCh38
rs745834124 Chr.20: 32987683-32987683 on Build GRCh38
rs753383146 Chr.20: 32987627-32987627 on Build GRCh38
rs760016112 Chr.20: 32987732-32987732 on Build GRCh38
rs995854143 Chr.20: 32987544-32987544 on Build GRCh38
rs79408847 Chr.20: 32987810-32987810 on Build GRCh38
rs751001803 Chr.20: 32987651-32987651 on Build GRCh38
rs34815261 Chr.20: 32987544-32987545 on Build GRCh38
rs912318167 Chr.20: 32987453-32987453 on Build GRCh38
rs903261719 Chr.20: 32987513-32987513 on Build GRCh38
rs757373537 Chr.20: 32987771-32987771 on Build GRCh38
rs761167616 Chr.20: 32987721-32987721 on Build GRCh38
rs769662035 Chr.20: 32987686-32987686 on Build GRCh38
rs760248766 Chr.20: 32987822-32987822 on Build GRCh38
rs914561562 Chr.20: 32987452-32987452 on Build GRCh38
rs535606221 Chr.20: 32987506-32987506 on Build GRCh38
rs767016137 Chr.20: 32987725-32987725 on Build GRCh38
rs763855078 Chr.20: 32987645-32987645 on Build GRCh38
rs557186131 Chr.20: 32987567-32987567 on Build GRCh38
rs942445417 Chr.20: 32987833-32987833 on Build GRCh38
rs1055749006 Chr.20: 32987448-32987448 on Build GRCh38
rs759347998 Chr.20: 32987614-32987614 on Build GRCh38
rs765618626 Chr.20: 32987658-32987658 on Build GRCh38
rs113041433 Chr.20: 32987860-32987860 on Build GRCh38
rs574570097 Chr.20: 32987475-32987475 on Build GRCh38
rs1006689298 Chr.20: 32987837-32987837 on Build GRCh38
rs988827560 Chr.20: 32987763-32987763 on Build GRCh38
rs776768027 Chr.20: 32987612-32987612 on Build GRCh38
rs924563795 Chr.20: 32987550-32987550 on Build GRCh38
rs368994571 Chr.20: 32987855-32987855 on Build GRCh38
rs890983081 Chr.20: 32987823-32987823 on Build GRCh38
rs144628893 Chr.20: 32987421-32987421 on Build GRCh38
rs142319289 Chr.20: 32987672-32987672 on Build GRCh38
rs566377806 Chr.20: 32987490-32987492 on Build GRCh38
rs866568042 Chr.20: 32987861-32987861 on Build GRCh38
rs951159609 Chr.20: 32987720-32987720 on Build GRCh38
rs749147888 Chr.20: 32987690-32987690 on Build GRCh38
rs914793562 Chr.20: 32987781-32987781 on Build GRCh38
rs769793129 Chr.20: 32987629-32987629 on Build GRCh38
rs1050864336 Chr.20: 32987451-32987451 on Build GRCh38
rs907502495 Chr.20: 32987416-32987416 on Build GRCh38
rs760380117 Chr.20: 32987431-32987431 on Build GRCh38
rs775245703 Chr.20: 32987624-32987624 on Build GRCh38
rs781446292 Chr.20: 32987773-32987773 on Build GRCh38
rs918364461 Chr.20: 32987715-32987715 on Build GRCh38
rs774107230 Chr.20: 32987714-32987714 on Build GRCh38
rs1045892224 Chr.20: 32987415-32987415 on Build GRCh38
rs562378771 Chr.20: 32987751-32987751 on Build GRCh38
rs780106231 Chr.20: 32987783-32987783 on Build GRCh38

View Full Product Details