Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33226810-33227318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA3
Entrez Gene ID
128861
Gene Name
BPI fold containing family A member 3
Gene Aliases
C20orf71, SPLUNC3
UniGene
Hs.360989
Gene Chromosome Location
Chr.20: 33217310-33227806 on Build GRCh38
Transcripts
NM_001042439.2 NM_178466.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4909659 c.661G>A p.V221M Chr.20: 33226969-33226969 on Build GRCh38
6300559 c.683T>C p.I228T Chr.20: 33226991-33226991 on Build GRCh38
2757347 c.622G>A p.V208I Chr.20: 33226930-33226930 on Build GRCh38
4386253 c.683T>A p.I228K Chr.20: 33226991-33226991 on Build GRCh38
5798145 c.634A>C p.I212L Chr.20: 33226942-33226942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963285493 Chr.20: 33227184-33227184 on Build GRCh38
rs555114886 Chr.20: 33227195-33227195 on Build GRCh38
rs554296975 Chr.20: 33227093-33227093 on Build GRCh38
rs761076464 Chr.20: 33226965-33226965 on Build GRCh38
rs965937595 Chr.20: 33227139-33227139 on Build GRCh38
rs986595889 Chr.20: 33226950-33226950 on Build GRCh38
rs750783675 Chr.20: 33226962-33226962 on Build GRCh38
rs768184513 Chr.20: 33226961-33226961 on Build GRCh38
rs913126242 Chr.20: 33226915-33226915 on Build GRCh38
rs573276663 Chr.20: 33227241-33227241 on Build GRCh38
rs912391364 Chr.20: 33226955-33226955 on Build GRCh38
rs766784394 Chr.20: 33226966-33226966 on Build GRCh38
rs986027229 Chr.20: 33226892-33226892 on Build GRCh38
rs984154942 Chr.20: 33227178-33227178 on Build GRCh38
rs374153892 Chr.20: 33226898-33226898 on Build GRCh38
rs758311579 Chr.20: 33226989-33226989 on Build GRCh38
rs750873527 Chr.20: 33226893-33226893 on Build GRCh38
rs750548402 Chr.20: 33227094-33227094 on Build GRCh38
rs778857381 Chr.20: 33226975-33226975 on Build GRCh38
rs199674478 Chr.20: 33227042-33227042 on Build GRCh38
rs185707742 Chr.20: 33227292-33227292 on Build GRCh38
rs750649586 Chr.20: 33227181-33227181 on Build GRCh38
rs544131759 Chr.20: 33227286-33227286 on Build GRCh38
rs949237600 Chr.20: 33227287-33227287 on Build GRCh38
rs150873854 Chr.20: 33226928-33226928 on Build GRCh38
rs887321765 Chr.20: 33227054-33227056 on Build GRCh38
rs539260827 Chr.20: 33227058-33227058 on Build GRCh38
rs529235189 Chr.20: 33227124-33227124 on Build GRCh38
rs769543822 Chr.20: 33227045-33227045 on Build GRCh38
rs972397239 Chr.20: 33227188-33227188 on Build GRCh38
rs367651598 Chr.20: 33226973-33226973 on Build GRCh38
rs921232703 Chr.20: 33226863-33226863 on Build GRCh38
rs746980314 Chr.20: 33227020-33227020 on Build GRCh38
rs954014230 Chr.20: 33227177-33227177 on Build GRCh38
rs571938603 Chr.20: 33227050-33227050 on Build GRCh38
rs754117001 Chr.20: 33226909-33226909 on Build GRCh38
rs114904196 Chr.20: 33226957-33226957 on Build GRCh38
rs3746391 Chr.20: 33226832-33226832 on Build GRCh38
rs904846473 Chr.20: 33227090-33227090 on Build GRCh38
rs139539413 Chr.20: 33226971-33226971 on Build GRCh38
rs200838072 Chr.20: 33226956-33226956 on Build GRCh38
rs1034090887 Chr.20: 33227097-33227097 on Build GRCh38
rs540311960 Chr.20: 33227148-33227148 on Build GRCh38
rs905473227 Chr.20: 33227293-33227293 on Build GRCh38
rs562367753 Chr.20: 33226855-33226855 on Build GRCh38
rs909159729 Chr.20: 33226895-33226895 on Build GRCh38
rs770664290 Chr.20: 33227021-33227021 on Build GRCh38
rs777533373 Chr.20: 33226944-33226944 on Build GRCh38
rs957364278 Chr.20: 33227101-33227101 on Build GRCh38
rs372164313 Chr.20: 33226958-33226958 on Build GRCh38
rs566874681 Chr.20: 33226930-33226930 on Build GRCh38
rs747516837 Chr.20: 33226897-33226897 on Build GRCh38
rs756190141 Chr.20: 33227183-33227183 on Build GRCh38
rs767795072 Chr.20: 33227154-33227154 on Build GRCh38
rs1016553809 Chr.20: 33227069-33227069 on Build GRCh38
rs781044608 Chr.20: 33227038-33227038 on Build GRCh38
rs751686888 Chr.20: 33227121-33227121 on Build GRCh38
rs561813816 Chr.20: 33226873-33226873 on Build GRCh38
rs377261494 Chr.20: 33226839-33226839 on Build GRCh38
rs909118405 Chr.20: 33227179-33227179 on Build GRCh38
rs754968574 Chr.20: 33226918-33226918 on Build GRCh38
rs779066653 Chr.20: 33226920-33226920 on Build GRCh38
rs1028434310 Chr.20: 33227166-33227166 on Build GRCh38
rs527737570 Chr.20: 33226945-33226945 on Build GRCh38
rs374914391 Chr.20: 33226849-33226849 on Build GRCh38
rs1046664706 Chr.20: 33227078-33227078 on Build GRCh38
rs967288091 Chr.20: 33226929-33226929 on Build GRCh38
rs770541338 Chr.20: 33226952-33226952 on Build GRCh38
rs572862356 Chr.20: 33227134-33227134 on Build GRCh38
rs1034554249 Chr.20: 33226851-33226852 on Build GRCh38
rs908543262 Chr.20: 33227266-33227266 on Build GRCh38
rs777729789 Chr.20: 33226994-33226994 on Build GRCh38
rs878903315 Chr.20: 33227095-33227095 on Build GRCh38
rs767450805 Chr.20: 33227092-33227092 on Build GRCh38
rs1008924921 Chr.20: 33227187-33227205 on Build GRCh38
rs754679360 Chr.20: 33227268-33227268 on Build GRCh38
rs1021084382 Chr.20: 33227131-33227131 on Build GRCh38
rs201210326 Chr.20: 33226908-33226908 on Build GRCh38
rs894093282 Chr.20: 33227107-33227107 on Build GRCh38
rs752882546 Chr.20: 33226976-33226976 on Build GRCh38
rs79189206 Chr.20: 33227085-33227085 on Build GRCh38

View Full Product Details