Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34980638-34981132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH7B
Entrez Gene ID
57644
Gene Name
myosin heavy chain 7B
Gene Aliases
MHC14, MYH14
UniGene
Hs.414122
Gene Chromosome Location
Chr.20: 34955835-35002437 on Build GRCh38
Transcripts
NM_020884.4 XM_006723840.3 XM_011528941.2 XM_017027986.1 XM_017027987.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118986 c.653+2T>G p.? Chr.20: 34981062-34981062 on Build GRCh38
443693 c.642C>T p.S214S Chr.20: 34981049-34981049 on Build GRCh38
1026142 c.603C>T p.N201N Chr.20: 34980712-34980712 on Build GRCh38
7101366 c.563G>A p.R188H Chr.20: 34980672-34980672 on Build GRCh38
4501060 c.579C>T p.P193P Chr.20: 34980688-34980688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761996955 Chr.20: 34981055-34981055 on Build GRCh38
rs561892954 Chr.20: 34980805-34980805 on Build GRCh38
rs146237112 Chr.20: 34981057-34981057 on Build GRCh38
rs374369942 Chr.20: 34980669-34980669 on Build GRCh38
rs375891946 Chr.20: 34980703-34980703 on Build GRCh38
rs370367964 Chr.20: 34981035-34981035 on Build GRCh38
rs961974716 Chr.20: 34981012-34981012 on Build GRCh38
rs368402495 Chr.20: 34980670-34980670 on Build GRCh38
rs934667896 Chr.20: 34980784-34980784 on Build GRCh38
rs993511727 Chr.20: 34980969-34980969 on Build GRCh38
rs372689911 Chr.20: 34981039-34981039 on Build GRCh38
rs578240950 Chr.20: 34980661-34980661 on Build GRCh38
rs916104652 Chr.20: 34980732-34980732 on Build GRCh38
rs369238070 Chr.20: 34980779-34980779 on Build GRCh38
rs991287682 Chr.20: 34981105-34981105 on Build GRCh38
rs762231855 Chr.20: 34981061-34981061 on Build GRCh38
rs994651192 Chr.20: 34980840-34980840 on Build GRCh38
rs781169401 Chr.20: 34980721-34980721 on Build GRCh38
rs886149246 Chr.20: 34980877-34980877 on Build GRCh38
rs372129944 Chr.20: 34980685-34980685 on Build GRCh38
rs747955411 Chr.20: 34981018-34981018 on Build GRCh38
rs531618249 Chr.20: 34980948-34980948 on Build GRCh38
rs763409138 Chr.20: 34981044-34981044 on Build GRCh38
rs376032551 Chr.20: 34980950-34980950 on Build GRCh38
rs762719894 Chr.20: 34980672-34980672 on Build GRCh38
rs775310408 Chr.20: 34980671-34980671 on Build GRCh38
rs371539151 Chr.20: 34980713-34980713 on Build GRCh38
rs983054690 Chr.20: 34980684-34980684 on Build GRCh38
rs61745058 Chr.20: 34981040-34981040 on Build GRCh38
rs900360042 Chr.20: 34980838-34980838 on Build GRCh38
rs901584107 Chr.20: 34980878-34980878 on Build GRCh38
rs1038930635 Chr.20: 34980818-34980818 on Build GRCh38
rs750584225 Chr.20: 34981080-34981080 on Build GRCh38
rs754837766 Chr.20: 34981011-34981011 on Build GRCh38
rs760163994 Chr.20: 34980700-34980700 on Build GRCh38
rs756100723 Chr.20: 34980731-34980731 on Build GRCh38
rs944658703 Chr.20: 34980806-34980806 on Build GRCh38
rs778618201 Chr.20: 34981013-34981013 on Build GRCh38
rs768273339 Chr.20: 34980740-34980740 on Build GRCh38
rs961821610 Chr.20: 34980762-34980762 on Build GRCh38
rs779959313 Chr.20: 34980734-34980734 on Build GRCh38
rs1027379728 Chr.20: 34980859-34980859 on Build GRCh38
rs1004512240 Chr.20: 34980925-34980925 on Build GRCh38
rs760762022 Chr.20: 34981089-34981089 on Build GRCh38
rs369320738 Chr.20: 34980760-34980760 on Build GRCh38
rs551248268 Chr.20: 34981097-34981097 on Build GRCh38
rs535913318 Chr.20: 34980682-34980682 on Build GRCh38
rs376078249 Chr.20: 34980728-34980728 on Build GRCh38
rs73905025 Chr.20: 34980773-34980773 on Build GRCh38
rs41305791 Chr.20: 34981024-34981024 on Build GRCh38
rs772900325 Chr.20: 34980764-34980764 on Build GRCh38
rs978929874 Chr.20: 34981071-34981071 on Build GRCh38
rs757388868 Chr.20: 34980717-34980717 on Build GRCh38
rs377012586 Chr.20: 34980785-34980785 on Build GRCh38
rs779975757 Chr.20: 34980997-34980997 on Build GRCh38
rs996355645 Chr.20: 34980884-34980884 on Build GRCh38
rs767765080 Chr.20: 34980988-34980988 on Build GRCh38
rs753888398 Chr.20: 34981111-34981111 on Build GRCh38
rs200034113 Chr.20: 34980995-34980995 on Build GRCh38
rs61745059 Chr.20: 34981041-34981041 on Build GRCh38
rs750565919 Chr.20: 34980992-34980992 on Build GRCh38
rs767727538 Chr.20: 34981069-34981069 on Build GRCh38
rs376965277 Chr.20: 34981020-34981020 on Build GRCh38
rs960359571 Chr.20: 34980954-34980954 on Build GRCh38
rs1015800857 Chr.20: 34980705-34980705 on Build GRCh38
rs545637890 Chr.20: 34980712-34980712 on Build GRCh38
rs540069100 Chr.20: 34980804-34980804 on Build GRCh38
rs902774726 Chr.20: 34980839-34980839 on Build GRCh38
rs776074381 Chr.20: 34980776-34980776 on Build GRCh38
rs757469986 Chr.20: 34980987-34980987 on Build GRCh38
rs200984501 Chr.20: 34980744-34980744 on Build GRCh38
rs777229146 Chr.20: 34980691-34980691 on Build GRCh38
rs367559910 Chr.20: 34980711-34980711 on Build GRCh38
rs764387751 Chr.20: 34980812-34980812 on Build GRCh38
rs753171719 Chr.20: 34980708-34980708 on Build GRCh38
rs371877031 Chr.20: 34980931-34980931 on Build GRCh38
rs111960403 Chr.20: 34980792-34980792 on Build GRCh38
rs981244125 Chr.20: 34980763-34980763 on Build GRCh38
rs770508327 Chr.20: 34981036-34981036 on Build GRCh38
rs749083292 Chr.20: 34980739-34980739 on Build GRCh38
rs749171130 Chr.20: 34980998-34980998 on Build GRCh38

View Full Product Details