Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35547205-35547718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
UniGene
Hs.472558
Gene Chromosome Location
Chr.20: 35542029-35557634 on Build GRCh38
Transcripts
NM_015966.2 NM_198398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6656103 c.461+7_461+8insG p.? Chr.20: 35547512-35547513 on Build GRCh38
1326782 c.422G>A p.R141H Chr.20: 35547466-35547466 on Build GRCh38
6583933 c.421C>T p.R141C Chr.20: 35547465-35547465 on Build GRCh38
6656105 c.461+7_461+8insG p.? Chr.20: 35547512-35547513 on Build GRCh38
5391926 c.454G>A p.D152N Chr.20: 35547498-35547498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540277990 Chr.20: 35547449-35547449 on Build GRCh38
rs142274713 Chr.20: 35547502-35547502 on Build GRCh38
rs200810973 Chr.20: 35547433-35547433 on Build GRCh38
rs200450712 Chr.20: 35547414-35547414 on Build GRCh38
rs528772096 Chr.20: 35547279-35547279 on Build GRCh38
rs1001431762 Chr.20: 35547324-35547324 on Build GRCh38
rs768853849 Chr.20: 35547488-35547488 on Build GRCh38
rs745660138 Chr.20: 35547250-35547250 on Build GRCh38
rs1040635297 Chr.20: 35547545-35547545 on Build GRCh38
rs781630771 Chr.20: 35547498-35547498 on Build GRCh38
rs975003695 Chr.20: 35547454-35547454 on Build GRCh38
rs373013568 Chr.20: 35547398-35547398 on Build GRCh38
rs34848077 Chr.20: 35547453-35547454 on Build GRCh38
rs201208533 Chr.20: 35547372-35547372 on Build GRCh38
rs376615939 Chr.20: 35547400-35547400 on Build GRCh38
rs761760973 Chr.20: 35547519-35547519 on Build GRCh38
rs770371796 Chr.20: 35547417-35547417 on Build GRCh38
rs757261785 Chr.20: 35547452-35547452 on Build GRCh38
rs527683953 Chr.20: 35547592-35547592 on Build GRCh38
rs755233679 Chr.20: 35547552-35547552 on Build GRCh38
rs1008004647 Chr.20: 35547575-35547575 on Build GRCh38
rs1049190148 Chr.20: 35547249-35547249 on Build GRCh38
rs865884255 Chr.20: 35547443-35547443 on Build GRCh38
rs578122081 Chr.20: 35547256-35547256 on Build GRCh38
rs556373726 Chr.20: 35547251-35547251 on Build GRCh38
rs765247551 Chr.20: 35547520-35547520 on Build GRCh38
rs181432942 Chr.20: 35547637-35547637 on Build GRCh38
rs984362405 Chr.20: 35547310-35547310 on Build GRCh38
rs1005278748 Chr.20: 35547624-35547624 on Build GRCh38
rs148786229 Chr.20: 35547435-35547435 on Build GRCh38
rs907029807 Chr.20: 35547291-35547291 on Build GRCh38
rs201814399 Chr.20: 35547434-35547434 on Build GRCh38
rs567406642 Chr.20: 35547633-35547633 on Build GRCh38
rs753656202 Chr.20: 35547448-35547448 on Build GRCh38
rs769108807 Chr.20: 35547404-35547404 on Build GRCh38
rs772422322 Chr.20: 35547376-35547376 on Build GRCh38
rs775104538 Chr.20: 35547512-35547513 on Build GRCh38
rs966217575 Chr.20: 35547579-35547580 on Build GRCh38
rs766502291 Chr.20: 35547544-35547544 on Build GRCh38
rs1014161504 Chr.20: 35547660-35547660 on Build GRCh38
rs958891745 Chr.20: 35547397-35547397 on Build GRCh38
rs957048298 Chr.20: 35547343-35547343 on Build GRCh38
rs958552968 Chr.20: 35547644-35547644 on Build GRCh38
rs781337665 Chr.20: 35547556-35547556 on Build GRCh38
rs763967570 Chr.20: 35547436-35547436 on Build GRCh38
rs201714249 Chr.20: 35547523-35547523 on Build GRCh38
rs1046724412 Chr.20: 35547620-35547620 on Build GRCh38
rs1034217951 Chr.20: 35547336-35547336 on Build GRCh38
rs779058707 Chr.20: 35547453-35547453 on Build GRCh38
rs767132794 Chr.20: 35547428-35547428 on Build GRCh38
rs187103039 Chr.20: 35547514-35547514 on Build GRCh38
rs199597735 Chr.20: 35547423-35547423 on Build GRCh38
rs111571893 Chr.20: 35547410-35547410 on Build GRCh38
rs561751402 Chr.20: 35547538-35547538 on Build GRCh38
rs770269912 Chr.20: 35547511-35547511 on Build GRCh38
rs762721281 Chr.20: 35547552-35547555 on Build GRCh38
rs745350676 Chr.20: 35547513-35547513 on Build GRCh38
rs900872550 Chr.20: 35547567-35547567 on Build GRCh38
rs1049055100 Chr.20: 35547508-35547508 on Build GRCh38
rs967231266 Chr.20: 35547673-35547673 on Build GRCh38
rs952309943 Chr.20: 35547619-35547619 on Build GRCh38
rs1024175993 Chr.20: 35547663-35547663 on Build GRCh38
rs377327385 Chr.20: 35547426-35547426 on Build GRCh38
rs750520078 Chr.20: 35547464-35547464 on Build GRCh38
rs224414 Chr.20: 35547515-35547515 on Build GRCh38
rs113834091 Chr.20: 35547692-35547692 on Build GRCh38
rs145119222 Chr.20: 35547587-35547587 on Build GRCh38
rs967429188 Chr.20: 35547573-35547573 on Build GRCh38
rs199738260 Chr.20: 35547512-35547512 on Build GRCh38
rs369141560 Chr.20: 35547407-35547407 on Build GRCh38
rs1019988227 Chr.20: 35547551-35547552 on Build GRCh38
rs758603835 Chr.20: 35547466-35547466 on Build GRCh38
rs868448649 Chr.20: 35547465-35547465 on Build GRCh38
rs986952530 Chr.20: 35547416-35547416 on Build GRCh38
rs79636727 Chr.20: 35547623-35547623 on Build GRCh38
rs779144737 Chr.20: 35547363-35547363 on Build GRCh38
rs189890419 Chr.20: 35547550-35547550 on Build GRCh38
rs747291377 Chr.20: 35547479-35547479 on Build GRCh38

View Full Product Details